Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 21906073-21906612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA7L
Entrez Gene ID
55536
Gene Name
cell division cycle associated 7 like
Gene Aliases
JPO2, R1, RAM2
UniGene
Hs.744069
Gene Chromosome Location
Chr.7: 21900899-21945924 on Build GRCh38
Transcripts
NM_001127370.2 NM_001127371.2 NM_018719.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5472392 c.775G>A p.A259T Chr.7: 21906435-21906435 on Build GRCh38
243587 c.731T>C p.V244A Chr.7: 21906590-21906590 on Build GRCh38
5528429 c.724T>C p.F242L Chr.7: 21906597-21906597 on Build GRCh38
7197043 c.882G>A p.A294A Chr.7: 21906328-21906328 on Build GRCh38
3923385 c.833C>T p.P278L Chr.7: 21906377-21906377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567337409 Chr.7: 21906579-21906579 on Build GRCh38
rs775452510 Chr.7: 21906490-21906490 on Build GRCh38
rs6958820 Chr.7: 21906340-21906340 on Build GRCh38
rs990709683 Chr.7: 21906478-21906478 on Build GRCh38
rs763009889 Chr.7: 21906548-21906548 on Build GRCh38
rs764612197 Chr.7: 21906429-21906429 on Build GRCh38
rs980189511 Chr.7: 21906103-21906103 on Build GRCh38
rs866565497 Chr.7: 21906233-21906233 on Build GRCh38
rs755505442 Chr.7: 21906313-21906313 on Build GRCh38
rs531578692 Chr.7: 21906510-21906510 on Build GRCh38
rs771925441 Chr.7: 21906530-21906530 on Build GRCh38
rs902890375 Chr.7: 21906102-21906102 on Build GRCh38
rs4445125 Chr.7: 21906222-21906222 on Build GRCh38
rs748785071 Chr.7: 21906466-21906466 on Build GRCh38
rs772970536 Chr.7: 21906471-21906474 on Build GRCh38
rs752101616 Chr.7: 21906496-21906496 on Build GRCh38
rs948414964 Chr.7: 21906396-21906396 on Build GRCh38
rs377604487 Chr.7: 21906413-21906413 on Build GRCh38
rs79856287 Chr.7: 21906454-21906454 on Build GRCh38
rs946317013 Chr.7: 21906210-21906210 on Build GRCh38
rs776219711 Chr.7: 21906257-21906257 on Build GRCh38
rs748591252 Chr.7: 21906492-21906493 on Build GRCh38
rs372198777 Chr.7: 21906227-21906227 on Build GRCh38
rs764460541 Chr.7: 21906137-21906137 on Build GRCh38
rs1023767944 Chr.7: 21906205-21906205 on Build GRCh38
rs369949371 Chr.7: 21906358-21906358 on Build GRCh38
rs762441667 Chr.7: 21906556-21906556 on Build GRCh38
rs1054241431 Chr.7: 21906151-21906151 on Build GRCh38
rs750600045 Chr.7: 21906298-21906298 on Build GRCh38
rs775082493 Chr.7: 21906407-21906407 on Build GRCh38
rs1046706851 Chr.7: 21906255-21906255 on Build GRCh38
rs1015204309 Chr.7: 21906424-21906424 on Build GRCh38
rs532195820 Chr.7: 21906157-21906157 on Build GRCh38
rs762865786 Chr.7: 21906341-21906341 on Build GRCh38
rs201719647 Chr.7: 21906415-21906415 on Build GRCh38
rs922366893 Chr.7: 21906437-21906437 on Build GRCh38
rs779113194 Chr.7: 21906481-21906482 on Build GRCh38
rs751659616 Chr.7: 21906155-21906155 on Build GRCh38
rs560390424 Chr.7: 21906443-21906443 on Build GRCh38
rs771809612 Chr.7: 21906254-21906254 on Build GRCh38
rs1005465017 Chr.7: 21906284-21906284 on Build GRCh38
rs372997607 Chr.7: 21906360-21906360 on Build GRCh38
rs760840017 Chr.7: 21906353-21906353 on Build GRCh38
rs540748332 Chr.7: 21906569-21906569 on Build GRCh38
rs772929155 Chr.7: 21906485-21906485 on Build GRCh38
rs375745418 Chr.7: 21906339-21906339 on Build GRCh38
rs780113859 Chr.7: 21906308-21906308 on Build GRCh38
rs549290765 Chr.7: 21906463-21906463 on Build GRCh38
rs770728898 Chr.7: 21906395-21906395 on Build GRCh38
rs779466315 Chr.7: 21906384-21906384 on Build GRCh38
rs186064203 Chr.7: 21906241-21906241 on Build GRCh38
rs548045503 Chr.7: 21906509-21906509 on Build GRCh38
rs779722757 Chr.7: 21906242-21906243 on Build GRCh38
rs200978833 Chr.7: 21906276-21906279 on Build GRCh38
rs534031047 Chr.7: 21906558-21906558 on Build GRCh38
rs145238615 Chr.7: 21906435-21906435 on Build GRCh38
rs146257728 Chr.7: 21906250-21906250 on Build GRCh38
rs765225320 Chr.7: 21906268-21906268 on Build GRCh38
rs200856074 Chr.7: 21906526-21906526 on Build GRCh38
rs1016216576 Chr.7: 21906421-21906421 on Build GRCh38
rs543951715 Chr.7: 21906285-21906285 on Build GRCh38
rs774230294 Chr.7: 21906551-21906551 on Build GRCh38
rs756302066 Chr.7: 21906304-21906304 on Build GRCh38
rs773019029 Chr.7: 21906539-21906539 on Build GRCh38
rs536974510 Chr.7: 21906232-21906232 on Build GRCh38
rs757664469 Chr.7: 21906438-21906438 on Build GRCh38
rs759666563 Chr.7: 21906418-21906418 on Build GRCh38
rs769492584 Chr.7: 21906220-21906220 on Build GRCh38
rs533091128 Chr.7: 21906272-21906272 on Build GRCh38
rs777536863 Chr.7: 21906329-21906329 on Build GRCh38
rs1045499113 Chr.7: 21906408-21906408 on Build GRCh38
rs751561480 Chr.7: 21906447-21906449 on Build GRCh38
rs748086892 Chr.7: 21906529-21906529 on Build GRCh38
rs776311696 Chr.7: 21906338-21906338 on Build GRCh38
rs898050182 Chr.7: 21906432-21906432 on Build GRCh38
rs369531494 Chr.7: 21906309-21906309 on Build GRCh38
rs568379504 Chr.7: 21906104-21906105 on Build GRCh38
rs748897088 Chr.7: 21906337-21906338 on Build GRCh38
rs375239883 Chr.7: 21906400-21906400 on Build GRCh38
rs894185447 Chr.7: 21906164-21906164 on Build GRCh38
rs768599738 Chr.7: 21906411-21906411 on Build GRCh38
rs767998433 Chr.7: 21906499-21906499 on Build GRCh38
rs372949340 Chr.7: 21906256-21906256 on Build GRCh38
rs767849344 Chr.7: 21906297-21906297 on Build GRCh38
rs992830549 Chr.7: 21906150-21906150 on Build GRCh38
rs139249411 Chr.7: 21906393-21906393 on Build GRCh38
rs757782682 Chr.7: 21906498-21906498 on Build GRCh38
rs866666863 Chr.7: 21906301-21906301 on Build GRCh38
rs368726304 Chr.7: 21906517-21906517 on Build GRCh38
rs924511731 Chr.7: 21906355-21906355 on Build GRCh38
rs752753569 Chr.7: 21906427-21906427 on Build GRCh38
rs764549352 Chr.7: 21906495-21906495 on Build GRCh38
rs748792271 Chr.7: 21906518-21906518 on Build GRCh38
rs571302464 Chr.7: 21906115-21906115 on Build GRCh38
rs913499331 Chr.7: 21906191-21906191 on Build GRCh38
rs201013188 Chr.7: 21906494-21906494 on Build GRCh38
rs780969974 Chr.7: 21906398-21906398 on Build GRCh38
rs74997026 Chr.7: 21906559-21906559 on Build GRCh38
rs754532598 Chr.7: 21906582-21906582 on Build GRCh38
rs372703469 Chr.7: 21906540-21906540 on Build GRCh38
rs762805490 Chr.7: 21906410-21906410 on Build GRCh38
rs780423280 Chr.7: 21906406-21906406 on Build GRCh38
rs769958757 Chr.7: 21906542-21906542 on Build GRCh38
rs758104439 Chr.7: 21906293-21906293 on Build GRCh38
rs141349806 Chr.7: 21906572-21906572 on Build GRCh38
rs768767328 Chr.7: 21906549-21906549 on Build GRCh38
rs370506709 Chr.7: 21906385-21906385 on Build GRCh38
rs926134535 Chr.7: 21906223-21906223 on Build GRCh38
rs375069121 Chr.7: 21906267-21906267 on Build GRCh38
rs905028335 Chr.7: 21906323-21906323 on Build GRCh38
rs1041610699 Chr.7: 21906101-21906101 on Build GRCh38
rs748589038 Chr.7: 21906324-21906324 on Build GRCh38
rs34578779 Chr.7: 21906370-21906370 on Build GRCh38
rs746650414 Chr.7: 21906330-21906330 on Build GRCh38
rs778002499 Chr.7: 21906386-21906386 on Build GRCh38
rs763552835 Chr.7: 21906428-21906428 on Build GRCh38
rs771556634 Chr.7: 21906587-21906587 on Build GRCh38
rs768528504 Chr.7: 21906470-21906470 on Build GRCh38
rs200255145 Chr.7: 21906521-21906521 on Build GRCh38
rs774317138 Chr.7: 21906476-21906476 on Build GRCh38
rs202228816 Chr.7: 21906544-21906544 on Build GRCh38
rs113324234 Chr.7: 21906174-21906174 on Build GRCh38
rs373221505 Chr.7: 21906346-21906346 on Build GRCh38
rs1054503511 Chr.7: 21906359-21906359 on Build GRCh38
rs140167617 Chr.7: 21906317-21906317 on Build GRCh38
rs755854899 Chr.7: 21906506-21906506 on Build GRCh38
rs370910032 Chr.7: 21906484-21906484 on Build GRCh38
rs745529677 Chr.7: 21906449-21906449 on Build GRCh38
rs749671239 Chr.7: 21906416-21906416 on Build GRCh38
rs770546760 Chr.7: 21906332-21906332 on Build GRCh38
rs150016835 Chr.7: 21906412-21906412 on Build GRCh38
rs201108706 Chr.7: 21906328-21906328 on Build GRCh38

View Full Product Details