Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25122344-25122840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYCS
Entrez Gene ID
54205
Gene Name
cytochrome c, somatic
Gene Aliases
CYC, HCS, THC4
UniGene
Hs.437060
Gene Chromosome Location
Chr.7: 25118651-25125361 on Build GRCh38
Transcripts
NM_018947.5

SNP Detail

SNP ID Chromosome Location
rs764558730 Chr.7: 25122422-25122427 on Build GRCh38
rs886062251 Chr.7: 25122751-25122751 on Build GRCh38
rs999077390 Chr.7: 25122576-25122576 on Build GRCh38
rs1041618911 Chr.7: 25122594-25122594 on Build GRCh38
rs867624943 Chr.7: 25122447-25122447 on Build GRCh38
rs553177713 Chr.7: 25122422-25122422 on Build GRCh38
rs950583238 Chr.7: 25122723-25122723 on Build GRCh38
rs189863080 Chr.7: 25122686-25122686 on Build GRCh38
rs762751602 Chr.7: 25122765-25122765 on Build GRCh38
rs542092170 Chr.7: 25122582-25122582 on Build GRCh38
rs535074932 Chr.7: 25122584-25122584 on Build GRCh38
rs938344841 Chr.7: 25122640-25122640 on Build GRCh38
rs1055283792 Chr.7: 25122478-25122478 on Build GRCh38
rs543873365 Chr.7: 25122752-25122752 on Build GRCh38
rs767509843 Chr.7: 25122617-25122617 on Build GRCh38
rs775529379 Chr.7: 25122763-25122763 on Build GRCh38
rs377461599 Chr.7: 25122592-25122592 on Build GRCh38
rs976113239 Chr.7: 25122431-25122431 on Build GRCh38
rs141279662 Chr.7: 25122419-25122419 on Build GRCh38
rs540807000 Chr.7: 25122749-25122749 on Build GRCh38
rs937864159 Chr.7: 25122756-25122757 on Build GRCh38
rs75291633 Chr.7: 25122407-25122407 on Build GRCh38
rs759660368 Chr.7: 25122677-25122677 on Build GRCh38
rs1018139470 Chr.7: 25122663-25122663 on Build GRCh38
rs973835192 Chr.7: 25122714-25122714 on Build GRCh38
rs550435948 Chr.7: 25122391-25122391 on Build GRCh38
rs990574985 Chr.7: 25122758-25122758 on Build GRCh38
rs914403896 Chr.7: 25122587-25122587 on Build GRCh38
rs922036276 Chr.7: 25122451-25122451 on Build GRCh38
rs952318917 Chr.7: 25122394-25122394 on Build GRCh38
rs928270766 Chr.7: 25122620-25122620 on Build GRCh38
rs527848307 Chr.7: 25122745-25122765 on Build GRCh38
rs776820188 Chr.7: 25122645-25122645 on Build GRCh38
rs181671136 Chr.7: 25122793-25122793 on Build GRCh38
rs937707143 Chr.7: 25122466-25122466 on Build GRCh38
rs893467749 Chr.7: 25122486-25122486 on Build GRCh38
rs36061386 Chr.7: 25122454-25122455 on Build GRCh38
rs576345659 Chr.7: 25122547-25122547 on Build GRCh38
rs370941753 Chr.7: 25122621-25122621 on Build GRCh38
rs771207801 Chr.7: 25122537-25122537 on Build GRCh38
rs1005100887 Chr.7: 25122768-25122768 on Build GRCh38
rs953610891 Chr.7: 25122483-25122483 on Build GRCh38
rs577601640 Chr.7: 25122721-25122741 on Build GRCh38
rs927741574 Chr.7: 25122753-25122753 on Build GRCh38
rs574649600 Chr.7: 25122404-25122404 on Build GRCh38
rs752150593 Chr.7: 25122448-25122448 on Build GRCh38
rs1011020165 Chr.7: 25122519-25122519 on Build GRCh38
rs1015174171 Chr.7: 25122779-25122779 on Build GRCh38
rs762398655 Chr.7: 25122549-25122550 on Build GRCh38
rs989984737 Chr.7: 25122570-25122570 on Build GRCh38

View Full Product Details