Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25122933-25123434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYCS
Entrez Gene ID
54205
Gene Name
cytochrome c, somatic
Gene Aliases
CYC, HCS, THC4
UniGene
Hs.437060
Gene Chromosome Location
Chr.7: 25118651-25125361 on Build GRCh38
Transcripts
NM_018947.5

SNP Detail

SNP ID Chromosome Location
rs28376346 Chr.7: 25123300-25123300 on Build GRCh38
rs570409351 Chr.7: 25123354-25123354 on Build GRCh38
rs530407636 Chr.7: 25123288-25123288 on Build GRCh38
rs923606418 Chr.7: 25123265-25123265 on Build GRCh38
rs1026697759 Chr.7: 25122996-25122996 on Build GRCh38
rs561477883 Chr.7: 25123220-25123220 on Build GRCh38
rs755259625 Chr.7: 25123213-25123213 on Build GRCh38
rs11548773 Chr.7: 25123315-25123315 on Build GRCh38
rs556498765 Chr.7: 25123204-25123204 on Build GRCh38
rs532984462 Chr.7: 25123164-25123164 on Build GRCh38
rs549785908 Chr.7: 25123236-25123236 on Build GRCh38
rs959177076 Chr.7: 25123074-25123074 on Build GRCh38
rs758762399 Chr.7: 25123132-25123132 on Build GRCh38
rs779379080 Chr.7: 25123031-25123031 on Build GRCh38
rs569634735 Chr.7: 25123302-25123302 on Build GRCh38
rs991038892 Chr.7: 25123154-25123154 on Build GRCh38
rs778338655 Chr.7: 25123140-25123140 on Build GRCh38
rs1034966608 Chr.7: 25123066-25123066 on Build GRCh38
rs753258621 Chr.7: 25123093-25123093 on Build GRCh38
rs11548774 Chr.7: 25123279-25123279 on Build GRCh38
rs943651119 Chr.7: 25123303-25123303 on Build GRCh38
rs528696386 Chr.7: 25123040-25123040 on Build GRCh38
rs11548775 Chr.7: 25123104-25123104 on Build GRCh38
rs886062252 Chr.7: 25123316-25123316 on Build GRCh38
rs933692984 Chr.7: 25123283-25123283 on Build GRCh38
rs1039583395 Chr.7: 25123328-25123328 on Build GRCh38
rs755713305 Chr.7: 25123002-25123002 on Build GRCh38

View Full Product Details