Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156292596-156293089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMP
Entrez Gene ID
112770
Gene Name
glycosylated lysosomal membrane protein
Gene Aliases
C1orf85, NCU-G1
UniGene
Hs.202522
Gene Chromosome Location
Chr.1: 156292687-156295689 on Build GRCh38
Transcripts
NM_001256604.1 NM_001256605.1 NM_001256608.1 NM_001256609.1 NM_144580.2 XM_011509117.2 XM_017000149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5983756 c.1201G>A p.E401K Chr.1: 156293064-156293064 on Build GRCh38
897175 c.1196A>G p.Y399C Chr.1: 156293069-156293069 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770023558 Chr.1: 156292991-156292991 on Build GRCh38
rs563358502 Chr.1: 156292958-156292958 on Build GRCh38
rs755625851 Chr.1: 156292629-156292629 on Build GRCh38
rs767186212 Chr.1: 156293024-156293024 on Build GRCh38
rs771362805 Chr.1: 156292996-156292996 on Build GRCh38
rs1047479497 Chr.1: 156292942-156292942 on Build GRCh38
rs1019686948 Chr.1: 156292959-156292959 on Build GRCh38
rs113550295 Chr.1: 156292846-156292846 on Build GRCh38
rs745742776 Chr.1: 156292973-156292973 on Build GRCh38
rs749531839 Chr.1: 156292886-156292886 on Build GRCh38
rs188090098 Chr.1: 156293030-156293030 on Build GRCh38
rs1020223674 Chr.1: 156292723-156292723 on Build GRCh38
rs559787191 Chr.1: 156292906-156292906 on Build GRCh38
rs760313889 Chr.1: 156293038-156293038 on Build GRCh38
rs889676835 Chr.1: 156292648-156292648 on Build GRCh38
rs975669333 Chr.1: 156292807-156292807 on Build GRCh38
rs8179445 Chr.1: 156292875-156292875 on Build GRCh38
rs199685689 Chr.1: 156292992-156292992 on Build GRCh38
rs763361490 Chr.1: 156292993-156292993 on Build GRCh38
rs1032083914 Chr.1: 156292857-156292857 on Build GRCh38
rs746817877 Chr.1: 156292792-156292792 on Build GRCh38
rs6800 Chr.1: 156292862-156292862 on Build GRCh38
rs8179431 Chr.1: 156292759-156292759 on Build GRCh38
rs931313391 Chr.1: 156292938-156292938 on Build GRCh38
rs113301950 Chr.1: 156292737-156292737 on Build GRCh38
rs551488125 Chr.1: 156293037-156293037 on Build GRCh38
rs113365142 Chr.1: 156292918-156292918 on Build GRCh38
rs957702689 Chr.1: 156292828-156292828 on Build GRCh38
rs753376766 Chr.1: 156293046-156293046 on Build GRCh38
rs949133386 Chr.1: 156292905-156292905 on Build GRCh38
rs768750449 Chr.1: 156292977-156292977 on Build GRCh38
rs1025733403 Chr.1: 156292633-156292633 on Build GRCh38
rs762390739 Chr.1: 156292988-156292988 on Build GRCh38
rs757053697 Chr.1: 156293058-156293058 on Build GRCh38
rs574178123 Chr.1: 156292850-156292850 on Build GRCh38
rs775002655 Chr.1: 156293014-156293014 on Build GRCh38
rs1049747204 Chr.1: 156292647-156292647 on Build GRCh38
rs148390696 Chr.1: 156292816-156292816 on Build GRCh38
rs931414610 Chr.1: 156293026-156293026 on Build GRCh38
rs544979845 Chr.1: 156292922-156292922 on Build GRCh38
rs999802209 Chr.1: 156292844-156292844 on Build GRCh38
rs528453983 Chr.1: 156293006-156293006 on Build GRCh38
rs778644291 Chr.1: 156293062-156293062 on Build GRCh38
rs368297351 Chr.1: 156293039-156293039 on Build GRCh38
rs760072039 Chr.1: 156293016-156293016 on Build GRCh38
rs768784829 Chr.1: 156292914-156292914 on Build GRCh38
rs1008412149 Chr.1: 156292703-156292703 on Build GRCh38
rs1008158380 Chr.1: 156292708-156292708 on Build GRCh38
rs995459586 Chr.1: 156292644-156292644 on Build GRCh38
rs987843569 Chr.1: 156293043-156293043 on Build GRCh38
rs898624919 Chr.1: 156292631-156292631 on Build GRCh38
rs913665087 Chr.1: 156292871-156292871 on Build GRCh38
rs34859560 Chr.1: 156292784-156292785 on Build GRCh38
rs748924635 Chr.1: 156292845-156292845 on Build GRCh38
rs781196734 Chr.1: 156292980-156292980 on Build GRCh38
rs112378379 Chr.1: 156292694-156292694 on Build GRCh38
rs887095366 Chr.1: 156292945-156292945 on Build GRCh38
rs183594199 Chr.1: 156292668-156292668 on Build GRCh38
rs758325072 Chr.1: 156292625-156292625 on Build GRCh38
rs748020606 Chr.1: 156292968-156292968 on Build GRCh38
rs981726948 Chr.1: 156292933-156292933 on Build GRCh38
rs899045441 Chr.1: 156292923-156292923 on Build GRCh38

View Full Product Details