Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156651369-156651875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAN
Entrez Gene ID
63827
Gene Name
brevican
Gene Aliases
BEHAB, CSPG7
UniGene
Hs.516904
Gene Chromosome Location
Chr.1: 156641948-156659532 on Build GRCh38
Transcripts
NM_021948.4 NM_198427.1 XM_011509866.1 XM_017002047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291958 c.1282C>T p.P428S Chr.1: 156651674-156651674 on Build GRCh38
1184519 c.1289C>T p.T430M Chr.1: 156651681-156651681 on Build GRCh38
6252992 c.1253G>T p.S418I Chr.1: 156651645-156651645 on Build GRCh38
4023361 c.1294C>A p.L432I Chr.1: 156651686-156651686 on Build GRCh38
3677412 c.1239C>T p.D413D Chr.1: 156651631-156651631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762878238 Chr.1: 156651440-156651440 on Build GRCh38
rs751639825 Chr.1: 156651677-156651677 on Build GRCh38
rs114326306 Chr.1: 156651696-156651696 on Build GRCh38
rs750707107 Chr.1: 156651575-156651575 on Build GRCh38
rs751346420 Chr.1: 156651444-156651444 on Build GRCh38
rs747397325 Chr.1: 156651407-156651407 on Build GRCh38
rs752328093 Chr.1: 156651485-156651496 on Build GRCh38
rs769007869 Chr.1: 156651472-156651472 on Build GRCh38
rs776426277 Chr.1: 156651734-156651734 on Build GRCh38
rs373345126 Chr.1: 156651597-156651597 on Build GRCh38
rs979371238 Chr.1: 156651553-156651553 on Build GRCh38
rs747776850 Chr.1: 156651485-156651485 on Build GRCh38
rs144857663 Chr.1: 156651466-156651466 on Build GRCh38
rs369888202 Chr.1: 156651430-156651430 on Build GRCh38
rs550450698 Chr.1: 156651681-156651681 on Build GRCh38
rs369041752 Chr.1: 156651596-156651596 on Build GRCh38
rs186250850 Chr.1: 156651762-156651762 on Build GRCh38
rs753293174 Chr.1: 156651683-156651683 on Build GRCh38
rs769435743 Chr.1: 156651627-156651627 on Build GRCh38
rs775260998 Chr.1: 156651729-156651729 on Build GRCh38
rs754811469 Chr.1: 156651808-156651808 on Build GRCh38
rs766392051 Chr.1: 156651666-156651666 on Build GRCh38
rs770559603 Chr.1: 156651635-156651635 on Build GRCh38
rs535641022 Chr.1: 156651702-156651702 on Build GRCh38
rs776239118 Chr.1: 156651429-156651429 on Build GRCh38
rs949759031 Chr.1: 156651633-156651633 on Build GRCh38
rs149715480 Chr.1: 156651620-156651620 on Build GRCh38
rs766446589 Chr.1: 156651443-156651443 on Build GRCh38
rs896952046 Chr.1: 156651779-156651779 on Build GRCh38
rs757462459 Chr.1: 156651562-156651562 on Build GRCh38
rs146863260 Chr.1: 156651460-156651460 on Build GRCh38
rs778267376 Chr.1: 156651697-156651697 on Build GRCh38
rs775792628 Chr.1: 156651406-156651406 on Build GRCh38
rs375645530 Chr.1: 156651586-156651586 on Build GRCh38
rs12065791 Chr.1: 156651459-156651459 on Build GRCh38
rs150989682 Chr.1: 156651532-156651532 on Build GRCh38
rs778527596 Chr.1: 156651717-156651717 on Build GRCh38
rs1031936896 Chr.1: 156651530-156651530 on Build GRCh38
rs774061754 Chr.1: 156651639-156651639 on Build GRCh38
rs747559084 Chr.1: 156651470-156651470 on Build GRCh38
rs763222307 Chr.1: 156651665-156651665 on Build GRCh38
rs999163446 Chr.1: 156651517-156651517 on Build GRCh38
rs373539890 Chr.1: 156651565-156651565 on Build GRCh38
rs201470193 Chr.1: 156651445-156651445 on Build GRCh38
rs943616791 Chr.1: 156651537-156651537 on Build GRCh38
rs547992858 Chr.1: 156651725-156651725 on Build GRCh38
rs745395356 Chr.1: 156651724-156651724 on Build GRCh38
rs147928348 Chr.1: 156651494-156651494 on Build GRCh38
rs758445352 Chr.1: 156651578-156651578 on Build GRCh38
rs1026321093 Chr.1: 156651492-156651492 on Build GRCh38
rs776921204 Chr.1: 156651484-156651484 on Build GRCh38
rs1026436540 Chr.1: 156651822-156651822 on Build GRCh38
rs147038529 Chr.1: 156651631-156651631 on Build GRCh38
rs745966917 Chr.1: 156651461-156651461 on Build GRCh38
rs763994015 Chr.1: 156651662-156651662 on Build GRCh38
rs749792099 Chr.1: 156651791-156651791 on Build GRCh38
rs1050099621 Chr.1: 156651752-156651752 on Build GRCh38
rs1040830970 Chr.1: 156651849-156651849 on Build GRCh38
rs564111183 Chr.1: 156651648-156651648 on Build GRCh38
rs755436243 Chr.1: 156651680-156651680 on Build GRCh38
rs867087628 Chr.1: 156651549-156651549 on Build GRCh38
rs753513734 Chr.1: 156651733-156651733 on Build GRCh38
rs1036996446 Chr.1: 156651399-156651399 on Build GRCh38
rs144024281 Chr.1: 156651509-156651509 on Build GRCh38
rs758854745 Chr.1: 156651463-156651463 on Build GRCh38
rs1023335380 Chr.1: 156651410-156651410 on Build GRCh38
rs760736766 Chr.1: 156651730-156651730 on Build GRCh38
rs889675925 Chr.1: 156651764-156651764 on Build GRCh38
rs760366866 Chr.1: 156651657-156651657 on Build GRCh38
rs200156625 Chr.1: 156651716-156651716 on Build GRCh38
rs201572906 Chr.1: 156651519-156651519 on Build GRCh38
rs532039779 Chr.1: 156651663-156651663 on Build GRCh38
rs772735056 Chr.1: 156651503-156651503 on Build GRCh38
rs752440615 Chr.1: 156651561-156651561 on Build GRCh38
rs759487112 Chr.1: 156651644-156651644 on Build GRCh38
rs147658469 Chr.1: 156651645-156651645 on Build GRCh38
rs767460070 Chr.1: 156651450-156651450 on Build GRCh38
rs754084504 Chr.1: 156651458-156651458 on Build GRCh38
rs769063004 Chr.1: 156651426-156651426 on Build GRCh38
rs772609352 Chr.1: 156651438-156651438 on Build GRCh38
rs767405069 Chr.1: 156651543-156651543 on Build GRCh38
rs759611993 Chr.1: 156651533-156651533 on Build GRCh38
rs748988983 Chr.1: 156651632-156651632 on Build GRCh38

View Full Product Details