Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156652166-156652675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAN
Entrez Gene ID
63827
Gene Name
brevican
Gene Aliases
BEHAB, CSPG7
UniGene
Hs.516904
Gene Chromosome Location
Chr.1: 156641948-156659532 on Build GRCh38
Transcripts
NM_021948.4 NM_198427.1 XM_011509866.1 XM_017002047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1725772 c.1612C>A p.P538T Chr.1: 156652562-156652562 on Build GRCh38
5905365 c.1642G>A p.E548K Chr.1: 156652592-156652592 on Build GRCh38
5568573 c.1671C>T p.S557S Chr.1: 156652621-156652621 on Build GRCh38
4529783 c.1641G>A p.R547R Chr.1: 156652591-156652591 on Build GRCh38
367609 c.1684G>T p.A562S Chr.1: 156652634-156652634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038040706 Chr.1: 156652317-156652317 on Build GRCh38
rs368105991 Chr.1: 156652258-156652258 on Build GRCh38
rs767481662 Chr.1: 156652294-156652294 on Build GRCh38
rs772341344 Chr.1: 156652428-156652428 on Build GRCh38
rs62001920 Chr.1: 156652547-156652547 on Build GRCh38
rs767671462 Chr.1: 156652218-156652218 on Build GRCh38
rs759120648 Chr.1: 156652643-156652643 on Build GRCh38
rs773734256 Chr.1: 156652621-156652621 on Build GRCh38
rs757070229 Chr.1: 156652584-156652584 on Build GRCh38
rs780615400 Chr.1: 156652499-156652499 on Build GRCh38
rs764311510 Chr.1: 156652438-156652438 on Build GRCh38
rs918531600 Chr.1: 156652324-156652324 on Build GRCh38
rs139064894 Chr.1: 156652495-156652495 on Build GRCh38
rs371198979 Chr.1: 156652361-156652361 on Build GRCh38
rs373894207 Chr.1: 156652476-156652476 on Build GRCh38
rs761018793 Chr.1: 156652325-156652325 on Build GRCh38
rs62001919 Chr.1: 156652401-156652401 on Build GRCh38
rs770681285 Chr.1: 156652502-156652502 on Build GRCh38
rs371827559 Chr.1: 156652612-156652612 on Build GRCh38
rs760788101 Chr.1: 156652433-156652433 on Build GRCh38
rs765764239 Chr.1: 156652366-156652366 on Build GRCh38
rs143971433 Chr.1: 156652524-156652524 on Build GRCh38
rs759241251 Chr.1: 156652557-156652557 on Build GRCh38
rs753139424 Chr.1: 156652221-156652221 on Build GRCh38
rs199717638 Chr.1: 156652650-156652650 on Build GRCh38
rs759508386 Chr.1: 156652202-156652202 on Build GRCh38
rs917386160 Chr.1: 156652207-156652207 on Build GRCh38
rs1000595108 Chr.1: 156652196-156652196 on Build GRCh38
rs775390953 Chr.1: 156652429-156652429 on Build GRCh38
rs769622186 Chr.1: 156652279-156652279 on Build GRCh38
rs746563491 Chr.1: 156652402-156652402 on Build GRCh38
rs766954928 Chr.1: 156652559-156652559 on Build GRCh38
rs756058000 Chr.1: 156652492-156652492 on Build GRCh38
rs141542995 Chr.1: 156652416-156652416 on Build GRCh38
rs769007410 Chr.1: 156652601-156652601 on Build GRCh38
rs773047689 Chr.1: 156652419-156652419 on Build GRCh38
rs746362652 Chr.1: 156652417-156652417 on Build GRCh38
rs763578354 Chr.1: 156652652-156652652 on Build GRCh38
rs199791981 Chr.1: 156652278-156652278 on Build GRCh38
rs971560790 Chr.1: 156652507-156652507 on Build GRCh38
rs751586663 Chr.1: 156652201-156652201 on Build GRCh38
rs777787904 Chr.1: 156652573-156652573 on Build GRCh38
rs763766842 Chr.1: 156652352-156652354 on Build GRCh38
rs759869163 Chr.1: 156652285-156652285 on Build GRCh38
rs768646668 Chr.1: 156652273-156652273 on Build GRCh38
rs929878801 Chr.1: 156652623-156652623 on Build GRCh38
rs776866917 Chr.1: 156652532-156652532 on Build GRCh38
rs149016541 Chr.1: 156652439-156652439 on Build GRCh38
rs745556190 Chr.1: 156652265-156652265 on Build GRCh38
rs762347714 Chr.1: 156652542-156652542 on Build GRCh38
rs780742859 Chr.1: 156652475-156652475 on Build GRCh38
rs776368239 Chr.1: 156652277-156652277 on Build GRCh38
rs750807184 Chr.1: 156652455-156652455 on Build GRCh38
rs865822902 Chr.1: 156652283-156652283 on Build GRCh38
rs753760220 Chr.1: 156652581-156652581 on Build GRCh38
rs773402783 Chr.1: 156652551-156652551 on Build GRCh38
rs994354668 Chr.1: 156652609-156652609 on Build GRCh38
rs754820500 Chr.1: 156652403-156652403 on Build GRCh38
rs753437509 Chr.1: 156652618-156652618 on Build GRCh38
rs752208249 Chr.1: 156652566-156652566 on Build GRCh38
rs1053513615 Chr.1: 156652477-156652477 on Build GRCh38
rs770457358 Chr.1: 156652550-156652550 on Build GRCh38
rs775651862 Chr.1: 156652303-156652303 on Build GRCh38
rs748265275 Chr.1: 156652611-156652611 on Build GRCh38
rs774591218 Chr.1: 156652640-156652640 on Build GRCh38
rs756922777 Chr.1: 156652246-156652246 on Build GRCh38
rs765643575 Chr.1: 156652440-156652440 on Build GRCh38
rs964705440 Chr.1: 156652195-156652195 on Build GRCh38
rs774931549 Chr.1: 156652646-156652646 on Build GRCh38
rs763433784 Chr.1: 156652634-156652634 on Build GRCh38
rs201720135 Chr.1: 156652555-156652555 on Build GRCh38
rs758086676 Chr.1: 156652372-156652372 on Build GRCh38
rs780294314 Chr.1: 156652504-156652504 on Build GRCh38
rs764739576 Chr.1: 156652516-156652516 on Build GRCh38
rs749567822 Chr.1: 156652599-156652599 on Build GRCh38
rs377376223 Chr.1: 156652351-156652351 on Build GRCh38
rs760147033 Chr.1: 156652647-156652647 on Build GRCh38
rs143303018 Chr.1: 156652375-156652375 on Build GRCh38
rs374184277 Chr.1: 156652244-156652244 on Build GRCh38
rs564173291 Chr.1: 156652354-156652354 on Build GRCh38
rs370780755 Chr.1: 156652243-156652243 on Build GRCh38
rs756004391 Chr.1: 156652567-156652567 on Build GRCh38
rs1056695 Chr.1: 156652460-156652460 on Build GRCh38
rs377653408 Chr.1: 156652406-156652406 on Build GRCh38
rs201162583 Chr.1: 156652198-156652198 on Build GRCh38
rs762475998 Chr.1: 156652456-156652456 on Build GRCh38
rs369751363 Chr.1: 156652471-156652471 on Build GRCh38
rs553305305 Chr.1: 156652378-156652378 on Build GRCh38
rs750034985 Chr.1: 156652367-156652367 on Build GRCh38
rs367672577 Chr.1: 156652552-156652552 on Build GRCh38
rs781385090 Chr.1: 156652605-156652605 on Build GRCh38
rs979837618 Chr.1: 156652268-156652268 on Build GRCh38
rs941070358 Chr.1: 156652301-156652301 on Build GRCh38
rs766074504 Chr.1: 156652459-156652459 on Build GRCh38
rs956271175 Chr.1: 156652200-156652200 on Build GRCh38
rs771310974 Chr.1: 156652637-156652637 on Build GRCh38
rs767187921 Chr.1: 156652645-156652645 on Build GRCh38
rs747261515 Chr.1: 156652513-156652513 on Build GRCh38
rs761391980 Chr.1: 156652272-156652272 on Build GRCh38
rs747171917 Chr.1: 156652600-156652600 on Build GRCh38
rs200016771 Chr.1: 156652422-156652422 on Build GRCh38
rs756701335 Chr.1: 156652230-156652230 on Build GRCh38

View Full Product Details