Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157519651-157520151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL5
Entrez Gene ID
83416
Gene Name
Fc receptor like 5
Gene Aliases
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820
UniGene
Hs.415950
Gene Chromosome Location
Chr.1: 157513377-157552520 on Build GRCh38
Transcripts
NM_001195388.1 NM_031281.2 XM_011510030.1 XM_011510031.1 XM_011510032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1295258 c.2642C>G p.P881R Chr.1: 157519761-157519761 on Build GRCh38
3996980 c.2658C>T p.A886A Chr.1: 157519745-157519745 on Build GRCh38
4810754 c.2642C>G p.P881R Chr.1: 157519761-157519761 on Build GRCh38
3996979 c.2658C>T p.A886A Chr.1: 157519745-157519745 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs141227043 Chr.1: 157519948-157519948 on Build GRCh38
rs527511661 Chr.1: 157520029-157520029 on Build GRCh38
rs114097690 Chr.1: 157520096-157520096 on Build GRCh38
rs947856141 Chr.1: 157519996-157519996 on Build GRCh38
rs3827757 Chr.1: 157519745-157519745 on Build GRCh38
rs571186858 Chr.1: 157519864-157519864 on Build GRCh38
rs747005781 Chr.1: 157519772-157519772 on Build GRCh38
rs765376362 Chr.1: 157519827-157519827 on Build GRCh38
rs748652043 Chr.1: 157519791-157519791 on Build GRCh38
rs371825236 Chr.1: 157519958-157519958 on Build GRCh38
rs961577872 Chr.1: 157519842-157519842 on Build GRCh38
rs772975523 Chr.1: 157519722-157519722 on Build GRCh38
rs575358297 Chr.1: 157519954-157519954 on Build GRCh38
rs573201847 Chr.1: 157519983-157519983 on Build GRCh38
rs554800698 Chr.1: 157519982-157519982 on Build GRCh38
rs762687627 Chr.1: 157519814-157519814 on Build GRCh38
rs3811032 Chr.1: 157519698-157519698 on Build GRCh38
rs781781694 Chr.1: 157519784-157519784 on Build GRCh38
rs912746648 Chr.1: 157519884-157519884 on Build GRCh38
rs770322375 Chr.1: 157519794-157519794 on Build GRCh38
rs780208492 Chr.1: 157519770-157519770 on Build GRCh38
rs201681764 Chr.1: 157519742-157519742 on Build GRCh38
rs899812459 Chr.1: 157519953-157519953 on Build GRCh38
rs540500917 Chr.1: 157520018-157520018 on Build GRCh38
rs35288621 Chr.1: 157519949-157519949 on Build GRCh38
rs750983516 Chr.1: 157519734-157519734 on Build GRCh38
rs577922295 Chr.1: 157520073-157520073 on Build GRCh38
rs750701176 Chr.1: 157519753-157519753 on Build GRCh38
rs561879669 Chr.1: 157519826-157519826 on Build GRCh38
rs762481997 Chr.1: 157519724-157519724 on Build GRCh38
rs750656097 Chr.1: 157519793-157519793 on Build GRCh38
rs760724274 Chr.1: 157519819-157519819 on Build GRCh38
rs968439951 Chr.1: 157519937-157519937 on Build GRCh38
rs955413816 Chr.1: 157519869-157519869 on Build GRCh38
rs917677299 Chr.1: 157519973-157519973 on Build GRCh38
rs148190057 Chr.1: 157519859-157519859 on Build GRCh38
rs765692848 Chr.1: 157519720-157519720 on Build GRCh38
rs935166063 Chr.1: 157519959-157519959 on Build GRCh38
rs911249432 Chr.1: 157519904-157519904 on Build GRCh38
rs879189449 Chr.1: 157519918-157519918 on Build GRCh38
rs995472311 Chr.1: 157519956-157519956 on Build GRCh38
rs992507183 Chr.1: 157519728-157519728 on Build GRCh38
rs553115467 Chr.1: 157520057-157520057 on Build GRCh38
rs773408669 Chr.1: 157519795-157519795 on Build GRCh38
rs1056179585 Chr.1: 157519715-157519715 on Build GRCh38
rs368910462 Chr.1: 157519747-157519747 on Build GRCh38
rs972857629 Chr.1: 157519853-157519853 on Build GRCh38
rs571376086 Chr.1: 157519748-157519748 on Build GRCh38
rs769291867 Chr.1: 157519777-157519777 on Build GRCh38
rs777517252 Chr.1: 157519752-157519752 on Build GRCh38
rs1056189516 Chr.1: 157519968-157519968 on Build GRCh38
rs1029760886 Chr.1: 157519856-157519856 on Build GRCh38
rs368264000 Chr.1: 157519717-157519717 on Build GRCh38
rs758511781 Chr.1: 157519759-157519759 on Build GRCh38
rs1332714 Chr.1: 157519726-157519726 on Build GRCh38
rs773914175 Chr.1: 157519820-157519820 on Build GRCh38
rs538374266 Chr.1: 157519881-157519881 on Build GRCh38
rs747585502 Chr.1: 157519714-157519714 on Build GRCh38
rs1013495581 Chr.1: 157520092-157520092 on Build GRCh38
rs1038635569 Chr.1: 157520125-157520125 on Build GRCh38
rs944300899 Chr.1: 157519914-157519914 on Build GRCh38
rs760866738 Chr.1: 157520036-157520036 on Build GRCh38
rs150745177 Chr.1: 157519965-157519965 on Build GRCh38

View Full Product Details