Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27243150-27243643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVX1
Entrez Gene ID
2128
Gene Name
even-skipped homeobox 1
Gene Aliases
EVX-1
UniGene
Hs.369879
Gene Chromosome Location
Chr.7: 27242545-27247819 on Build GRCh38
Transcripts
NM_001304519.1 NM_001304520.1 NM_001989.4 NR_120507.1

Gene Symbol
EVX1-AS
Entrez Gene ID
101410536
Gene Name
EVX1 antisense RNA
Gene Aliases
EVX1-AS1
UniGene
Hs.687800
Gene Chromosome Location
Chr.7: 27241429-27247229 on Build GRCh38
Transcripts
NM_001304519.1 NM_001304520.1 NM_001989.4 NR_120507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5737891 c.305A>G p.D102G Chr.7: 27243335-27243335 on Build GRCh38
1131608 c.409G>A p.E137K Chr.7: 27243439-27243439 on Build GRCh38
6481909 c.424A>C p.K142Q Chr.7: 27243454-27243454 on Build GRCh38
6434886 c.410A>G p.E137G Chr.7: 27243440-27243440 on Build GRCh38
746298 c.423C>A p.S141R Chr.7: 27243453-27243453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757316376 Chr.7: 27243296-27243296 on Build GRCh38
rs776746533 Chr.7: 27243273-27243273 on Build GRCh38
rs993150459 Chr.7: 27243217-27243217 on Build GRCh38
rs1025927333 Chr.7: 27243220-27243220 on Build GRCh38
rs781625288 Chr.7: 27243429-27243429 on Build GRCh38
rs778102845 Chr.7: 27243417-27243417 on Build GRCh38
rs1050773381 Chr.7: 27243472-27243472 on Build GRCh38
rs759552042 Chr.7: 27243345-27243345 on Build GRCh38
rs563339648 Chr.7: 27243236-27243236 on Build GRCh38
rs977968177 Chr.7: 27243546-27243546 on Build GRCh38
rs544700966 Chr.7: 27243232-27243232 on Build GRCh38
rs761644611 Chr.7: 27243448-27243448 on Build GRCh38
rs546856629 Chr.7: 27243290-27243290 on Build GRCh38
rs933312252 Chr.7: 27243563-27243563 on Build GRCh38
rs779653755 Chr.7: 27243413-27243413 on Build GRCh38
rs756790055 Chr.7: 27243225-27243225 on Build GRCh38
rs750354243 Chr.7: 27243473-27243473 on Build GRCh38
rs772291612 Chr.7: 27243319-27243319 on Build GRCh38
rs868857633 Chr.7: 27243221-27243221 on Build GRCh38
rs754717437 Chr.7: 27243416-27243416 on Build GRCh38
rs141446680 Chr.7: 27243504-27243504 on Build GRCh38
rs917153256 Chr.7: 27243318-27243318 on Build GRCh38
rs769937342 Chr.7: 27243348-27243348 on Build GRCh38
rs777511332 Chr.7: 27243210-27243210 on Build GRCh38
rs755241999 Chr.7: 27243194-27243194 on Build GRCh38
rs968154364 Chr.7: 27243280-27243280 on Build GRCh38
rs966656572 Chr.7: 27243525-27243525 on Build GRCh38
rs750401561 Chr.7: 27243394-27243394 on Build GRCh38
rs576986682 Chr.7: 27243421-27243421 on Build GRCh38
rs538993502 Chr.7: 27243415-27243415 on Build GRCh38
rs752365793 Chr.7: 27243502-27243502 on Build GRCh38
rs924618538 Chr.7: 27243562-27243562 on Build GRCh38
rs774720259 Chr.7: 27243445-27243445 on Build GRCh38
rs368309805 Chr.7: 27243361-27243361 on Build GRCh38
rs776078595 Chr.7: 27243275-27243275 on Build GRCh38
rs1003718445 Chr.7: 27243426-27243426 on Build GRCh38
rs753454722 Chr.7: 27243482-27243482 on Build GRCh38
rs371249285 Chr.7: 27243501-27243501 on Build GRCh38
rs767443946 Chr.7: 27243463-27243463 on Build GRCh38
rs763278751 Chr.7: 27243359-27243359 on Build GRCh38
rs778248979 Chr.7: 27243320-27243320 on Build GRCh38
rs769733573 Chr.7: 27243430-27243430 on Build GRCh38
rs1031020320 Chr.7: 27243585-27243585 on Build GRCh38
rs777123254 Chr.7: 27243332-27243332 on Build GRCh38
rs750021188 Chr.7: 27243307-27243307 on Build GRCh38
rs917943781 Chr.7: 27243599-27243599 on Build GRCh38
rs148774441 Chr.7: 27243238-27243238 on Build GRCh38
rs761838789 Chr.7: 27243368-27243368 on Build GRCh38
rs183219836 Chr.7: 27243247-27243247 on Build GRCh38
rs758215109 Chr.7: 27243209-27243209 on Build GRCh38
rs751462613 Chr.7: 27243363-27243363 on Build GRCh38
rs779516582 Chr.7: 27243317-27243317 on Build GRCh38
rs186975658 Chr.7: 27243621-27243621 on Build GRCh38
rs1037871537 Chr.7: 27243606-27243607 on Build GRCh38
rs780837066 Chr.7: 27243302-27243302 on Build GRCh38
rs1000998666 Chr.7: 27243567-27243567 on Build GRCh38
rs754446648 Chr.7: 27243498-27243498 on Build GRCh38
rs765307739 Chr.7: 27243279-27243279 on Build GRCh38
rs1033939449 Chr.7: 27243235-27243235 on Build GRCh38
rs780894214 Chr.7: 27243231-27243231 on Build GRCh38
rs758083639 Chr.7: 27243288-27243288 on Build GRCh38
rs764809415 Chr.7: 27243499-27243499 on Build GRCh38
rs34266329 Chr.7: 27243543-27243543 on Build GRCh38
rs577545670 Chr.7: 27243169-27243169 on Build GRCh38
rs775538165 Chr.7: 27243352-27243352 on Build GRCh38
rs899456755 Chr.7: 27243325-27243325 on Build GRCh38
rs755691091 Chr.7: 27243316-27243316 on Build GRCh38
rs747500263 Chr.7: 27243323-27243323 on Build GRCh38
rs1007041941 Chr.7: 27243274-27243274 on Build GRCh38
rs761209465 Chr.7: 27243250-27243250 on Build GRCh38
rs768990201 Chr.7: 27243444-27243444 on Build GRCh38
rs993706585 Chr.7: 27243327-27243327 on Build GRCh38
rs775692773 Chr.7: 27243436-27243436 on Build GRCh38
rs545634008 Chr.7: 27243510-27243510 on Build GRCh38
rs759398187 Chr.7: 27243380-27243380 on Build GRCh38
rs746992899 Chr.7: 27243224-27243224 on Build GRCh38
rs767496829 Chr.7: 27243372-27243372 on Build GRCh38
rs752497574 Chr.7: 27243202-27243202 on Build GRCh38
rs528315654 Chr.7: 27243286-27243286 on Build GRCh38
rs899404417 Chr.7: 27243203-27243203 on Build GRCh38
rs771295828 Chr.7: 27243422-27243422 on Build GRCh38
rs760714073 Chr.7: 27243475-27243475 on Build GRCh38
rs751515921 Chr.7: 27243295-27243295 on Build GRCh38
rs750953182 Chr.7: 27243470-27243470 on Build GRCh38
rs757575772 Chr.7: 27243509-27243509 on Build GRCh38
rs530631267 Chr.7: 27243237-27243237 on Build GRCh38
rs763075504 Chr.7: 27243437-27243437 on Build GRCh38
rs989342734 Chr.7: 27243483-27243483 on Build GRCh38
rs955833398 Chr.7: 27243249-27243249 on Build GRCh38
rs765793689 Chr.7: 27243477-27243477 on Build GRCh38
rs971336524 Chr.7: 27243613-27243613 on Build GRCh38
rs1016446502 Chr.7: 27243172-27243172 on Build GRCh38
rs772500369 Chr.7: 27243239-27243239 on Build GRCh38
rs993811292 Chr.7: 27243438-27243438 on Build GRCh38
rs769629286 Chr.7: 27243234-27243234 on Build GRCh38
rs956808598 Chr.7: 27243588-27243588 on Build GRCh38
rs968585625 Chr.7: 27243529-27243529 on Build GRCh38
rs959094532 Chr.7: 27243457-27243457 on Build GRCh38
rs780361632 Chr.7: 27243593-27243593 on Build GRCh38
rs771491861 Chr.7: 27243324-27243324 on Build GRCh38
rs766423021 Chr.7: 27243263-27243263 on Build GRCh38
rs889301180 Chr.7: 27243313-27243313 on Build GRCh38
rs755531580 Chr.7: 27243396-27243396 on Build GRCh38

View Full Product Details