Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30600932-30601433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GARS
Entrez Gene ID
2617
Gene Name
glycyl-tRNA synthetase
Gene Aliases
CMT2D, DSMAV, GlyRS, HMN5, SMAD1
UniGene
Hs.404321
Gene Chromosome Location
Chr.7: 30594735-30634033 on Build GRCh38
Transcripts
NM_001316772.1 NM_002047.3 XM_006715686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5951292 c.517_521delGAACA p.E173fs*12 Chr.7: 30601148-30601152 on Build GRCh38
5951276 c.524T>A p.I175N Chr.7: 30601155-30601155 on Build GRCh38
6669351 c.563_564delTT p.L189fs*11 Chr.7: 30601194-30601195 on Build GRCh38
6548124 c.447C>T p.D149D Chr.7: 30601078-30601078 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540427205 Chr.7: 30601369-30601369 on Build GRCh38
rs758783290 Chr.7: 30601205-30601205 on Build GRCh38
rs755839241 Chr.7: 30601188-30601188 on Build GRCh38
rs771420365 Chr.7: 30601225-30601225 on Build GRCh38
rs199760356 Chr.7: 30601212-30601212 on Build GRCh38
rs186309742 Chr.7: 30601044-30601044 on Build GRCh38
rs375532339 Chr.7: 30601007-30601007 on Build GRCh38
rs773738689 Chr.7: 30601135-30601135 on Build GRCh38
rs370452042 Chr.7: 30601042-30601042 on Build GRCh38
rs774732829 Chr.7: 30601165-30601165 on Build GRCh38
rs368297381 Chr.7: 30601123-30601123 on Build GRCh38
rs752426125 Chr.7: 30601059-30601059 on Build GRCh38
rs771009601 Chr.7: 30601147-30601147 on Build GRCh38
rs527491538 Chr.7: 30600958-30600958 on Build GRCh38
rs934418828 Chr.7: 30601265-30601265 on Build GRCh38
rs914260908 Chr.7: 30601289-30601289 on Build GRCh38
rs376772628 Chr.7: 30601193-30601193 on Build GRCh38
rs745908895 Chr.7: 30601238-30601238 on Build GRCh38
rs569183579 Chr.7: 30601144-30601144 on Build GRCh38
rs772510751 Chr.7: 30601033-30601033 on Build GRCh38
rs767204701 Chr.7: 30601183-30601183 on Build GRCh38
rs775601213 Chr.7: 30601249-30601249 on Build GRCh38
rs893344608 Chr.7: 30600970-30600970 on Build GRCh38
rs1002947680 Chr.7: 30601022-30601022 on Build GRCh38
rs373782750 Chr.7: 30601046-30601046 on Build GRCh38
rs756853213 Chr.7: 30601090-30601090 on Build GRCh38
rs570753336 Chr.7: 30600988-30600988 on Build GRCh38
rs995659808 Chr.7: 30601026-30601026 on Build GRCh38
rs948965363 Chr.7: 30601315-30601315 on Build GRCh38
rs754718810 Chr.7: 30601038-30601038 on Build GRCh38
rs372221055 Chr.7: 30601166-30601166 on Build GRCh38
rs772899197 Chr.7: 30601168-30601168 on Build GRCh38
rs10280491 Chr.7: 30601373-30601373 on Build GRCh38
rs781387343 Chr.7: 30601236-30601236 on Build GRCh38
rs879200062 Chr.7: 30601217-30601217 on Build GRCh38
rs762875205 Chr.7: 30601174-30601174 on Build GRCh38
rs1044679573 Chr.7: 30601351-30601351 on Build GRCh38
rs137852644 Chr.7: 30601179-30601179 on Build GRCh38
rs554776015 Chr.7: 30601272-30601272 on Build GRCh38
rs925400735 Chr.7: 30601230-30601230 on Build GRCh38
rs367915362 Chr.7: 30601102-30601102 on Build GRCh38
rs765081631 Chr.7: 30601055-30601055 on Build GRCh38
rs766404526 Chr.7: 30601043-30601043 on Build GRCh38
rs769882122 Chr.7: 30601241-30601241 on Build GRCh38
rs1044932173 Chr.7: 30600997-30600997 on Build GRCh38
rs751558842 Chr.7: 30601177-30601177 on Build GRCh38
rs1049737836 Chr.7: 30601311-30601311 on Build GRCh38
rs779541328 Chr.7: 30601189-30601189 on Build GRCh38
rs772606983 Chr.7: 30601129-30601129 on Build GRCh38
rs763935450 Chr.7: 30601175-30601175 on Build GRCh38
rs757196240 Chr.7: 30601181-30601181 on Build GRCh38
rs573056537 Chr.7: 30601329-30601329 on Build GRCh38
rs755718829 Chr.7: 30601115-30601115 on Build GRCh38
rs62446863 Chr.7: 30601300-30601300 on Build GRCh38
rs773303002 Chr.7: 30601034-30601034 on Build GRCh38
rs778588443 Chr.7: 30601045-30601046 on Build GRCh38
rs554596840 Chr.7: 30600972-30600972 on Build GRCh38
rs758414117 Chr.7: 30601070-30601070 on Build GRCh38
rs377056075 Chr.7: 30601119-30601119 on Build GRCh38
rs201959920 Chr.7: 30601196-30601196 on Build GRCh38
rs759865366 Chr.7: 30601162-30601162 on Build GRCh38
rs776505252 Chr.7: 30601148-30601148 on Build GRCh38
rs989744262 Chr.7: 30601278-30601278 on Build GRCh38
rs961598440 Chr.7: 30601024-30601024 on Build GRCh38
rs372198757 Chr.7: 30601109-30601109 on Build GRCh38
rs751505742 Chr.7: 30601081-30601081 on Build GRCh38
rs139026353 Chr.7: 30601028-30601028 on Build GRCh38
rs764096597 Chr.7: 30601074-30601074 on Build GRCh38

View Full Product Details