Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30653249-30653742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR2
Entrez Gene ID
1395
Gene Name
corticotropin releasing hormone receptor 2
Gene Aliases
CRF-RB, CRF2, CRFR2, HM-CRF
UniGene
Hs.546246
Gene Chromosome Location
Chr.7: 30651844-30700103 on Build GRCh38
Transcripts
NM_001202475.1 NM_001202481.1 NM_001202482.1 NM_001202483.1 NM_001883.4 XM_011515127.2 XM_011515128.2 XM_011515129.2 XM_017011751.1 XM_017011752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635459 c.1181C>T p.T394M Chr.7: 30653473-30653473 on Build GRCh38
6635473 c.1160T>C p.M387T Chr.7: 30653536-30653536 on Build GRCh38
122358 c.1137C>G p.D379E Chr.7: 30653559-30653559 on Build GRCh38
4946207 c.1191G>A p.V397V Chr.7: 30653586-30653586 on Build GRCh38
6635471 c.1108C>T p.R370* Chr.7: 30653546-30653546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780561724 Chr.7: 30653432-30653432 on Build GRCh38
rs781666287 Chr.7: 30653424-30653425 on Build GRCh38
rs763155899 Chr.7: 30653571-30653571 on Build GRCh38
rs8192493 Chr.7: 30653408-30653408 on Build GRCh38
rs866153291 Chr.7: 30653370-30653370 on Build GRCh38
rs141315235 Chr.7: 30653449-30653449 on Build GRCh38
rs756762492 Chr.7: 30653426-30653426 on Build GRCh38
rs143275254 Chr.7: 30653598-30653598 on Build GRCh38
rs772634618 Chr.7: 30653531-30653531 on Build GRCh38
rs746454825 Chr.7: 30653517-30653518 on Build GRCh38
rs775532993 Chr.7: 30653644-30653644 on Build GRCh38
rs528785294 Chr.7: 30653386-30653386 on Build GRCh38
rs540775655 Chr.7: 30653498-30653498 on Build GRCh38
rs752866618 Chr.7: 30653469-30653469 on Build GRCh38
rs773864067 Chr.7: 30653450-30653450 on Build GRCh38
rs747361357 Chr.7: 30653628-30653628 on Build GRCh38
rs775168983 Chr.7: 30653444-30653444 on Build GRCh38
rs984282654 Chr.7: 30653308-30653308 on Build GRCh38
rs751563535 Chr.7: 30653488-30653488 on Build GRCh38
rs140591778 Chr.7: 30653473-30653473 on Build GRCh38
rs746890889 Chr.7: 30653425-30653425 on Build GRCh38
rs77113016 Chr.7: 30653465-30653465 on Build GRCh38
rs780797242 Chr.7: 30653512-30653512 on Build GRCh38
rs745402159 Chr.7: 30653517-30653517 on Build GRCh38
rs377066091 Chr.7: 30653437-30653437 on Build GRCh38
rs771274285 Chr.7: 30653634-30653634 on Build GRCh38
rs746319940 Chr.7: 30653642-30653642 on Build GRCh38
rs757121565 Chr.7: 30653501-30653501 on Build GRCh38
rs368317467 Chr.7: 30653643-30653643 on Build GRCh38
rs1032084775 Chr.7: 30653691-30653691 on Build GRCh38
rs1050143476 Chr.7: 30653347-30653347 on Build GRCh38
rs925614677 Chr.7: 30653639-30653639 on Build GRCh38
rs748545748 Chr.7: 30653446-30653446 on Build GRCh38
rs767377653 Chr.7: 30653593-30653593 on Build GRCh38
rs967266962 Chr.7: 30653661-30653661 on Build GRCh38
rs374100148 Chr.7: 30653589-30653589 on Build GRCh38
rs753606266 Chr.7: 30653602-30653602 on Build GRCh38
rs143322026 Chr.7: 30653500-30653500 on Build GRCh38
rs550672911 Chr.7: 30653454-30653454 on Build GRCh38
rs763934084 Chr.7: 30653583-30653583 on Build GRCh38
rs8192492 Chr.7: 30653546-30653546 on Build GRCh38
rs894891453 Chr.7: 30653715-30653715 on Build GRCh38
rs549812068 Chr.7: 30653530-30653530 on Build GRCh38
rs781562417 Chr.7: 30653635-30653635 on Build GRCh38
rs777151430 Chr.7: 30653548-30653548 on Build GRCh38
rs545097587 Chr.7: 30653318-30653318 on Build GRCh38
rs942080647 Chr.7: 30653284-30653284 on Build GRCh38
rs1000576664 Chr.7: 30653687-30653687 on Build GRCh38
rs1041787391 Chr.7: 30653356-30653356 on Build GRCh38
rs564864801 Chr.7: 30653442-30653442 on Build GRCh38
rs917033270 Chr.7: 30653577-30653577 on Build GRCh38
rs143693159 Chr.7: 30653533-30653533 on Build GRCh38
rs937042664 Chr.7: 30653320-30653320 on Build GRCh38
rs201296723 Chr.7: 30653565-30653565 on Build GRCh38
rs778319099 Chr.7: 30653607-30653607 on Build GRCh38
rs543221809 Chr.7: 30653379-30653379 on Build GRCh38
rs761262194 Chr.7: 30653453-30653453 on Build GRCh38
rs1050180655 Chr.7: 30653396-30653396 on Build GRCh38
rs952954179 Chr.7: 30653511-30653511 on Build GRCh38
rs373708051 Chr.7: 30653472-30653472 on Build GRCh38
rs151248954 Chr.7: 30653597-30653597 on Build GRCh38
rs984788167 Chr.7: 30653526-30653526 on Build GRCh38
rs367951133 Chr.7: 30653545-30653545 on Build GRCh38
rs754657829 Chr.7: 30653605-30653605 on Build GRCh38
rs992560867 Chr.7: 30653572-30653572 on Build GRCh38
rs765405966 Chr.7: 30653468-30653468 on Build GRCh38
rs759553448 Chr.7: 30653549-30653549 on Build GRCh38
rs199502493 Chr.7: 30653596-30653596 on Build GRCh38
rs575735314 Chr.7: 30653359-30653359 on Build GRCh38
rs772231611 Chr.7: 30653456-30653456 on Build GRCh38
rs900465146 Chr.7: 30653319-30653319 on Build GRCh38
rs546130607 Chr.7: 30653667-30653667 on Build GRCh38
rs768969599 Chr.7: 30653641-30653641 on Build GRCh38
rs779869891 Chr.7: 30653519-30653519 on Build GRCh38
rs188297269 Chr.7: 30653665-30653665 on Build GRCh38
rs755602103 Chr.7: 30653518-30653518 on Build GRCh38
rs202000400 Chr.7: 30653542-30653542 on Build GRCh38
rs370392453 Chr.7: 30653570-30653570 on Build GRCh38
rs563471497 Chr.7: 30653323-30653323 on Build GRCh38
rs79671818 Chr.7: 30653588-30653588 on Build GRCh38
rs916917706 Chr.7: 30653333-30653333 on Build GRCh38
rs1025925076 Chr.7: 30653278-30653278 on Build GRCh38
rs891043329 Chr.7: 30653353-30653353 on Build GRCh38
rs146600078 Chr.7: 30653380-30653380 on Build GRCh38

View Full Product Details