Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30664783-30665280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR2
Entrez Gene ID
1395
Gene Name
corticotropin releasing hormone receptor 2
Gene Aliases
CRF-RB, CRF2, CRFR2, HM-CRF
UniGene
Hs.546246
Gene Chromosome Location
Chr.7: 30651844-30700103 on Build GRCh38
Transcripts
NM_001202475.1 NM_001202481.1 NM_001202482.1 NM_001202483.1 NM_001883.4 XM_011515127.2 XM_011515128.2 XM_011515129.2 XM_017011751.1 XM_017011752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3028396 c.393C>A p.R131R Chr.7: 30665178-30665178 on Build GRCh38
3028397 c.514C>T p.R172C Chr.7: 30665180-30665180 on Build GRCh38
4162172 c.494T>G p.M165R Chr.7: 30665119-30665119 on Build GRCh38
3028398 c.433C>T p.R145C Chr.7: 30665180-30665180 on Build GRCh38
3028394 c.516C>A p.R172R Chr.7: 30665178-30665178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996899613 Chr.7: 30664991-30664991 on Build GRCh38
rs978234050 Chr.7: 30665202-30665202 on Build GRCh38
rs550502361 Chr.7: 30665124-30665124 on Build GRCh38
rs764427829 Chr.7: 30665073-30665073 on Build GRCh38
rs898239225 Chr.7: 30665137-30665137 on Build GRCh38
rs770131785 Chr.7: 30665238-30665238 on Build GRCh38
rs958491491 Chr.7: 30664821-30664821 on Build GRCh38
rs1042762266 Chr.7: 30664985-30664985 on Build GRCh38
rs764266631 Chr.7: 30665148-30665148 on Build GRCh38
rs374876028 Chr.7: 30665109-30665109 on Build GRCh38
rs776086218 Chr.7: 30665128-30665128 on Build GRCh38
rs992636710 Chr.7: 30664864-30664864 on Build GRCh38
rs955786861 Chr.7: 30665031-30665031 on Build GRCh38
rs756897258 Chr.7: 30665031-30665032 on Build GRCh38
rs747783455 Chr.7: 30665118-30665118 on Build GRCh38
rs747834580 Chr.7: 30665192-30665192 on Build GRCh38
rs902706014 Chr.7: 30665190-30665190 on Build GRCh38
rs1003884946 Chr.7: 30664902-30664902 on Build GRCh38
rs1028605197 Chr.7: 30665010-30665010 on Build GRCh38
rs150872538 Chr.7: 30665082-30665082 on Build GRCh38
rs532304085 Chr.7: 30665106-30665106 on Build GRCh38
rs376545794 Chr.7: 30665056-30665056 on Build GRCh38
rs939855115 Chr.7: 30665107-30665107 on Build GRCh38
rs557586574 Chr.7: 30664872-30664872 on Build GRCh38
rs1045950895 Chr.7: 30664885-30664885 on Build GRCh38
rs750208111 Chr.7: 30665165-30665165 on Build GRCh38
rs576019745 Chr.7: 30664889-30664889 on Build GRCh38
rs756036342 Chr.7: 30665099-30665099 on Build GRCh38
rs774840630 Chr.7: 30665039-30665039 on Build GRCh38
rs776826662 Chr.7: 30665120-30665120 on Build GRCh38
rs76778629 Chr.7: 30665059-30665059 on Build GRCh38
rs777437048 Chr.7: 30665212-30665212 on Build GRCh38
rs200091848 Chr.7: 30665042-30665042 on Build GRCh38
rs774855744 Chr.7: 30665179-30665179 on Build GRCh38
rs765402556 Chr.7: 30665060-30665060 on Build GRCh38
rs915800608 Chr.7: 30664810-30664810 on Build GRCh38
rs773933160 Chr.7: 30665047-30665047 on Build GRCh38
rs745866563 Chr.7: 30665091-30665091 on Build GRCh38
rs751766093 Chr.7: 30665081-30665081 on Build GRCh38
rs1038110342 Chr.7: 30665110-30665110 on Build GRCh38
rs750730176 Chr.7: 30665164-30665174 on Build GRCh38
rs966858198 Chr.7: 30665048-30665048 on Build GRCh38
rs1038077801 Chr.7: 30664958-30664958 on Build GRCh38
rs898208128 Chr.7: 30664979-30664979 on Build GRCh38
rs143109330 Chr.7: 30665119-30665119 on Build GRCh38
rs774055102 Chr.7: 30665114-30665114 on Build GRCh38
rs758667239 Chr.7: 30665019-30665019 on Build GRCh38
rs572905690 Chr.7: 30665057-30665057 on Build GRCh38
rs949927229 Chr.7: 30664862-30664862 on Build GRCh38
rs751889993 Chr.7: 30665150-30665150 on Build GRCh38
rs778104497 Chr.7: 30665020-30665020 on Build GRCh38
rs908419152 Chr.7: 30665094-30665094 on Build GRCh38
rs949524731 Chr.7: 30664909-30664909 on Build GRCh38
rs781196298 Chr.7: 30665028-30665028 on Build GRCh38
rs927219307 Chr.7: 30664807-30664807 on Build GRCh38
rs748756901 Chr.7: 30665040-30665040 on Build GRCh38
rs746826366 Chr.7: 30665021-30665021 on Build GRCh38
rs191752150 Chr.7: 30665157-30665157 on Build GRCh38
rs754720556 Chr.7: 30664978-30664978 on Build GRCh38
rs370570799 Chr.7: 30665065-30665065 on Build GRCh38
rs745790176 Chr.7: 30665032-30665032 on Build GRCh38
rs146042188 Chr.7: 30665170-30665170 on Build GRCh38
rs748820749 Chr.7: 30665063-30665063 on Build GRCh38
rs780385856 Chr.7: 30665223-30665224 on Build GRCh38
rs545294772 Chr.7: 30664868-30664868 on Build GRCh38
rs548124161 Chr.7: 30665247-30665247 on Build GRCh38
rs577428438 Chr.7: 30665049-30665049 on Build GRCh38
rs777302611 Chr.7: 30664945-30664945 on Build GRCh38
rs763132126 Chr.7: 30665129-30665129 on Build GRCh38
rs958626492 Chr.7: 30665151-30665156 on Build GRCh38
rs371329676 Chr.7: 30665199-30665199 on Build GRCh38
rs757400435 Chr.7: 30665152-30665152 on Build GRCh38
rs201981872 Chr.7: 30665050-30665050 on Build GRCh38
rs578098304 Chr.7: 30664805-30664805 on Build GRCh38
rs756684910 Chr.7: 30665223-30665223 on Build GRCh38
rs770794509 Chr.7: 30665022-30665022 on Build GRCh38
rs759910018 Chr.7: 30665123-30665123 on Build GRCh38
rs746235196 Chr.7: 30665233-30665233 on Build GRCh38
rs778368609 Chr.7: 30665180-30665180 on Build GRCh38
rs924003549 Chr.7: 30664998-30665001 on Build GRCh38
rs184589106 Chr.7: 30665171-30665171 on Build GRCh38
rs932350085 Chr.7: 30665145-30665145 on Build GRCh38
rs767926372 Chr.7: 30665032-30665032 on Build GRCh38
rs779991159 Chr.7: 30665105-30665105 on Build GRCh38

View Full Product Details