Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30968834-30969378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRHR
Entrez Gene ID
2692
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
UniGene
Hs.733003
Gene Chromosome Location
Chr.7: 30964021-30979531 on Build GRCh38
Transcripts
NM_000823.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3923489 c.79G>A p.E27K Chr.7: 30968855-30968855 on Build GRCh38
32398 c.133G>A p.A45T Chr.7: 30968909-30968909 on Build GRCh38
7111705 c.79G>C p.E27Q Chr.7: 30968855-30968855 on Build GRCh38
6576922 c.195G>A p.W65* Chr.7: 30969097-30969097 on Build GRCh38
4479134 c.229C>T p.P77S Chr.7: 30969131-30969131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930160852 Chr.7: 30969305-30969305 on Build GRCh38
rs865904751 Chr.7: 30969170-30969170 on Build GRCh38
rs751885823 Chr.7: 30969166-30969166 on Build GRCh38
rs764582464 Chr.7: 30968896-30968896 on Build GRCh38
rs762629722 Chr.7: 30969043-30969043 on Build GRCh38
rs368404708 Chr.7: 30968953-30968953 on Build GRCh38
rs977460905 Chr.7: 30968964-30968964 on Build GRCh38
rs752793580 Chr.7: 30969072-30969072 on Build GRCh38
rs192109771 Chr.7: 30968991-30968991 on Build GRCh38
rs768112214 Chr.7: 30969115-30969115 on Build GRCh38
rs748443344 Chr.7: 30968946-30968946 on Build GRCh38
rs989948340 Chr.7: 30969151-30969151 on Build GRCh38
rs199857325 Chr.7: 30968986-30968986 on Build GRCh38
rs763709666 Chr.7: 30969051-30969051 on Build GRCh38
rs754256570 Chr.7: 30968935-30968935 on Build GRCh38
rs781077252 Chr.7: 30969177-30969177 on Build GRCh38
rs758585465 Chr.7: 30969150-30969150 on Build GRCh38
rs922813339 Chr.7: 30969302-30969302 on Build GRCh38
rs774857155 Chr.7: 30969041-30969041 on Build GRCh38
rs760129030 Chr.7: 30969135-30969135 on Build GRCh38
rs761056409 Chr.7: 30969054-30969054 on Build GRCh38
rs28371559 Chr.7: 30969282-30969282 on Build GRCh38
rs759110872 Chr.7: 30968872-30968872 on Build GRCh38
rs201978703 Chr.7: 30968911-30968911 on Build GRCh38
rs773323831 Chr.7: 30968912-30968914 on Build GRCh38
rs1008727395 Chr.7: 30969323-30969323 on Build GRCh38
rs146723194 Chr.7: 30968902-30968902 on Build GRCh38
rs965353400 Chr.7: 30968888-30968888 on Build GRCh38
rs769403061 Chr.7: 30969019-30969019 on Build GRCh38
rs776859854 Chr.7: 30969132-30969132 on Build GRCh38
rs755430803 Chr.7: 30969067-30969067 on Build GRCh38
rs764209354 Chr.7: 30969155-30969155 on Build GRCh38
rs34284452 Chr.7: 30969213-30969213 on Build GRCh38
rs577536745 Chr.7: 30969099-30969099 on Build GRCh38
rs942863057 Chr.7: 30969002-30969002 on Build GRCh38
rs376235715 Chr.7: 30969014-30969014 on Build GRCh38
rs554963225 Chr.7: 30969344-30969344 on Build GRCh38
rs28371558 Chr.7: 30969139-30969139 on Build GRCh38
rs948242768 Chr.7: 30968997-30968997 on Build GRCh38
rs563078420 Chr.7: 30969103-30969103 on Build GRCh38
rs540740642 Chr.7: 30969076-30969076 on Build GRCh38
rs4988496 Chr.7: 30969071-30969071 on Build GRCh38
rs777751805 Chr.7: 30968974-30968974 on Build GRCh38
rs770962010 Chr.7: 30968863-30968863 on Build GRCh38
rs544345041 Chr.7: 30969275-30969275 on Build GRCh38
rs745989289 Chr.7: 30969001-30969001 on Build GRCh38
rs200887870 Chr.7: 30968981-30968981 on Build GRCh38
rs879337316 Chr.7: 30969261-30969261 on Build GRCh38
rs867537571 Chr.7: 30969097-30969097 on Build GRCh38
rs773941149 Chr.7: 30969053-30969053 on Build GRCh38
rs121918117 Chr.7: 30969116-30969116 on Build GRCh38
rs768237840 Chr.7: 30969348-30969349 on Build GRCh38
rs765640577 Chr.7: 30969138-30969138 on Build GRCh38
rs141571344 Chr.7: 30968927-30968927 on Build GRCh38
rs762580559 Chr.7: 30968909-30968909 on Build GRCh38
rs4988495 Chr.7: 30968992-30968992 on Build GRCh38
rs535744163 Chr.7: 30969314-30969314 on Build GRCh38
rs766587678 Chr.7: 30969063-30969063 on Build GRCh38
rs565445448 Chr.7: 30969091-30969091 on Build GRCh38
rs570325656 Chr.7: 30968944-30968944 on Build GRCh38
rs527349420 Chr.7: 30969252-30969252 on Build GRCh38
rs373112876 Chr.7: 30969011-30969011 on Build GRCh38
rs1036622816 Chr.7: 30969277-30969277 on Build GRCh38
rs568313679 Chr.7: 30969299-30969299 on Build GRCh38
rs755092627 Chr.7: 30968937-30968937 on Build GRCh38
rs143429066 Chr.7: 30968869-30968869 on Build GRCh38
rs754386566 Chr.7: 30969066-30969066 on Build GRCh38
rs755889034 Chr.7: 30969199-30969199 on Build GRCh38
rs757412947 Chr.7: 30969171-30969171 on Build GRCh38
rs201893637 Chr.7: 30969102-30969102 on Build GRCh38
rs958079133 Chr.7: 30969148-30969148 on Build GRCh38
rs35071506 Chr.7: 30969256-30969256 on Build GRCh38
rs185014978 Chr.7: 30969269-30969269 on Build GRCh38
rs1047290898 Chr.7: 30969348-30969348 on Build GRCh38
rs761399375 Chr.7: 30969121-30969121 on Build GRCh38
rs967674479 Chr.7: 30969191-30969191 on Build GRCh38
rs769587851 Chr.7: 30969109-30969109 on Build GRCh38
rs139126021 Chr.7: 30969349-30969350 on Build GRCh38
rs200679786 Chr.7: 30969347-30969348 on Build GRCh38
rs958401552 Chr.7: 30968929-30968929 on Build GRCh38
rs756658887 Chr.7: 30968920-30968920 on Build GRCh38
rs775025721 Chr.7: 30969110-30969110 on Build GRCh38
rs760761162 Chr.7: 30968857-30968857 on Build GRCh38
rs930342802 Chr.7: 30969290-30969290 on Build GRCh38
rs940355976 Chr.7: 30969036-30969036 on Build GRCh38
rs141243007 Chr.7: 30969194-30969194 on Build GRCh38
rs547825255 Chr.7: 30969128-30969128 on Build GRCh38
rs1050194454 Chr.7: 30969083-30969083 on Build GRCh38
rs745733075 Chr.7: 30969018-30969018 on Build GRCh38
rs369751652 Chr.7: 30969073-30969073 on Build GRCh38
rs770802637 Chr.7: 30968985-30968985 on Build GRCh38
rs745628562 Chr.7: 30969192-30969192 on Build GRCh38
rs888896902 Chr.7: 30969309-30969309 on Build GRCh38
rs140417128 Chr.7: 30968917-30968917 on Build GRCh38
rs994960254 Chr.7: 30969075-30969075 on Build GRCh38
rs375206873 Chr.7: 30968914-30968914 on Build GRCh38

View Full Product Details