Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31549347-31549855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC129
Entrez Gene ID
223075
Gene Name
coiled-coil domain containing 129
Gene Aliases
-
UniGene
Hs.224269
Gene Chromosome Location
Chr.7: 31514071-31658720 on Build GRCh38
Transcripts
NM_001257967.1 NM_001257968.1 NR_047565.1 XM_011515213.1 XM_017011871.1 XM_017011872.1

SNP Detail

SNP ID Chromosome Location
rs932736878 Chr.7: 31549554-31549554 on Build GRCh38
rs185080697 Chr.7: 31549399-31549399 on Build GRCh38
rs566949861 Chr.7: 31549737-31549737 on Build GRCh38
rs541780775 Chr.7: 31549511-31549511 on Build GRCh38
rs959776083 Chr.7: 31549753-31549753 on Build GRCh38
rs143420740 Chr.7: 31549631-31549631 on Build GRCh38
rs530349399 Chr.7: 31549497-31549497 on Build GRCh38
rs139971214 Chr.7: 31549545-31549545 on Build GRCh38
rs979785775 Chr.7: 31549445-31549445 on Build GRCh38
rs959674024 Chr.7: 31549491-31549491 on Build GRCh38
rs967998560 Chr.7: 31549776-31549776 on Build GRCh38
rs149257277 Chr.7: 31549825-31549825 on Build GRCh38
rs1021949514 Chr.7: 31549757-31549757 on Build GRCh38
rs569773629 Chr.7: 31549768-31549768 on Build GRCh38
rs1001255481 Chr.7: 31549701-31549701 on Build GRCh38
rs926528345 Chr.7: 31549823-31549823 on Build GRCh38
rs776585187 Chr.7: 31549781-31549781 on Build GRCh38
rs771055289 Chr.7: 31549488-31549488 on Build GRCh38
rs754675270 Chr.7: 31549435-31549435 on Build GRCh38
rs888728726 Chr.7: 31549542-31549542 on Build GRCh38
rs914194173 Chr.7: 31549506-31549506 on Build GRCh38
rs969781029 Chr.7: 31549436-31549436 on Build GRCh38
rs1026824781 Chr.7: 31549661-31549661 on Build GRCh38
rs189973663 Chr.7: 31549624-31549624 on Build GRCh38
rs533619953 Chr.7: 31549616-31549616 on Build GRCh38
rs891197810 Chr.7: 31549581-31549581 on Build GRCh38
rs1049818050 Chr.7: 31549571-31549571 on Build GRCh38
rs10248545 Chr.7: 31549652-31549652 on Build GRCh38
rs980678169 Chr.7: 31549811-31549811 on Build GRCh38
rs117297547 Chr.7: 31549463-31549463 on Build GRCh38
rs543816671 Chr.7: 31549456-31549456 on Build GRCh38
rs1039905874 Chr.7: 31549612-31549612 on Build GRCh38
rs917300905 Chr.7: 31549510-31549510 on Build GRCh38
rs998325210 Chr.7: 31549646-31549646 on Build GRCh38
rs1027229895 Chr.7: 31549374-31549374 on Build GRCh38
rs956452929 Chr.7: 31549696-31549696 on Build GRCh38
rs921731839 Chr.7: 31549680-31549680 on Build GRCh38
rs548587999 Chr.7: 31549665-31549665 on Build GRCh38
rs528878586 Chr.7: 31549629-31549629 on Build GRCh38
rs112315387 Chr.7: 31549512-31549512 on Build GRCh38
rs1009264004 Chr.7: 31549708-31549708 on Build GRCh38

View Full Product Details