Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 31554390-31554889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC129
Entrez Gene ID
223075
Gene Name
coiled-coil domain containing 129
Gene Aliases
-
UniGene
Hs.224269
Gene Chromosome Location
Chr.7: 31514071-31658720 on Build GRCh38
Transcripts
NM_001257967.1 NM_001257968.1 NM_194300.3 NR_047565.1 XM_011515213.1 XM_017011871.1 XM_017011872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
145198 c.209T>A p.F70Y Chr.7: 31554520-31554520 on Build GRCh38
7359721 c.212+2T>G p.? Chr.7: 31554525-31554525 on Build GRCh38
7359719 c.290+2T>G p.? Chr.7: 31554525-31554525 on Build GRCh38
1496617 c.166G>T p.D56Y Chr.7: 31554477-31554477 on Build GRCh38
1329867 c.189G>A p.Q63Q Chr.7: 31554500-31554500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765617651 Chr.7: 31554492-31554492 on Build GRCh38
rs746748979 Chr.7: 31554534-31554535 on Build GRCh38
rs774810851 Chr.7: 31554838-31554838 on Build GRCh38
rs560277245 Chr.7: 31554795-31554795 on Build GRCh38
rs200034335 Chr.7: 31554500-31554500 on Build GRCh38
rs764471903 Chr.7: 31554502-31554502 on Build GRCh38
rs749235459 Chr.7: 31554473-31554473 on Build GRCh38
rs987963583 Chr.7: 31554574-31554574 on Build GRCh38
rs775654781 Chr.7: 31554493-31554493 on Build GRCh38
rs770427577 Chr.7: 31554557-31554557 on Build GRCh38
rs755920022 Chr.7: 31554519-31554519 on Build GRCh38
rs574568785 Chr.7: 31554731-31554731 on Build GRCh38
rs1017616052 Chr.7: 31554613-31554613 on Build GRCh38
rs200507019 Chr.7: 31554456-31554456 on Build GRCh38
rs781720577 Chr.7: 31554776-31554776 on Build GRCh38
rs757308052 Chr.7: 31554514-31554514 on Build GRCh38
rs750639860 Chr.7: 31554515-31554515 on Build GRCh38
rs556209592 Chr.7: 31554703-31554703 on Build GRCh38
rs372723381 Chr.7: 31554859-31554859 on Build GRCh38
rs774028957 Chr.7: 31554479-31554479 on Build GRCh38
rs558038802 Chr.7: 31554580-31554580 on Build GRCh38
rs757505766 Chr.7: 31554425-31554425 on Build GRCh38
rs200078907 Chr.7: 31554535-31554535 on Build GRCh38
rs773189011 Chr.7: 31554535-31554536 on Build GRCh38
rs781345316 Chr.7: 31554444-31554444 on Build GRCh38
rs897048223 Chr.7: 31554847-31554847 on Build GRCh38
rs938309032 Chr.7: 31554789-31554789 on Build GRCh38
rs78453087 Chr.7: 31554524-31554524 on Build GRCh38
rs948428649 Chr.7: 31554507-31554507 on Build GRCh38
rs934782382 Chr.7: 31554434-31554434 on Build GRCh38
rs747698756 Chr.7: 31554545-31554545 on Build GRCh38
rs768410299 Chr.7: 31554474-31554474 on Build GRCh38
rs534039239 Chr.7: 31554697-31554697 on Build GRCh38
rs966317086 Chr.7: 31554624-31554624 on Build GRCh38
rs1027751905 Chr.7: 31554581-31554581 on Build GRCh38
rs928308724 Chr.7: 31554753-31554753 on Build GRCh38
rs956252767 Chr.7: 31554665-31554665 on Build GRCh38
rs200315406 Chr.7: 31554560-31554560 on Build GRCh38
rs755133738 Chr.7: 31554484-31554484 on Build GRCh38
rs773053944 Chr.7: 31554547-31554547 on Build GRCh38
rs12155214 Chr.7: 31554644-31554644 on Build GRCh38
rs747611396 Chr.7: 31554481-31554481 on Build GRCh38
rs751664156 Chr.7: 31554509-31554509 on Build GRCh38
rs745714084 Chr.7: 31554452-31554452 on Build GRCh38
rs979824542 Chr.7: 31554737-31554737 on Build GRCh38
rs948458331 Chr.7: 31554710-31554710 on Build GRCh38
rs1015007549 Chr.7: 31554777-31554777 on Build GRCh38
rs990695504 Chr.7: 31554672-31554672 on Build GRCh38
rs1014997769 Chr.7: 31554440-31554440 on Build GRCh38
rs770929032 Chr.7: 31554547-31554553 on Build GRCh38
rs201046422 Chr.7: 31554537-31554537 on Build GRCh38
rs1005049050 Chr.7: 31554757-31554757 on Build GRCh38
rs540436650 Chr.7: 31554460-31554460 on Build GRCh38
rs754963857 Chr.7: 31554531-31554531 on Build GRCh38
rs2286710 Chr.7: 31554552-31554552 on Build GRCh38
rs759882354 Chr.7: 31554489-31554489 on Build GRCh38
rs1005449564 Chr.7: 31554416-31554416 on Build GRCh38
rs887846442 Chr.7: 31554599-31554599 on Build GRCh38
rs768968479 Chr.7: 31554524-31554527 on Build GRCh38
rs1039395469 Chr.7: 31554622-31554622 on Build GRCh38
rs1007581503 Chr.7: 31554604-31554604 on Build GRCh38
rs369275747 Chr.7: 31554469-31554469 on Build GRCh38
rs1054215154 Chr.7: 31554435-31554435 on Build GRCh38
rs775958475 Chr.7: 31554559-31554559 on Build GRCh38
rs568184446 Chr.7: 31554488-31554488 on Build GRCh38
rs982029418 Chr.7: 31554539-31554539 on Build GRCh38
rs371691830 Chr.7: 31554534-31554534 on Build GRCh38
rs201836843 Chr.7: 31554536-31554536 on Build GRCh38
rs764636129 Chr.7: 31554568-31554568 on Build GRCh38
rs776361517 Chr.7: 31554824-31554827 on Build GRCh38
rs541951342 Chr.7: 31554767-31554767 on Build GRCh38
rs976341000 Chr.7: 31554632-31554632 on Build GRCh38
rs771700230 Chr.7: 31554546-31554546 on Build GRCh38

View Full Product Details