Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 32999847-33000388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP9
Entrez Gene ID
11328
Gene Name
FK506 binding protein 9
Gene Aliases
FKBP60, FKBP63, PPIase
UniGene
Hs.103934
Gene Chromosome Location
Chr.7: 32957404-33006931 on Build GRCh38
Transcripts
NM_001284341.1 NM_001284343.1 NM_007270.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
300056 c.1336A>G p.I446V Chr.7: 33000224-33000224 on Build GRCh38
334774 c.615C>T p.C205C Chr.7: 33000199-33000199 on Build GRCh38
1089254 c.660T>C p.Y220Y Chr.7: 33000244-33000244 on Build GRCh38
1089252 c.626A>C p.K209T Chr.7: 33000210-33000210 on Build GRCh38
275179 c.1283G>A p.G428E Chr.7: 33000171-33000171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048244137 Chr.7: 33000084-33000084 on Build GRCh38
rs755142682 Chr.7: 33000163-33000163 on Build GRCh38
rs758571417 Chr.7: 33000197-33000197 on Build GRCh38
rs754296702 Chr.7: 33000160-33000160 on Build GRCh38
rs766930304 Chr.7: 33000263-33000263 on Build GRCh38
rs543328342 Chr.7: 33000297-33000297 on Build GRCh38
rs771054225 Chr.7: 33000213-33000213 on Build GRCh38
rs201129637 Chr.7: 33000096-33000096 on Build GRCh38
rs924391012 Chr.7: 32999921-32999921 on Build GRCh38
rs971168032 Chr.7: 32999897-32999897 on Build GRCh38
rs1024226015 Chr.7: 33000212-33000212 on Build GRCh38
rs746750732 Chr.7: 33000079-33000079 on Build GRCh38
rs377018562 Chr.7: 33000063-33000063 on Build GRCh38
rs761166680 Chr.7: 33000257-33000257 on Build GRCh38
rs566489199 Chr.7: 33000061-33000061 on Build GRCh38
rs767975179 Chr.7: 33000123-33000123 on Build GRCh38
rs762562679 Chr.7: 33000109-33000109 on Build GRCh38
rs775240947 Chr.7: 33000241-33000241 on Build GRCh38
rs779245562 Chr.7: 33000166-33000166 on Build GRCh38
rs769297248 Chr.7: 33000229-33000229 on Build GRCh38
rs770862136 Chr.7: 33000088-33000088 on Build GRCh38
rs762324524 Chr.7: 33000243-33000243 on Build GRCh38
rs369317981 Chr.7: 33000274-33000274 on Build GRCh38
rs369594659 Chr.7: 33000178-33000178 on Build GRCh38
rs757237298 Chr.7: 33000305-33000305 on Build GRCh38
rs894387433 Chr.7: 33000174-33000174 on Build GRCh38
rs533537841 Chr.7: 33000072-33000072 on Build GRCh38
rs943632085 Chr.7: 33000039-33000039 on Build GRCh38
rs776550035 Chr.7: 33000223-33000223 on Build GRCh38
rs756417253 Chr.7: 33000142-33000142 on Build GRCh38
rs766738302 Chr.7: 33000157-33000157 on Build GRCh38
rs774838321 Chr.7: 33000098-33000098 on Build GRCh38
rs369275537 Chr.7: 33000273-33000273 on Build GRCh38
rs145311636 Chr.7: 33000035-33000035 on Build GRCh38
rs532391350 Chr.7: 32999870-32999870 on Build GRCh38
rs554943247 Chr.7: 33000298-33000298 on Build GRCh38
rs759103068 Chr.7: 33000094-33000094 on Build GRCh38
rs377272641 Chr.7: 33000252-33000252 on Build GRCh38
rs910788129 Chr.7: 33000001-33000001 on Build GRCh38
rs750953924 Chr.7: 33000136-33000136 on Build GRCh38
rs376122812 Chr.7: 33000285-33000285 on Build GRCh38
rs373607944 Chr.7: 33000354-33000354 on Build GRCh38
rs201284807 Chr.7: 33000199-33000199 on Build GRCh38
rs1055076709 Chr.7: 33000114-33000114 on Build GRCh38
rs550794875 Chr.7: 32999999-32999999 on Build GRCh38
rs956995618 Chr.7: 32999944-32999944 on Build GRCh38
rs977536168 Chr.7: 32999899-32999899 on Build GRCh38
rs781228986 Chr.7: 33000311-33000311 on Build GRCh38
rs1024592850 Chr.7: 32999892-32999892 on Build GRCh38
rs373354190 Chr.7: 33000280-33000280 on Build GRCh38
rs773312000 Chr.7: 33000078-33000078 on Build GRCh38
rs202084262 Chr.7: 33000200-33000200 on Build GRCh38
rs776355757 Chr.7: 33000091-33000091 on Build GRCh38
rs372601051 Chr.7: 33000228-33000228 on Build GRCh38
rs764926975 Chr.7: 33000324-33000325 on Build GRCh38
rs61259431 Chr.7: 33000040-33000040 on Build GRCh38
rs746947172 Chr.7: 33000205-33000205 on Build GRCh38
rs773646822 Chr.7: 33000256-33000256 on Build GRCh38
rs569030731 Chr.7: 33000002-33000002 on Build GRCh38
rs58586080 Chr.7: 33000316-33000316 on Build GRCh38
rs964956642 Chr.7: 33000319-33000319 on Build GRCh38
rs375699561 Chr.7: 33000292-33000292 on Build GRCh38
rs748319140 Chr.7: 33000065-33000065 on Build GRCh38
rs989831538 Chr.7: 32999946-32999946 on Build GRCh38

View Full Product Details