Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36855355-36855849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO1
Entrez Gene ID
9844
Gene Name
engulfment and cell motility 1
Gene Aliases
CED-12, CED12, ELMO-1
UniGene
Hs.434989
Gene Chromosome Location
Chr.7: 36852906-37449409 on Build GRCh38
Transcripts
NM_001039459.2 NM_001206480.2 NM_001206482.1 NM_014800.10 NM_130442.3 NR_038120.1 XM_005249919.2 XM_006715805.1 XM_011515654.1 XM_017012838.1 XM_017012839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6436826 c.2038G>A p.D680N Chr.7: 36855697-36855697 on Build GRCh38
7366129 c.1127G>T p.G376V Chr.7: 36855714-36855714 on Build GRCh38
1161677 c.2176T>C p.C726R Chr.7: 36855559-36855559 on Build GRCh38
6110339 c.1207C>A p.L403M Chr.7: 36855634-36855634 on Build GRCh38
5358159 c.2067G>A p.L689L Chr.7: 36855668-36855668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757503801 Chr.7: 36855661-36855661 on Build GRCh38
rs751558501 Chr.7: 36855522-36855522 on Build GRCh38
rs748010154 Chr.7: 36855803-36855803 on Build GRCh38
rs542140804 Chr.7: 36855824-36855824 on Build GRCh38
rs201616410 Chr.7: 36855687-36855687 on Build GRCh38
rs764481575 Chr.7: 36855730-36855730 on Build GRCh38
rs750500849 Chr.7: 36855757-36855757 on Build GRCh38
rs958651939 Chr.7: 36855527-36855527 on Build GRCh38
rs138633855 Chr.7: 36855587-36855587 on Build GRCh38
rs757295660 Chr.7: 36855723-36855723 on Build GRCh38
rs757405131 Chr.7: 36855530-36855530 on Build GRCh38
rs986799793 Chr.7: 36855670-36855670 on Build GRCh38
rs756037174 Chr.7: 36855689-36855689 on Build GRCh38
rs200650822 Chr.7: 36855515-36855515 on Build GRCh38
rs984811545 Chr.7: 36855739-36855739 on Build GRCh38
rs149257728 Chr.7: 36855611-36855611 on Build GRCh38
rs766206964 Chr.7: 36855778-36855778 on Build GRCh38
rs911454272 Chr.7: 36855671-36855671 on Build GRCh38
rs747851553 Chr.7: 36855408-36855408 on Build GRCh38
rs774095292 Chr.7: 36855550-36855550 on Build GRCh38
rs779767064 Chr.7: 36855543-36855543 on Build GRCh38
rs770393657 Chr.7: 36855721-36855721 on Build GRCh38
rs374523052 Chr.7: 36855774-36855774 on Build GRCh38
rs962203529 Chr.7: 36855568-36855568 on Build GRCh38
rs918674508 Chr.7: 36855610-36855610 on Build GRCh38
rs930688281 Chr.7: 36855409-36855409 on Build GRCh38
rs769728173 Chr.7: 36855539-36855539 on Build GRCh38
rs554868519 Chr.7: 36855478-36855478 on Build GRCh38
rs774585018 Chr.7: 36855734-36855734 on Build GRCh38
rs964094394 Chr.7: 36855608-36855608 on Build GRCh38
rs142425368 Chr.7: 36855719-36855719 on Build GRCh38
rs1041433452 Chr.7: 36855790-36855790 on Build GRCh38
rs892851067 Chr.7: 36855599-36855599 on Build GRCh38
rs747766842 Chr.7: 36855710-36855710 on Build GRCh38
rs377709624 Chr.7: 36855602-36855602 on Build GRCh38
rs115635114 Chr.7: 36855463-36855463 on Build GRCh38
rs894159444 Chr.7: 36855826-36855826 on Build GRCh38
rs943249869 Chr.7: 36855473-36855473 on Build GRCh38
rs34022626 Chr.7: 36855669-36855670 on Build GRCh38
rs368359705 Chr.7: 36855462-36855462 on Build GRCh38
rs575599293 Chr.7: 36855817-36855817 on Build GRCh38
rs906980907 Chr.7: 36855798-36855798 on Build GRCh38
rs751754505 Chr.7: 36855645-36855645 on Build GRCh38
rs1034259774 Chr.7: 36855508-36855508 on Build GRCh38
rs919315359 Chr.7: 36855402-36855402 on Build GRCh38
rs765898655 Chr.7: 36855619-36855619 on Build GRCh38
rs137942550 Chr.7: 36855683-36855683 on Build GRCh38
rs776112181 Chr.7: 36855722-36855722 on Build GRCh38
rs749009029 Chr.7: 36855692-36855692 on Build GRCh38
rs570043981 Chr.7: 36855400-36855400 on Build GRCh38
rs745767117 Chr.7: 36855533-36855533 on Build GRCh38
rs1002548174 Chr.7: 36855498-36855498 on Build GRCh38
rs767784847 Chr.7: 36855738-36855738 on Build GRCh38
rs943370974 Chr.7: 36855783-36855783 on Build GRCh38
rs752998607 Chr.7: 36855620-36855620 on Build GRCh38
rs866035568 Chr.7: 36855704-36855704 on Build GRCh38
rs749169817 Chr.7: 36855544-36855544 on Build GRCh38
rs1054649854 Chr.7: 36855701-36855701 on Build GRCh38
rs13246439 Chr.7: 36855737-36855737 on Build GRCh38
rs866540184 Chr.7: 36855474-36855474 on Build GRCh38
rs778708825 Chr.7: 36855801-36855801 on Build GRCh38
rs758531963 Chr.7: 36855500-36855500 on Build GRCh38
rs768548471 Chr.7: 36855697-36855697 on Build GRCh38
rs985166813 Chr.7: 36855434-36855434 on Build GRCh38
rs761208793 Chr.7: 36855552-36855552 on Build GRCh38
rs772830948 Chr.7: 36855716-36855716 on Build GRCh38
rs182459066 Chr.7: 36855690-36855690 on Build GRCh38
rs149292247 Chr.7: 36855792-36855792 on Build GRCh38
rs938326627 Chr.7: 36855808-36855808 on Build GRCh38
rs892866005 Chr.7: 36855428-36855428 on Build GRCh38
rs778602057 Chr.7: 36855698-36855698 on Build GRCh38
rs771861381 Chr.7: 36855712-36855712 on Build GRCh38
rs753763165 Chr.7: 36855791-36855791 on Build GRCh38
rs143442077 Chr.7: 36855621-36855621 on Build GRCh38
rs371172929 Chr.7: 36855674-36855674 on Build GRCh38

View Full Product Details