Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 40001726-40002236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
UniGene
Hs.233552
Gene Chromosome Location
Chr.7: 39949501-40097134 on Build GRCh38
Transcripts
NM_003718.4 NM_031267.3 XM_011515597.2 XM_017012750.1 XM_017012751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5635517 c.2269C>T p.R757W Chr.7: 40001947-40001947 on Build GRCh38
6275339 c.2353+7G>T p.? Chr.7: 40002038-40002038 on Build GRCh38
4644799 c.2274G>A p.Q758Q Chr.7: 40001952-40001952 on Build GRCh38
6625377 c.2252G>A p.R751Q Chr.7: 40001930-40001930 on Build GRCh38
5482121 c.2289T>C p.S763S Chr.7: 40001967-40001967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570076650 Chr.7: 40002182-40002182 on Build GRCh38
rs200605545 Chr.7: 40002012-40002012 on Build GRCh38
rs1029554374 Chr.7: 40002198-40002198 on Build GRCh38
rs745528119 Chr.7: 40001918-40001919 on Build GRCh38
rs1023503658 Chr.7: 40002068-40002068 on Build GRCh38
rs55838651 Chr.7: 40001958-40001958 on Build GRCh38
rs887249522 Chr.7: 40001808-40001808 on Build GRCh38
rs17496388 Chr.7: 40002203-40002203 on Build GRCh38
rs756741273 Chr.7: 40001813-40001813 on Build GRCh38
rs370851800 Chr.7: 40001831-40001831 on Build GRCh38
rs548941535 Chr.7: 40002024-40002024 on Build GRCh38
rs202079430 Chr.7: 40002054-40002054 on Build GRCh38
rs750743833 Chr.7: 40001854-40001854 on Build GRCh38
rs556237734 Chr.7: 40002145-40002145 on Build GRCh38
rs540707119 Chr.7: 40001818-40001818 on Build GRCh38
rs931507542 Chr.7: 40001789-40001789 on Build GRCh38
rs755149077 Chr.7: 40001922-40001922 on Build GRCh38
rs756296289 Chr.7: 40001857-40001857 on Build GRCh38
rs769546328 Chr.7: 40001975-40001975 on Build GRCh38
rs1039218902 Chr.7: 40001776-40001776 on Build GRCh38
rs763213335 Chr.7: 40002085-40002085 on Build GRCh38
rs755709850 Chr.7: 40001939-40001940 on Build GRCh38
rs983785427 Chr.7: 40001767-40001769 on Build GRCh38
rs766621965 Chr.7: 40002059-40002059 on Build GRCh38
rs964772915 Chr.7: 40002190-40002190 on Build GRCh38
rs150544209 Chr.7: 40002114-40002114 on Build GRCh38
rs534423422 Chr.7: 40002066-40002066 on Build GRCh38
rs548030752 Chr.7: 40002078-40002078 on Build GRCh38
rs549473641 Chr.7: 40001947-40001947 on Build GRCh38
rs774968302 Chr.7: 40002041-40002041 on Build GRCh38
rs17496380 Chr.7: 40002196-40002196 on Build GRCh38
rs376578929 Chr.7: 40001785-40001785 on Build GRCh38
rs777432797 Chr.7: 40001959-40001959 on Build GRCh38
rs1048602176 Chr.7: 40001792-40001792 on Build GRCh38
rs780272203 Chr.7: 40001913-40001913 on Build GRCh38
rs945611373 Chr.7: 40002096-40002096 on Build GRCh38
rs202152509 Chr.7: 40002180-40002180 on Build GRCh38
rs149553872 Chr.7: 40002070-40002071 on Build GRCh38
rs866823436 Chr.7: 40002191-40002191 on Build GRCh38
rs746775910 Chr.7: 40001976-40001976 on Build GRCh38
rs987378937 Chr.7: 40002146-40002146 on Build GRCh38
rs1000647403 Chr.7: 40002103-40002103 on Build GRCh38
rs911792934 Chr.7: 40002151-40002151 on Build GRCh38
rs749126237 Chr.7: 40001750-40001750 on Build GRCh38
rs920355297 Chr.7: 40001786-40001786 on Build GRCh38
rs201512928 Chr.7: 40001926-40001926 on Build GRCh38
rs369643634 Chr.7: 40002006-40002006 on Build GRCh38
rs780713745 Chr.7: 40001877-40001877 on Build GRCh38
rs972648004 Chr.7: 40002195-40002195 on Build GRCh38
rs961309212 Chr.7: 40001884-40001884 on Build GRCh38
rs753983903 Chr.7: 40001919-40001919 on Build GRCh38
rs755233257 Chr.7: 40002073-40002073 on Build GRCh38
rs762303779 Chr.7: 40001835-40001835 on Build GRCh38
rs563811515 Chr.7: 40001852-40001852 on Build GRCh38
rs779670998 Chr.7: 40002047-40002050 on Build GRCh38
rs571947675 Chr.7: 40001769-40001769 on Build GRCh38
rs894883794 Chr.7: 40002144-40002144 on Build GRCh38
rs762323988 Chr.7: 40002048-40002048 on Build GRCh38
rs1033869699 Chr.7: 40002134-40002134 on Build GRCh38
rs568358047 Chr.7: 40002055-40002055 on Build GRCh38
rs893840381 Chr.7: 40002095-40002095 on Build GRCh38
rs769039387 Chr.7: 40002075-40002076 on Build GRCh38
rs539931564 Chr.7: 40002131-40002131 on Build GRCh38

View Full Product Details