Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43639158-43639654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COA1
Entrez Gene ID
55744
Gene Name
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases
C7orf44, MITRAC15
UniGene
Hs.268887
Gene Chromosome Location
Chr.7: 43608452-43729541 on Build GRCh38
Transcripts
NM_001321197.1 NM_001321198.1 NM_001321199.1 NM_001321200.1 NM_001321201.1 NM_018224.3 NR_047701.1 NR_135580.1 NR_135581.1 NR_135582.1 NR_135583.1 XM_005249807.4 XM_006715752.2 XM_017012414.1 XM_017012415.1 XM_017012416.1 XM_017012417.1 XM_017012418.1 XM_017012419.1 XM_017012421.1 XM_017012422.1 XM_017012423.1 XM_017012424.1 XM_017012425.1 XM_017012426.1 XM_017012427.1 XM_017012428.1 XM_017012429.1 XM_017012430.1 XM_017012431.1 XM_017012432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7116472 c.436G>A p.E146K Chr.7: 43639587-43639587 on Build GRCh38
6632061 c.380G>A p.G127D Chr.7: 43639643-43639643 on Build GRCh38
1089837 c.417C>T p.N139N Chr.7: 43639606-43639606 on Build GRCh38
1622876 c.432A>C p.K144N Chr.7: 43639591-43639591 on Build GRCh38
7161726 c.391C>T p.P131S Chr.7: 43639632-43639632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902887360 Chr.7: 43639217-43639217 on Build GRCh38
rs945900317 Chr.7: 43639502-43639502 on Build GRCh38
rs575978836 Chr.7: 43639354-43639354 on Build GRCh38
rs962748223 Chr.7: 43639516-43639516 on Build GRCh38
rs1044039 Chr.7: 43639398-43639398 on Build GRCh38
rs537301566 Chr.7: 43639589-43639589 on Build GRCh38
rs763387725 Chr.7: 43639605-43639605 on Build GRCh38
rs376700205 Chr.7: 43639572-43639572 on Build GRCh38
rs753785726 Chr.7: 43639552-43639552 on Build GRCh38
rs771752515 Chr.7: 43639575-43639575 on Build GRCh38
rs558753350 Chr.7: 43639403-43639403 on Build GRCh38
rs550280163 Chr.7: 43639290-43639294 on Build GRCh38
rs868303769 Chr.7: 43639604-43639604 on Build GRCh38
rs559137859 Chr.7: 43639632-43639632 on Build GRCh38
rs376882360 Chr.7: 43639624-43639624 on Build GRCh38
rs74583449 Chr.7: 43639228-43639233 on Build GRCh38
rs576928319 Chr.7: 43639473-43639473 on Build GRCh38
rs969953331 Chr.7: 43639298-43639298 on Build GRCh38
rs780158797 Chr.7: 43639534-43639534 on Build GRCh38
rs373930511 Chr.7: 43639606-43639606 on Build GRCh38
rs1011034081 Chr.7: 43639283-43639284 on Build GRCh38
rs756166648 Chr.7: 43639531-43639531 on Build GRCh38
rs921138909 Chr.7: 43639554-43639554 on Build GRCh38
rs958494103 Chr.7: 43639455-43639455 on Build GRCh38
rs951444701 Chr.7: 43639338-43639338 on Build GRCh38
rs1054161048 Chr.7: 43639230-43639231 on Build GRCh38
rs761977200 Chr.7: 43639616-43639616 on Build GRCh38
rs934340779 Chr.7: 43639619-43639619 on Build GRCh38
rs145808464 Chr.7: 43639220-43639225 on Build GRCh38
rs992627332 Chr.7: 43639468-43639468 on Build GRCh38
rs1023725752 Chr.7: 43639287-43639287 on Build GRCh38
rs200930949 Chr.7: 43639228-43639228 on Build GRCh38
rs138770666 Chr.7: 43639587-43639587 on Build GRCh38
rs76044398 Chr.7: 43639622-43639622 on Build GRCh38
rs745428154 Chr.7: 43639613-43639615 on Build GRCh38
rs750265343 Chr.7: 43639350-43639350 on Build GRCh38
rs770380189 Chr.7: 43639578-43639578 on Build GRCh38
rs776599475 Chr.7: 43639324-43639325 on Build GRCh38
rs775267037 Chr.7: 43639607-43639607 on Build GRCh38
rs1051441859 Chr.7: 43639215-43639215 on Build GRCh38
rs892858320 Chr.7: 43639263-43639264 on Build GRCh38
rs777189024 Chr.7: 43639577-43639577 on Build GRCh38
rs778711047 Chr.7: 43639568-43639568 on Build GRCh38
rs77224833 Chr.7: 43639511-43639511 on Build GRCh38
rs779666717 Chr.7: 43639434-43639434 on Build GRCh38
rs770746683 Chr.7: 43639286-43639286 on Build GRCh38
rs935983379 Chr.7: 43639628-43639628 on Build GRCh38
rs754823202 Chr.7: 43639561-43639561 on Build GRCh38
rs1035665088 Chr.7: 43639324-43639324 on Build GRCh38
rs1003838577 Chr.7: 43639303-43639304 on Build GRCh38
rs556929670 Chr.7: 43639200-43639200 on Build GRCh38

View Full Product Details