Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43787824-43788324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLVRA
Entrez Gene ID
644
Gene Name
biliverdin reductase A
Gene Aliases
BLVR, BVR, BVRA
UniGene
Hs.488143
Gene Chromosome Location
Chr.7: 43758153-43807342 on Build GRCh38
Transcripts
NM_000712.3 NM_001253823.1 XM_011515474.2 XM_017012520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
746843 c.67C>T p.R23W Chr.7: 43787958-43787958 on Build GRCh38
188813 c.131C>T p.S44L Chr.7: 43788022-43788022 on Build GRCh38
7357568 c.116T>A p.L39Q Chr.7: 43788007-43788007 on Build GRCh38
7303987 c.35T>C p.V12A Chr.7: 43787926-43787926 on Build GRCh38
5809537 c.44G>A p.G15D Chr.7: 43787935-43787935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138715080 Chr.7: 43787949-43787949 on Build GRCh38
rs751142620 Chr.7: 43787873-43787873 on Build GRCh38
rs778261680 Chr.7: 43787986-43787986 on Build GRCh38
rs17245911 Chr.7: 43787921-43787921 on Build GRCh38
rs200837553 Chr.7: 43788077-43788077 on Build GRCh38
rs201359042 Chr.7: 43787922-43787922 on Build GRCh38
rs539405061 Chr.7: 43788296-43788296 on Build GRCh38
rs767091448 Chr.7: 43788047-43788047 on Build GRCh38
rs1011986155 Chr.7: 43788072-43788072 on Build GRCh38
rs761588401 Chr.7: 43788046-43788046 on Build GRCh38
rs923872648 Chr.7: 43788209-43788209 on Build GRCh38
rs749890069 Chr.7: 43787955-43787955 on Build GRCh38
rs904739195 Chr.7: 43788116-43788116 on Build GRCh38
rs775434942 Chr.7: 43788017-43788017 on Build GRCh38
rs1026577333 Chr.7: 43788087-43788087 on Build GRCh38
rs762296012 Chr.7: 43788088-43788088 on Build GRCh38
rs1025476643 Chr.7: 43788259-43788259 on Build GRCh38
rs1023105069 Chr.7: 43787887-43787887 on Build GRCh38
rs771090611 Chr.7: 43787920-43787920 on Build GRCh38
rs755595790 Chr.7: 43787958-43787958 on Build GRCh38
rs772600465 Chr.7: 43788115-43788115 on Build GRCh38
rs974851043 Chr.7: 43788190-43788190 on Build GRCh38
rs954575433 Chr.7: 43788198-43788198 on Build GRCh38
rs369440039 Chr.7: 43788052-43788052 on Build GRCh38
rs11545804 Chr.7: 43787906-43787906 on Build GRCh38
rs747274098 Chr.7: 43787993-43787993 on Build GRCh38
rs535577617 Chr.7: 43788175-43788175 on Build GRCh38
rs765939375 Chr.7: 43788058-43788058 on Build GRCh38
rs530389071 Chr.7: 43788010-43788010 on Build GRCh38
rs747180057 Chr.7: 43787916-43787916 on Build GRCh38
rs145679201 Chr.7: 43787907-43787907 on Build GRCh38
rs387906596 Chr.7: 43788022-43788022 on Build GRCh38
rs1019821173 Chr.7: 43788233-43788233 on Build GRCh38
rs17239544 Chr.7: 43788170-43788170 on Build GRCh38
rs576624997 Chr.7: 43788076-43788076 on Build GRCh38
rs936342726 Chr.7: 43788014-43788014 on Build GRCh38
rs762650138 Chr.7: 43787866-43787866 on Build GRCh38
rs387906595 Chr.7: 43787943-43787943 on Build GRCh38
rs916234753 Chr.7: 43788238-43788238 on Build GRCh38
rs775346802 Chr.7: 43787936-43787936 on Build GRCh38
rs950907652 Chr.7: 43788135-43788135 on Build GRCh38
rs1003572724 Chr.7: 43788085-43788085 on Build GRCh38
rs936568343 Chr.7: 43788212-43788212 on Build GRCh38
rs879292423 Chr.7: 43788276-43788276 on Build GRCh38
rs978289817 Chr.7: 43788208-43788208 on Build GRCh38
rs756820050 Chr.7: 43787880-43787880 on Build GRCh38
rs780514948 Chr.7: 43787882-43787882 on Build GRCh38
rs564994689 Chr.7: 43787857-43787857 on Build GRCh38
rs765120206 Chr.7: 43787856-43787856 on Build GRCh38
rs139391855 Chr.7: 43788242-43788242 on Build GRCh38
rs2074795 Chr.7: 43788172-43788172 on Build GRCh38
rs368057250 Chr.7: 43787948-43787948 on Build GRCh38
rs1006237207 Chr.7: 43788163-43788163 on Build GRCh38
rs763832285 Chr.7: 43787944-43787944 on Build GRCh38
rs140584840 Chr.7: 43787904-43787904 on Build GRCh38
rs763748106 Chr.7: 43787867-43787867 on Build GRCh38
rs17245925 Chr.7: 43788054-43788054 on Build GRCh38
rs1006626912 Chr.7: 43788200-43788200 on Build GRCh38
rs984044750 Chr.7: 43787961-43787961 on Build GRCh38
rs771053277 Chr.7: 43787995-43787995 on Build GRCh38
rs575605650 Chr.7: 43788263-43788263 on Build GRCh38
rs923130648 Chr.7: 43788197-43788197 on Build GRCh38
rs17245918 Chr.7: 43788000-43788000 on Build GRCh38
rs535265391 Chr.7: 43788140-43788140 on Build GRCh38
rs183114141 Chr.7: 43788018-43788018 on Build GRCh38
rs953752957 Chr.7: 43787858-43787858 on Build GRCh38
rs140295067 Chr.7: 43787954-43787954 on Build GRCh38
rs965234195 Chr.7: 43788239-43788239 on Build GRCh38
rs758855722 Chr.7: 43787985-43787985 on Build GRCh38
rs779568837 Chr.7: 43787973-43787973 on Build GRCh38
rs753216810 Chr.7: 43787974-43787974 on Build GRCh38
rs1802849 Chr.7: 43787996-43787996 on Build GRCh38
rs754413382 Chr.7: 43787885-43787885 on Build GRCh38
rs906234700 Chr.7: 43788281-43788281 on Build GRCh38

View Full Product Details