Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44107462-44107952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AEBP1
Entrez Gene ID
165
Gene Name
AE binding protein 1
Gene Aliases
ACLP
UniGene
Hs.439463
Gene Chromosome Location
Chr.7: 44104320-44114565 on Build GRCh38
Transcripts
NM_001129.4 XM_011515162.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
485357 c.853G>A p.E285K Chr.7: 44107922-44107922 on Build GRCh38
3663207 c.642T>C p.H214H Chr.7: 44107485-44107485 on Build GRCh38
453090 c.697C>G p.L233V Chr.7: 44107658-44107658 on Build GRCh38
453091 c.817C>A p.P273T Chr.7: 44107886-44107886 on Build GRCh38
5430183 c.714G>A p.Q238Q Chr.7: 44107675-44107675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774499700 Chr.7: 44107891-44107891 on Build GRCh38
rs767627263 Chr.7: 44107723-44107723 on Build GRCh38
rs777002650 Chr.7: 44107496-44107496 on Build GRCh38
rs115538527 Chr.7: 44107616-44107616 on Build GRCh38
rs761597363 Chr.7: 44107690-44107690 on Build GRCh38
rs779461143 Chr.7: 44107740-44107740 on Build GRCh38
rs762250650 Chr.7: 44107503-44107503 on Build GRCh38
rs142763648 Chr.7: 44107673-44107673 on Build GRCh38
rs199888866 Chr.7: 44107521-44107521 on Build GRCh38
rs768249465 Chr.7: 44107748-44107748 on Build GRCh38
rs754231247 Chr.7: 44107733-44107733 on Build GRCh38
rs753695601 Chr.7: 44107528-44107528 on Build GRCh38
rs765260103 Chr.7: 44107534-44107534 on Build GRCh38
rs985310264 Chr.7: 44107800-44107800 on Build GRCh38
rs773075478 Chr.7: 44107699-44107699 on Build GRCh38
rs141311453 Chr.7: 44107700-44107700 on Build GRCh38
rs374676651 Chr.7: 44107921-44107921 on Build GRCh38
rs761972892 Chr.7: 44107887-44107887 on Build GRCh38
rs779044302 Chr.7: 44107820-44107820 on Build GRCh38
rs768864068 Chr.7: 44107495-44107495 on Build GRCh38
rs750822533 Chr.7: 44107901-44107901 on Build GRCh38
rs561259360 Chr.7: 44107914-44107914 on Build GRCh38
rs758958315 Chr.7: 44107512-44107512 on Build GRCh38
rs758385259 Chr.7: 44107633-44107633 on Build GRCh38
rs771478590 Chr.7: 44107679-44107679 on Build GRCh38
rs752164012 Chr.7: 44107905-44107905 on Build GRCh38
rs757626030 Chr.7: 44107738-44107738 on Build GRCh38
rs773779720 Chr.7: 44107510-44107510 on Build GRCh38
rs36040343 Chr.7: 44107594-44107594 on Build GRCh38
rs747658564 Chr.7: 44107827-44107827 on Build GRCh38
rs748420024 Chr.7: 44107668-44107668 on Build GRCh38
rs201137709 Chr.7: 44107615-44107615 on Build GRCh38
rs919846958 Chr.7: 44107788-44107788 on Build GRCh38
rs368747482 Chr.7: 44107867-44107867 on Build GRCh38
rs182097426 Chr.7: 44107625-44107625 on Build GRCh38
rs375816630 Chr.7: 44107753-44107753 on Build GRCh38
rs1013697124 Chr.7: 44107929-44107929 on Build GRCh38
rs996248410 Chr.7: 44107599-44107599 on Build GRCh38
rs766981749 Chr.7: 44107902-44107902 on Build GRCh38
rs773178679 Chr.7: 44107892-44107894 on Build GRCh38
rs200279382 Chr.7: 44107824-44107824 on Build GRCh38
rs754064375 Chr.7: 44107818-44107818 on Build GRCh38
rs867536357 Chr.7: 44107847-44107847 on Build GRCh38
rs746426034 Chr.7: 44107743-44107743 on Build GRCh38
rs544375149 Chr.7: 44107906-44107906 on Build GRCh38
rs912983756 Chr.7: 44107888-44107888 on Build GRCh38
rs2537188 Chr.7: 44107886-44107886 on Build GRCh38
rs200209047 Chr.7: 44107759-44107759 on Build GRCh38
rs201157904 Chr.7: 44107729-44107729 on Build GRCh38
rs771996997 Chr.7: 44107872-44107872 on Build GRCh38
rs777463013 Chr.7: 44107854-44107854 on Build GRCh38
rs749706932 Chr.7: 44107678-44107678 on Build GRCh38
rs755639223 Chr.7: 44107844-44107844 on Build GRCh38
rs755121488 Chr.7: 44107651-44107651 on Build GRCh38
rs775886011 Chr.7: 44107496-44107496 on Build GRCh38
rs771439583 Chr.7: 44107597-44107597 on Build GRCh38
rs772807772 Chr.7: 44107769-44107769 on Build GRCh38
rs755207231 Chr.7: 44107579-44107579 on Build GRCh38
rs750288301 Chr.7: 44107632-44107632 on Build GRCh38
rs770206330 Chr.7: 44107592-44107592 on Build GRCh38
rs756870217 Chr.7: 44107917-44107917 on Build GRCh38
rs766336537 Chr.7: 44107706-44107706 on Build GRCh38
rs762798082 Chr.7: 44107777-44107777 on Build GRCh38
rs890895132 Chr.7: 44107568-44107568 on Build GRCh38
rs112591957 Chr.7: 44107808-44107808 on Build GRCh38
rs760817355 Chr.7: 44107811-44107811 on Build GRCh38
rs777183157 Chr.7: 44107908-44107908 on Build GRCh38
rs148932183 Chr.7: 44107645-44107645 on Build GRCh38
rs764369368 Chr.7: 44107817-44107817 on Build GRCh38
rs746524312 Chr.7: 44107682-44107682 on Build GRCh38
rs758384725 Chr.7: 44107558-44107558 on Build GRCh38
rs928500448 Chr.7: 44107687-44107687 on Build GRCh38
rs749468361 Chr.7: 44107584-44107585 on Build GRCh38
rs775469777 Chr.7: 44107874-44107874 on Build GRCh38
rs992139534 Chr.7: 44107487-44107487 on Build GRCh38
rs773689779 Chr.7: 44107593-44107593 on Build GRCh38
rs780872112 Chr.7: 44107758-44107758 on Build GRCh38
rs1021588353 Chr.7: 44107576-44107576 on Build GRCh38
rs748511797 Chr.7: 44107581-44107581 on Build GRCh38
rs376465080 Chr.7: 44107580-44107580 on Build GRCh38
rs201963190 Chr.7: 44107694-44107694 on Build GRCh38
rs774791727 Chr.7: 44107681-44107681 on Build GRCh38
rs747297608 Chr.7: 44107494-44107494 on Build GRCh38
rs1015059460 Chr.7: 44107843-44107843 on Build GRCh38
rs372015098 Chr.7: 44107661-44107661 on Build GRCh38
rs745744525 Chr.7: 44107871-44107871 on Build GRCh38
rs866170571 Chr.7: 44107865-44107865 on Build GRCh38
rs1053005862 Chr.7: 44107732-44107732 on Build GRCh38
rs760792254 Chr.7: 44107879-44107879 on Build GRCh38
rs374508688 Chr.7: 44107559-44107559 on Build GRCh38
rs747152934 Chr.7: 44107561-44107561 on Build GRCh38
rs367737639 Chr.7: 44107639-44107639 on Build GRCh38
rs748935568 Chr.7: 44107859-44107859 on Build GRCh38
rs774319124 Chr.7: 44107794-44107794 on Build GRCh38
rs755070851 Chr.7: 44107764-44107764 on Build GRCh38
rs752782990 Chr.7: 44107804-44107804 on Build GRCh38
rs751513362 Chr.7: 44107718-44107718 on Build GRCh38
rs574188983 Chr.7: 44107821-44107821 on Build GRCh38
rs553811931 Chr.7: 44107683-44107683 on Build GRCh38
rs544420240 Chr.7: 44107868-44107868 on Build GRCh38
rs777939416 Chr.7: 44107730-44107730 on Build GRCh38
rs35147172 Chr.7: 44107566-44107566 on Build GRCh38
rs759594768 Chr.7: 44107719-44107719 on Build GRCh38
rs979852412 Chr.7: 44107685-44107685 on Build GRCh38
rs780125197 Chr.7: 44107485-44107485 on Build GRCh38
rs764128287 Chr.7: 44107882-44107882 on Build GRCh38
rs368244699 Chr.7: 44107605-44107605 on Build GRCh38
rs759504950 Chr.7: 44107803-44107803 on Build GRCh38
rs768974633 Chr.7: 44107853-44107855 on Build GRCh38
rs758987524 Chr.7: 44107823-44107823 on Build GRCh38
rs1043465087 Chr.7: 44107750-44107750 on Build GRCh38
rs750430662 Chr.7: 44107551-44107551 on Build GRCh38
rs145045450 Chr.7: 44107822-44107822 on Build GRCh38
rs113528346 Chr.7: 44107807-44107807 on Build GRCh38
rs769492722 Chr.7: 44107762-44107762 on Build GRCh38
rs201736457 Chr.7: 44107527-44107527 on Build GRCh38
rs151068311 Chr.7: 44107684-44107684 on Build GRCh38
rs374348558 Chr.7: 44107520-44107520 on Build GRCh38
rs111934840 Chr.7: 44107701-44107701 on Build GRCh38
rs770683133 Chr.7: 44107790-44107790 on Build GRCh38
rs756442121 Chr.7: 44107669-44107669 on Build GRCh38
rs375315010 Chr.7: 44107672-44107672 on Build GRCh38
rs200342317 Chr.7: 44107631-44107631 on Build GRCh38

View Full Product Details