Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44571419-44571918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX56
Entrez Gene ID
54606
Gene Name
DEAD-box helicase 56
Gene Aliases
DDX21, DDX26, NOH61
UniGene
Hs.654762
Gene Chromosome Location
Chr.7: 44565417-44574538 on Build GRCh38
Transcripts
NM_001257189.1 NM_019082.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5472412 c.772C>T p.R258W Chr.7: 44571610-44571610 on Build GRCh38
7245595 c.721G>C p.E241Q Chr.7: 44571661-44571661 on Build GRCh38
3881259 c.729C>T p.D243D Chr.7: 44571653-44571653 on Build GRCh38
5450930 c.764C>T p.S255L Chr.7: 44571618-44571618 on Build GRCh38
6641988 c.817C>T p.R273W Chr.7: 44571565-44571565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775963725 Chr.7: 44571450-44571450 on Build GRCh38
rs753581119 Chr.7: 44571895-44571895 on Build GRCh38
rs994901999 Chr.7: 44571874-44571874 on Build GRCh38
rs577267203 Chr.7: 44571605-44571605 on Build GRCh38
rs945819203 Chr.7: 44571597-44571597 on Build GRCh38
rs200906446 Chr.7: 44571718-44571718 on Build GRCh38
rs141491106 Chr.7: 44571647-44571647 on Build GRCh38
rs967071477 Chr.7: 44571848-44571848 on Build GRCh38
rs138531895 Chr.7: 44571640-44571640 on Build GRCh38
rs138175613 Chr.7: 44571862-44571862 on Build GRCh38
rs1001430797 Chr.7: 44571641-44571641 on Build GRCh38
rs763840415 Chr.7: 44571507-44571507 on Build GRCh38
rs746510981 Chr.7: 44571643-44571643 on Build GRCh38
rs41279636 Chr.7: 44571558-44571558 on Build GRCh38
rs754183846 Chr.7: 44571725-44571725 on Build GRCh38
rs372992961 Chr.7: 44571760-44571760 on Build GRCh38
rs755220533 Chr.7: 44571551-44571551 on Build GRCh38
rs897834777 Chr.7: 44571739-44571739 on Build GRCh38
rs759621867 Chr.7: 44571700-44571700 on Build GRCh38
rs200177535 Chr.7: 44571499-44571499 on Build GRCh38
rs776086145 Chr.7: 44571653-44571653 on Build GRCh38
rs751786562 Chr.7: 44571539-44571539 on Build GRCh38
rs373548733 Chr.7: 44571784-44571784 on Build GRCh38
rs537020657 Chr.7: 44571705-44571705 on Build GRCh38
rs925716268 Chr.7: 44571887-44571887 on Build GRCh38
rs746566080 Chr.7: 44571572-44571572 on Build GRCh38
rs773024707 Chr.7: 44571676-44571676 on Build GRCh38
rs41279639 Chr.7: 44571636-44571636 on Build GRCh38
rs747856771 Chr.7: 44571665-44571665 on Build GRCh38
rs766456412 Chr.7: 44571610-44571610 on Build GRCh38
rs200165658 Chr.7: 44571603-44571603 on Build GRCh38
rs750936470 Chr.7: 44571740-44571740 on Build GRCh38
rs755165770 Chr.7: 44571626-44571626 on Build GRCh38
rs922101918 Chr.7: 44571570-44571570 on Build GRCh38
rs779714226 Chr.7: 44571812-44571812 on Build GRCh38
rs1004871919 Chr.7: 44571601-44571601 on Build GRCh38
rs762889439 Chr.7: 44571687-44571687 on Build GRCh38
rs765074860 Chr.7: 44571513-44571513 on Build GRCh38
rs769089417 Chr.7: 44571446-44571446 on Build GRCh38
rs953529602 Chr.7: 44571515-44571515 on Build GRCh38
rs779390948 Chr.7: 44571738-44571738 on Build GRCh38
rs752966392 Chr.7: 44571634-44571634 on Build GRCh38
rs145895899 Chr.7: 44571609-44571609 on Build GRCh38
rs932153168 Chr.7: 44571777-44571777 on Build GRCh38
rs1027760102 Chr.7: 44571750-44571750 on Build GRCh38
rs200686546 Chr.7: 44571574-44571574 on Build GRCh38
rs776405497 Chr.7: 44571580-44571580 on Build GRCh38
rs749987638 Chr.7: 44571729-44571729 on Build GRCh38
rs748585109 Chr.7: 44571475-44571475 on Build GRCh38
rs140702133 Chr.7: 44571496-44571496 on Build GRCh38
rs780782944 Chr.7: 44571766-44571766 on Build GRCh38
rs890431512 Chr.7: 44571588-44571588 on Build GRCh38
rs766405612 Chr.7: 44571694-44571694 on Build GRCh38
rs766584541 Chr.7: 44571525-44571525 on Build GRCh38
rs370069185 Chr.7: 44571782-44571782 on Build GRCh38
rs754433166 Chr.7: 44571742-44571742 on Build GRCh38
rs988402450 Chr.7: 44571791-44571791 on Build GRCh38
rs374060771 Chr.7: 44571688-44571689 on Build GRCh38
rs914104689 Chr.7: 44571830-44571830 on Build GRCh38
rs375038290 Chr.7: 44571573-44571573 on Build GRCh38
rs773826192 Chr.7: 44571479-44571479 on Build GRCh38
rs775066678 Chr.7: 44571498-44571498 on Build GRCh38
rs367882554 Chr.7: 44571510-44571510 on Build GRCh38
rs767845229 Chr.7: 44571633-44571633 on Build GRCh38
rs928297580 Chr.7: 44571447-44571447 on Build GRCh38
rs988432489 Chr.7: 44571662-44571662 on Build GRCh38
rs217375 Chr.7: 44571478-44571478 on Build GRCh38
rs758962465 Chr.7: 44571492-44571492 on Build GRCh38
rs1036330484 Chr.7: 44571861-44571861 on Build GRCh38
rs781613764 Chr.7: 44571552-44571552 on Build GRCh38
rs375188739 Chr.7: 44571769-44571769 on Build GRCh38
rs759194163 Chr.7: 44571743-44571744 on Build GRCh38
rs750375632 Chr.7: 44571516-44571516 on Build GRCh38
rs774495551 Chr.7: 44571695-44571695 on Build GRCh38
rs988466969 Chr.7: 44571497-44571497 on Build GRCh38
rs751665943 Chr.7: 44571612-44571612 on Build GRCh38
rs201691567 Chr.7: 44571668-44571668 on Build GRCh38
rs561304472 Chr.7: 44571474-44571474 on Build GRCh38
rs187799703 Chr.7: 44571786-44571786 on Build GRCh38
rs372022006 Chr.7: 44571565-44571565 on Build GRCh38
rs200964208 Chr.7: 44571755-44571755 on Build GRCh38
rs373129289 Chr.7: 44571564-44571564 on Build GRCh38
rs1052462686 Chr.7: 44571587-44571587 on Build GRCh38
rs201308009 Chr.7: 44571559-44571559 on Build GRCh38
rs968405518 Chr.7: 44571758-44571758 on Build GRCh38
rs758349449 Chr.7: 44571518-44571518 on Build GRCh38
rs753603280 Chr.7: 44571509-44571509 on Build GRCh38
rs776190071 Chr.7: 44571586-44571586 on Build GRCh38
rs369278330 Chr.7: 44571753-44571753 on Build GRCh38
rs767650743 Chr.7: 44571704-44571704 on Build GRCh38

View Full Product Details