Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45068328-45068824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2
Entrez Gene ID
83605
Gene Name
CCM2 scaffolding protein
Gene Aliases
C7orf22, OSM, PP10187
UniGene
Hs.148272
Gene Chromosome Location
Chr.7: 44999746-45076470 on Build GRCh38
Transcripts
NM_001029835.2 NM_001167934.1 NM_001167935.1 NM_031443.3 NR_030770.1 XM_006715785.3 XM_006715786.3 XM_011515561.2 XM_011515562.1 XM_011515563.2 XM_011515564.2 XM_017012671.1 XM_017012672.1 XM_017012673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3923681 c.554C>T p.S185F Chr.7: 45068461-45068461 on Build GRCh38
7270676 c.536-1G>T p.? Chr.7: 45068442-45068442 on Build GRCh38
5078324 c.576G>T p.A192A Chr.7: 45068483-45068483 on Build GRCh38
6020495 c.596G>C p.S199T Chr.7: 45068503-45068503 on Build GRCh38
3263242 c.625G>C p.G209R Chr.7: 45068532-45068532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145671290 Chr.7: 45068519-45068519 on Build GRCh38
rs774896337 Chr.7: 45068513-45068513 on Build GRCh38
rs574711870 Chr.7: 45068367-45068367 on Build GRCh38
rs748895941 Chr.7: 45068696-45068696 on Build GRCh38
rs528670285 Chr.7: 45068710-45068710 on Build GRCh38
rs766127952 Chr.7: 45068528-45068528 on Build GRCh38
rs751420337 Chr.7: 45068529-45068529 on Build GRCh38
rs779850799 Chr.7: 45068459-45068459 on Build GRCh38
rs951910506 Chr.7: 45068561-45068561 on Build GRCh38
rs907156998 Chr.7: 45068679-45068679 on Build GRCh38
rs948068706 Chr.7: 45068633-45068633 on Build GRCh38
rs748247533 Chr.7: 45068499-45068538 on Build GRCh38
rs945166100 Chr.7: 45068706-45068706 on Build GRCh38
rs936845344 Chr.7: 45068756-45068756 on Build GRCh38
rs373239614 Chr.7: 45068523-45068523 on Build GRCh38
rs148547810 Chr.7: 45068482-45068482 on Build GRCh38
rs763666971 Chr.7: 45068397-45068397 on Build GRCh38
rs776079371 Chr.7: 45068520-45068520 on Build GRCh38
rs761271260 Chr.7: 45068616-45068616 on Build GRCh38
rs961491758 Chr.7: 45068466-45068466 on Build GRCh38
rs752071985 Chr.7: 45068394-45068394 on Build GRCh38
rs774845080 Chr.7: 45068592-45068592 on Build GRCh38
rs377135433 Chr.7: 45068590-45068590 on Build GRCh38
rs769929401 Chr.7: 45068503-45068503 on Build GRCh38
rs1044264866 Chr.7: 45068667-45068667 on Build GRCh38
rs201140198 Chr.7: 45068597-45068597 on Build GRCh38
rs749998555 Chr.7: 45068524-45068524 on Build GRCh38
rs925459410 Chr.7: 45068720-45068720 on Build GRCh38
rs964738207 Chr.7: 45068434-45068434 on Build GRCh38
rs1038199581 Chr.7: 45068568-45068568 on Build GRCh38
rs369353708 Chr.7: 45068461-45068461 on Build GRCh38
rs1011288316 Chr.7: 45068800-45068800 on Build GRCh38
rs904081511 Chr.7: 45068351-45068351 on Build GRCh38
rs984654716 Chr.7: 45068580-45068581 on Build GRCh38
rs780931748 Chr.7: 45068537-45068537 on Build GRCh38
rs964341276 Chr.7: 45068618-45068619 on Build GRCh38
rs771346534 Chr.7: 45068589-45068589 on Build GRCh38
rs764741384 Chr.7: 45068408-45068408 on Build GRCh38
rs1031505009 Chr.7: 45068774-45068774 on Build GRCh38
rs373136857 Chr.7: 45068505-45068505 on Build GRCh38
rs138789643 Chr.7: 45068390-45068390 on Build GRCh38
rs137852843 Chr.7: 45068563-45068563 on Build GRCh38
rs557108717 Chr.7: 45068436-45068436 on Build GRCh38
rs758091168 Chr.7: 45068526-45068526 on Build GRCh38
rs141353947 Chr.7: 45068521-45068521 on Build GRCh38
rs775824096 Chr.7: 45068599-45068599 on Build GRCh38
rs749490859 Chr.7: 45068569-45068569 on Build GRCh38
rs975731234 Chr.7: 45068621-45068621 on Build GRCh38
rs187724008 Chr.7: 45068506-45068506 on Build GRCh38
rs200358025 Chr.7: 45068538-45068538 on Build GRCh38
rs753358163 Chr.7: 45068407-45068407 on Build GRCh38
rs13231913 Chr.7: 45068366-45068366 on Build GRCh38
rs756106358 Chr.7: 45068547-45068547 on Build GRCh38
rs777930252 Chr.7: 45068555-45068555 on Build GRCh38
rs760310677 Chr.7: 45068781-45068781 on Build GRCh38
rs754893851 Chr.7: 45068532-45068532 on Build GRCh38
rs772611495 Chr.7: 45068671-45068671 on Build GRCh38
rs760117074 Chr.7: 45068518-45068518 on Build GRCh38
rs750097466 Chr.7: 45068419-45068419 on Build GRCh38
rs771372937 Chr.7: 45068512-45068512 on Build GRCh38
rs933463775 Chr.7: 45068487-45068487 on Build GRCh38
rs754943918 Chr.7: 45068468-45068468 on Build GRCh38
rs1002642416 Chr.7: 45068760-45068760 on Build GRCh38
rs760170287 Chr.7: 45068395-45068395 on Build GRCh38
rs990219871 Chr.7: 45068380-45068380 on Build GRCh38
rs377719633 Chr.7: 45068530-45068530 on Build GRCh38
rs748232087 Chr.7: 45068488-45068488 on Build GRCh38
rs772492249 Chr.7: 45068598-45068598 on Build GRCh38

View Full Product Details