Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45074186-45074690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2
Entrez Gene ID
83605
Gene Name
CCM2 scaffolding protein
Gene Aliases
C7orf22, OSM, PP10187
UniGene
Hs.148272
Gene Chromosome Location
Chr.7: 44999746-45076470 on Build GRCh38
Transcripts
NM_001029835.2 NM_001167934.1 NM_001167935.1 NM_031443.3 NR_030770.1 XM_006715785.3 XM_006715786.3 XM_011515561.2 XM_011515562.1 XM_011515563.2 XM_011515564.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726589 c.1086C>T p.L362L Chr.7: 45074377-45074377 on Build GRCh38
5872768 c.1097G>A p.S366N Chr.7: 45074388-45074388 on Build GRCh38
6622880 c.982C>T p.R328C Chr.7: 45074273-45074273 on Build GRCh38
1450982 c.1090G>A p.G364R Chr.7: 45074381-45074381 on Build GRCh38
1450983 c.1099C>T p.R367C Chr.7: 45074390-45074390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772690362 Chr.7: 45074345-45074345 on Build GRCh38
rs964429862 Chr.7: 45074427-45074427 on Build GRCh38
rs150428392 Chr.7: 45074334-45074334 on Build GRCh38
rs763682175 Chr.7: 45074224-45074224 on Build GRCh38
rs760062640 Chr.7: 45074221-45074221 on Build GRCh38
rs963331929 Chr.7: 45074455-45074455 on Build GRCh38
rs1053265471 Chr.7: 45074624-45074624 on Build GRCh38
rs766187723 Chr.7: 45074299-45074299 on Build GRCh38
rs751441309 Chr.7: 45074398-45074398 on Build GRCh38
rs929116812 Chr.7: 45074651-45074651 on Build GRCh38
rs540000312 Chr.7: 45074212-45074212 on Build GRCh38
rs533279125 Chr.7: 45074239-45074239 on Build GRCh38
rs535112809 Chr.7: 45074280-45074280 on Build GRCh38
rs980043440 Chr.7: 45074557-45074557 on Build GRCh38
rs776469228 Chr.7: 45074463-45074463 on Build GRCh38
rs928432467 Chr.7: 45074569-45074569 on Build GRCh38
rs113529574 Chr.7: 45074486-45074486 on Build GRCh38
rs762043544 Chr.7: 45074426-45074426 on Build GRCh38
rs760137287 Chr.7: 45074270-45074270 on Build GRCh38
rs1046457226 Chr.7: 45074628-45074628 on Build GRCh38
rs1042776746 Chr.7: 45074634-45074634 on Build GRCh38
rs201215825 Chr.7: 45074416-45074416 on Build GRCh38
rs561714324 Chr.7: 45074216-45074216 on Build GRCh38
rs973416766 Chr.7: 45074460-45074460 on Build GRCh38
rs1047567876 Chr.7: 45074665-45074665 on Build GRCh38
rs983914121 Chr.7: 45074419-45074419 on Build GRCh38
rs145332010 Chr.7: 45074615-45074615 on Build GRCh38
rs762647976 Chr.7: 45074384-45074384 on Build GRCh38
rs756276859 Chr.7: 45074324-45074324 on Build GRCh38
rs925536551 Chr.7: 45074591-45074591 on Build GRCh38
rs779374000 Chr.7: 45074343-45074343 on Build GRCh38
rs778075744 Chr.7: 45074331-45074331 on Build GRCh38
rs2289370 Chr.7: 45074490-45074490 on Build GRCh38
rs558327316 Chr.7: 45074485-45074485 on Build GRCh38
rs766059576 Chr.7: 45074385-45074385 on Build GRCh38
rs761242413 Chr.7: 45074353-45074353 on Build GRCh38
rs749607359 Chr.7: 45074265-45074265 on Build GRCh38
rs771358794 Chr.7: 45074371-45074371 on Build GRCh38
rs754986248 Chr.7: 45074315-45074315 on Build GRCh38
rs867158013 Chr.7: 45074261-45074261 on Build GRCh38
rs754081301 Chr.7: 45074437-45074437 on Build GRCh38
rs750790401 Chr.7: 45074507-45074507 on Build GRCh38
rs546816313 Chr.7: 45074323-45074323 on Build GRCh38
rs757617442 Chr.7: 45074449-45074449 on Build GRCh38
rs751551628 Chr.7: 45074245-45074245 on Build GRCh38
rs966836655 Chr.7: 45074494-45074494 on Build GRCh38
rs532261504 Chr.7: 45074635-45074637 on Build GRCh38
rs769322559 Chr.7: 45074369-45074369 on Build GRCh38
rs938389428 Chr.7: 45074231-45074231 on Build GRCh38
rs373072247 Chr.7: 45074267-45074267 on Build GRCh38
rs141561098 Chr.7: 45074219-45074219 on Build GRCh38
rs758199311 Chr.7: 45074240-45074240 on Build GRCh38
rs997263357 Chr.7: 45074465-45074465 on Build GRCh38
rs775873072 Chr.7: 45074351-45074351 on Build GRCh38
rs551511374 Chr.7: 45074263-45074263 on Build GRCh38
rs556953955 Chr.7: 45074380-45074380 on Build GRCh38
rs766302113 Chr.7: 45074441-45074441 on Build GRCh38
rs764317578 Chr.7: 45074435-45074435 on Build GRCh38
rs779895886 Chr.7: 45074244-45074244 on Build GRCh38
rs761438201 Chr.7: 45074277-45074277 on Build GRCh38
rs1005856659 Chr.7: 45074356-45074356 on Build GRCh38
rs754966956 Chr.7: 45074250-45074250 on Build GRCh38
rs781129836 Chr.7: 45074317-45074317 on Build GRCh38
rs759379755 Chr.7: 45074414-45074414 on Build GRCh38
rs756971672 Chr.7: 45074233-45074233 on Build GRCh38
rs958547202 Chr.7: 45074606-45074606 on Build GRCh38
rs140520179 Chr.7: 45074308-45074308 on Build GRCh38
rs372739455 Chr.7: 45074274-45074274 on Build GRCh38
rs544192218 Chr.7: 45074659-45074659 on Build GRCh38
rs948608056 Chr.7: 45074516-45074516 on Build GRCh38
rs2289372 Chr.7: 45074627-45074627 on Build GRCh38
rs907044131 Chr.7: 45074320-45074320 on Build GRCh38
rs562742746 Chr.7: 45074228-45074228 on Build GRCh38
rs781385360 Chr.7: 45074259-45074259 on Build GRCh38
rs1030877825 Chr.7: 45074350-45074350 on Build GRCh38
rs887645827 Chr.7: 45074668-45074668 on Build GRCh38
rs750076141 Chr.7: 45074282-45074282 on Build GRCh38
rs190686229 Chr.7: 45074420-45074420 on Build GRCh38
rs746355834 Chr.7: 45074344-45074344 on Build GRCh38
rs181315668 Chr.7: 45074220-45074220 on Build GRCh38
rs2289371 Chr.7: 45074625-45074625 on Build GRCh38
rs112504276 Chr.7: 45074338-45074338 on Build GRCh38

View Full Product Details