Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48192867-48193367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA13
Entrez Gene ID
154664
Gene Name
ATP binding cassette subfamily A member 13
Gene Aliases
-
UniGene
Hs.226568
Gene Chromosome Location
Chr.7: 48171424-48647497 on Build GRCh38
Transcripts
NM_152701.4 XM_011515130.2 XM_011515131.2 XM_011515132.2 XM_011515133.2 XM_011515134.2 XM_011515137.2 XM_011515138.2 XM_011515139.2 XM_011515141.2 XM_011515142.2 XM_011515143.2 XM_011515144.2 XM_011515145.2 XM_011515146.2 XM_011515147.2 XM_011515148.2 XM_011515149.2 XM_017011767.1 XM_017011768.1 XM_017011769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6430624 c.107T>C p.L36P Chr.7: 48192996-48192996 on Build GRCh38
6382739 c.85G>T p.E29* Chr.7: 48192974-48192974 on Build GRCh38
4596796 c.131G>A p.R44H Chr.7: 48193020-48193020 on Build GRCh38
5809275 c.70-4C>T p.? Chr.7: 48192955-48192955 on Build GRCh38
1090230 c.157G>A p.D53N Chr.7: 48193046-48193046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748051084 Chr.7: 48193226-48193226 on Build GRCh38
rs1049802387 Chr.7: 48193263-48193263 on Build GRCh38
rs907301116 Chr.7: 48193293-48193293 on Build GRCh38
rs990505998 Chr.7: 48192917-48192917 on Build GRCh38
rs1043612404 Chr.7: 48193132-48193132 on Build GRCh38
rs757124318 Chr.7: 48192999-48192999 on Build GRCh38
rs867958801 Chr.7: 48193131-48193131 on Build GRCh38
rs914932974 Chr.7: 48192920-48192920 on Build GRCh38
rs1034432695 Chr.7: 48193308-48193308 on Build GRCh38
rs887808265 Chr.7: 48193091-48193091 on Build GRCh38
rs145368784 Chr.7: 48193203-48193203 on Build GRCh38
rs772387525 Chr.7: 48193075-48193075 on Build GRCh38
rs149161491 Chr.7: 48193227-48193227 on Build GRCh38
rs370921866 Chr.7: 48192983-48192983 on Build GRCh38
rs559638720 Chr.7: 48192934-48192935 on Build GRCh38
rs566894266 Chr.7: 48193020-48193020 on Build GRCh38
rs779052168 Chr.7: 48193042-48193042 on Build GRCh38
rs750544056 Chr.7: 48192914-48192914 on Build GRCh38
rs1011824492 Chr.7: 48192950-48192950 on Build GRCh38
rs1014934355 Chr.7: 48193238-48193238 on Build GRCh38
rs755871574 Chr.7: 48192928-48192928 on Build GRCh38
rs182813684 Chr.7: 48193119-48193119 on Build GRCh38
rs187453025 Chr.7: 48193166-48193166 on Build GRCh38
rs991181128 Chr.7: 48193143-48193143 on Build GRCh38
rs977722994 Chr.7: 48193175-48193175 on Build GRCh38
rs560240987 Chr.7: 48193321-48193321 on Build GRCh38
rs961322073 Chr.7: 48193021-48193021 on Build GRCh38
rs904596220 Chr.7: 48192960-48192960 on Build GRCh38
rs189981035 Chr.7: 48193205-48193205 on Build GRCh38
rs775546437 Chr.7: 48193080-48193080 on Build GRCh38
rs758441818 Chr.7: 48193264-48193264 on Build GRCh38
rs535885402 Chr.7: 48193104-48193104 on Build GRCh38
rs994138475 Chr.7: 48193255-48193255 on Build GRCh38
rs375210341 Chr.7: 48193044-48193044 on Build GRCh38
rs961581876 Chr.7: 48193251-48193251 on Build GRCh38
rs1056820419 Chr.7: 48193088-48193088 on Build GRCh38
rs919805254 Chr.7: 48193057-48193057 on Build GRCh38
rs972945018 Chr.7: 48193048-48193048 on Build GRCh38
rs143298995 Chr.7: 48193280-48193280 on Build GRCh38
rs959489106 Chr.7: 48193324-48193324 on Build GRCh38
rs569853882 Chr.7: 48193129-48193129 on Build GRCh38
rs952578177 Chr.7: 48193289-48193289 on Build GRCh38
rs1035228522 Chr.7: 48193125-48193125 on Build GRCh38
rs535147792 Chr.7: 48193236-48193236 on Build GRCh38
rs979847704 Chr.7: 48193070-48193070 on Build GRCh38
rs967722195 Chr.7: 48193149-48193149 on Build GRCh38
rs888007720 Chr.7: 48193270-48193270 on Build GRCh38
rs749815623 Chr.7: 48192934-48192934 on Build GRCh38
rs996870610 Chr.7: 48193067-48193067 on Build GRCh38
rs996249221 Chr.7: 48193202-48193202 on Build GRCh38
rs797010644 Chr.7: 48192934-48192935 on Build GRCh38
rs186242785 Chr.7: 48192907-48192907 on Build GRCh38
rs1003387642 Chr.7: 48193123-48193124 on Build GRCh38
rs1004821588 Chr.7: 48193097-48193097 on Build GRCh38
rs754755975 Chr.7: 48193076-48193076 on Build GRCh38
rs766686036 Chr.7: 48193012-48193012 on Build GRCh38
rs183048157 Chr.7: 48193303-48193303 on Build GRCh38
rs769149581 Chr.7: 48192933-48192934 on Build GRCh38
rs904714534 Chr.7: 48193200-48193200 on Build GRCh38
rs1020242935 Chr.7: 48193144-48193171 on Build GRCh38
rs745929108 Chr.7: 48193059-48193059 on Build GRCh38
rs913438932 Chr.7: 48193121-48193121 on Build GRCh38
rs1014981516 Chr.7: 48193100-48193100 on Build GRCh38
rs191026398 Chr.7: 48192948-48192948 on Build GRCh38
rs1011410445 Chr.7: 48192921-48192934 on Build GRCh38
rs566261966 Chr.7: 48193319-48193319 on Build GRCh38
rs546968040 Chr.7: 48193019-48193019 on Build GRCh38

View Full Product Details