Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55018824-55019363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201283.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652229 c.84G>T p.K28N Chr.7: 55019361-55019361 on Build GRCh38
6953026 c.70G>T p.A24S Chr.7: 55019347-55019347 on Build GRCh38
402884 c.63G>T p.A21A Chr.7: 55019340-55019340 on Build GRCh38
6652254 c.46G>A p.A16T Chr.7: 55019323-55019323 on Build GRCh38
6916445 c.14G>C p.G5A Chr.7: 55019291-55019291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010599071 Chr.7: 55019316-55019316 on Build GRCh38
rs567894670 Chr.7: 55019315-55019315 on Build GRCh38
rs888955272 Chr.7: 55018947-55018947 on Build GRCh38
rs377241852 Chr.7: 55018842-55018842 on Build GRCh38
rs749433287 Chr.7: 55019285-55019285 on Build GRCh38
rs933251199 Chr.7: 55018905-55018905 on Build GRCh38
rs923752281 Chr.7: 55018896-55018896 on Build GRCh38
rs761183109 Chr.7: 55019296-55019296 on Build GRCh38
rs754259847 Chr.7: 55019299-55019299 on Build GRCh38
rs781240283 Chr.7: 55019257-55019257 on Build GRCh38
rs555477367 Chr.7: 55019215-55019215 on Build GRCh38
rs369581368 Chr.7: 55019298-55019298 on Build GRCh38
rs866678715 Chr.7: 55019111-55019111 on Build GRCh38
rs911854407 Chr.7: 55019072-55019072 on Build GRCh38
rs895170118 Chr.7: 55019004-55019004 on Build GRCh38
rs866177915 Chr.7: 55018983-55018983 on Build GRCh38
rs773069229 Chr.7: 55019284-55019284 on Build GRCh38
rs971948093 Chr.7: 55018900-55018900 on Build GRCh38
rs886062378 Chr.7: 55019041-55019041 on Build GRCh38
rs978286218 Chr.7: 55019035-55019035 on Build GRCh38
rs1036210424 Chr.7: 55019088-55019088 on Build GRCh38
rs919211832 Chr.7: 55018901-55018901 on Build GRCh38
rs897206305 Chr.7: 55019094-55019094 on Build GRCh38
rs1031135482 Chr.7: 55019062-55019063 on Build GRCh38
rs1027002893 Chr.7: 55018995-55018995 on Build GRCh38
rs375923119 Chr.7: 55019246-55019246 on Build GRCh38
rs866936692 Chr.7: 55019334-55019334 on Build GRCh38
rs17288924 Chr.7: 55018968-55018968 on Build GRCh38
rs1028735720 Chr.7: 55019336-55019336 on Build GRCh38
rs373129709 Chr.7: 55019338-55019338 on Build GRCh38
rs1048537252 Chr.7: 55019132-55019132 on Build GRCh38
rs537165657 Chr.7: 55019209-55019209 on Build GRCh38
rs712829 Chr.7: 55019062-55019062 on Build GRCh38
rs780865931 Chr.7: 55019333-55019333 on Build GRCh38
rs754527029 Chr.7: 55019329-55019329 on Build GRCh38
rs958454879 Chr.7: 55018854-55018854 on Build GRCh38
rs941739455 Chr.7: 55018892-55018892 on Build GRCh38
rs746536427 Chr.7: 55019240-55019240 on Build GRCh38
rs775964175 Chr.7: 55019295-55019295 on Build GRCh38
rs890276117 Chr.7: 55018926-55018928 on Build GRCh38
rs769658183 Chr.7: 55019276-55019276 on Build GRCh38
rs551488510 Chr.7: 55019144-55019144 on Build GRCh38
rs1025264514 Chr.7: 55019014-55019014 on Build GRCh38
rs1025127429 Chr.7: 55018845-55018845 on Build GRCh38
rs562250109 Chr.7: 55018919-55018919 on Build GRCh38
rs712830 Chr.7: 55019087-55019087 on Build GRCh38
rs1051656710 Chr.7: 55018924-55018924 on Build GRCh38
rs368795806 Chr.7: 55018997-55018997 on Build GRCh38
rs766029589 Chr.7: 55019310-55019310 on Build GRCh38
rs767651648 Chr.7: 55019293-55019293 on Build GRCh38
rs971299577 Chr.7: 55019025-55019025 on Build GRCh38
rs566284155 Chr.7: 55019068-55019068 on Build GRCh38
rs942722858 Chr.7: 55018959-55018959 on Build GRCh38
rs529539316 Chr.7: 55018939-55018939 on Build GRCh38
rs550360947 Chr.7: 55018853-55018853 on Build GRCh38
rs940570715 Chr.7: 55018948-55018948 on Build GRCh38
rs869245861 Chr.7: 55019196-55019196 on Build GRCh38
rs759582787 Chr.7: 55019291-55019291 on Build GRCh38
rs757936561 Chr.7: 55019307-55019307 on Build GRCh38
rs989083320 Chr.7: 55019063-55019063 on Build GRCh38
rs748159429 Chr.7: 55019275-55019275 on Build GRCh38
rs995883443 Chr.7: 55018982-55018982 on Build GRCh38
rs1037522485 Chr.7: 55018957-55018957 on Build GRCh38
rs921583827 Chr.7: 55019064-55019064 on Build GRCh38
rs41371445 Chr.7: 55018891-55018891 on Build GRCh38
rs780885208 Chr.7: 55019005-55019005 on Build GRCh38
rs774487133 Chr.7: 55019290-55019290 on Build GRCh38
rs570107879 Chr.7: 55019202-55019202 on Build GRCh38
rs895139219 Chr.7: 55019172-55019172 on Build GRCh38
rs987343349 Chr.7: 55019067-55019067 on Build GRCh38
rs991209541 Chr.7: 55018874-55018874 on Build GRCh38
rs17288945 Chr.7: 55019234-55019234 on Build GRCh38
rs771210838 Chr.7: 55019288-55019288 on Build GRCh38

View Full Product Details