Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56051445-56051953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6A
Entrez Gene ID
908
Gene Name
chaperonin containing TCP1 subunit 6A
Gene Aliases
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
UniGene
Hs.82916
Gene Chromosome Location
Chr.7: 56051685-56063989 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3 NM_004577.3 XM_005271773.1 XM_005271774.1 XM_005271775.1 XM_005271776.1 XM_006715760.1 XM_006715761.1 XM_011515461.1 XM_017012466.1 XM_017012467.1 XM_017012468.1 XM_017012469.1

Gene Symbol
PSPH
Entrez Gene ID
5723
Gene Name
phosphoserine phosphatase
Gene Aliases
PSP, PSPHD
UniGene
Hs.512656
Gene Chromosome Location
Chr.7: 56010363-56051604 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3 NM_004577.3 XM_005271773.1 XM_005271774.1 XM_005271775.1 XM_005271776.1 XM_006715760.1 XM_006715761.1 XM_011515461.1 XM_017012466.1 XM_017012467.1 XM_017012468.1 XM_017012469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4450583 c.71_100del30 p.S25_L34del10 Chr.7: 56051919-56051948 on Build GRCh38
5696266 c.40G>T p.A14S Chr.7: 56051888-56051888 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765291447 Chr.7: 56051703-56051703 on Build GRCh38
rs766631041 Chr.7: 56051832-56051832 on Build GRCh38
rs535381274 Chr.7: 56051782-56051782 on Build GRCh38
rs779908729 Chr.7: 56051847-56051847 on Build GRCh38
rs866860999 Chr.7: 56051853-56051853 on Build GRCh38
rs1010511213 Chr.7: 56051764-56051764 on Build GRCh38
rs747982869 Chr.7: 56051866-56051866 on Build GRCh38
rs895702455 Chr.7: 56051751-56051751 on Build GRCh38
rs1010046013 Chr.7: 56051624-56051624 on Build GRCh38
rs1041532508 Chr.7: 56051771-56051771 on Build GRCh38
rs968806855 Chr.7: 56051503-56051503 on Build GRCh38
rs1056930245 Chr.7: 56051601-56051601 on Build GRCh38
rs1020464242 Chr.7: 56051625-56051625 on Build GRCh38
rs746493351 Chr.7: 56051854-56051854 on Build GRCh38
rs768319112 Chr.7: 56051859-56051859 on Build GRCh38
rs190794562 Chr.7: 56051566-56051566 on Build GRCh38
rs942108353 Chr.7: 56051706-56051706 on Build GRCh38
rs774606575 Chr.7: 56051933-56051933 on Build GRCh38
rs756676392 Chr.7: 56051839-56051839 on Build GRCh38
rs545599589 Chr.7: 56051791-56051791 on Build GRCh38
rs1056980916 Chr.7: 56051744-56051744 on Build GRCh38
rs895648900 Chr.7: 56051620-56051620 on Build GRCh38
rs1018002520 Chr.7: 56051775-56051775 on Build GRCh38
rs1035471409 Chr.7: 56051487-56051487 on Build GRCh38
rs34822782 Chr.7: 56051478-56051478 on Build GRCh38
rs760291328 Chr.7: 56051687-56051687 on Build GRCh38
rs886062396 Chr.7: 56051527-56051527 on Build GRCh38
rs563602633 Chr.7: 56051822-56051822 on Build GRCh38
rs964992649 Chr.7: 56051636-56051636 on Build GRCh38
rs963466545 Chr.7: 56051813-56051813 on Build GRCh38
rs867241037 Chr.7: 56051615-56051615 on Build GRCh38
rs767991539 Chr.7: 56051835-56051835 on Build GRCh38
rs1010394875 Chr.7: 56051500-56051500 on Build GRCh38
rs751637412 Chr.7: 56051833-56051833 on Build GRCh38
rs762875896 Chr.7: 56051905-56051905 on Build GRCh38
rs1019999867 Chr.7: 56051502-56051502 on Build GRCh38
rs894290431 Chr.7: 56051486-56051486 on Build GRCh38
rs886062398 Chr.7: 56051561-56051561 on Build GRCh38
rs1037653165 Chr.7: 56051561-56051561 on Build GRCh38
rs180755860 Chr.7: 56051787-56051787 on Build GRCh38
rs983610722 Chr.7: 56051698-56051698 on Build GRCh38
rs578186989 Chr.7: 56051788-56051788 on Build GRCh38
rs933469329 Chr.7: 56051538-56051538 on Build GRCh38
rs1001406975 Chr.7: 56051585-56051585 on Build GRCh38
rs187219685 Chr.7: 56051554-56051554 on Build GRCh38
rs574372841 Chr.7: 56051770-56051770 on Build GRCh38
rs978421911 Chr.7: 56051504-56051504 on Build GRCh38
rs769788272 Chr.7: 56051885-56051885 on Build GRCh38
rs569749358 Chr.7: 56051674-56051674 on Build GRCh38
rs772819900 Chr.7: 56051902-56051902 on Build GRCh38
rs773085914 Chr.7: 56051817-56051817 on Build GRCh38
rs565582529 Chr.7: 56051628-56051628 on Build GRCh38
rs148855313 Chr.7: 56051529-56051529 on Build GRCh38
rs547177613 Chr.7: 56051599-56051599 on Build GRCh38
rs1015091404 Chr.7: 56051806-56051806 on Build GRCh38
rs927343831 Chr.7: 56051820-56051820 on Build GRCh38
rs561809908 Chr.7: 56051555-56051555 on Build GRCh38
rs563812934 Chr.7: 56051526-56051526 on Build GRCh38
rs539255120 Chr.7: 56051634-56051634 on Build GRCh38
rs551153059 Chr.7: 56051673-56051673 on Build GRCh38
rs555370989 Chr.7: 56051731-56051731 on Build GRCh38
rs764896645 Chr.7: 56051815-56051815 on Build GRCh38
rs766499405 Chr.7: 56051914-56051914 on Build GRCh38
rs559290263 Chr.7: 56051719-56051719 on Build GRCh38
rs904308540 Chr.7: 56051772-56051772 on Build GRCh38
rs927292840 Chr.7: 56051723-56051723 on Build GRCh38
rs955064298 Chr.7: 56051696-56051696 on Build GRCh38
rs905759611 Chr.7: 56051476-56051476 on Build GRCh38
rs542638354 Chr.7: 56051841-56051841 on Build GRCh38
rs868684313 Chr.7: 56051889-56051889 on Build GRCh38
rs997827517 Chr.7: 56051785-56051785 on Build GRCh38
rs575719815 Chr.7: 56051831-56051831 on Build GRCh38
rs539413143 Chr.7: 56051613-56051613 on Build GRCh38
rs761665214 Chr.7: 56051812-56051812 on Build GRCh38
rs560984863 Chr.7: 56051845-56051845 on Build GRCh38
rs974998705 Chr.7: 56051665-56051665 on Build GRCh38
rs937363029 Chr.7: 56051821-56051821 on Build GRCh38
rs545470619 Chr.7: 56051520-56051520 on Build GRCh38
rs942041067 Chr.7: 56051560-56051560 on Build GRCh38
rs886062397 Chr.7: 56051557-56051557 on Build GRCh38
rs776307829 Chr.7: 56051865-56051865 on Build GRCh38
rs959858760 Chr.7: 56051499-56051499 on Build GRCh38
rs753242916 Chr.7: 56051836-56051836 on Build GRCh38
rs749666879 Chr.7: 56051844-56051844 on Build GRCh38
rs1041975364 Chr.7: 56051837-56051837 on Build GRCh38
rs755111167 Chr.7: 56051834-56051834 on Build GRCh38
rs999937558 Chr.7: 56051773-56051773 on Build GRCh38
rs1017950032 Chr.7: 56051630-56051630 on Build GRCh38

View Full Product Details