Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56057893-56058407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6A
Entrez Gene ID
908
Gene Name
chaperonin containing TCP1 subunit 6A
Gene Aliases
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
UniGene
Hs.82916
Gene Chromosome Location
Chr.7: 56051685-56063989 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6415221 c.668A>T p.K223I Chr.7: 56058046-56058046 on Build GRCh38
6034147 c.726-9_763>44 p.? Chr.7: 56058353-56058399 on Build GRCh38
6416967 c.639C>A p.D213E Chr.7: 56058017-56058017 on Build GRCh38
3267084 c.725+1G>A p.? Chr.7: 56058104-56058104 on Build GRCh38
6178016 c.683C>T p.A228V Chr.7: 56058061-56058061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181192271 Chr.7: 56058220-56058220 on Build GRCh38
rs189719119 Chr.7: 56058156-56058156 on Build GRCh38
rs375113562 Chr.7: 56057961-56057961 on Build GRCh38
rs758136626 Chr.7: 56058102-56058102 on Build GRCh38
rs760643265 Chr.7: 56057920-56057920 on Build GRCh38
rs567948145 Chr.7: 56058080-56058080 on Build GRCh38
rs533956429 Chr.7: 56058162-56058162 on Build GRCh38
rs764781227 Chr.7: 56058266-56058270 on Build GRCh38
rs369940433 Chr.7: 56058143-56058143 on Build GRCh38
rs768461065 Chr.7: 56058129-56058129 on Build GRCh38
rs368742773 Chr.7: 56058358-56058358 on Build GRCh38
rs199970063 Chr.7: 56058086-56058086 on Build GRCh38
rs139223504 Chr.7: 56058056-56058056 on Build GRCh38
rs768137658 Chr.7: 56058071-56058071 on Build GRCh38
rs998778940 Chr.7: 56057922-56057922 on Build GRCh38
rs148087479 Chr.7: 56058151-56058151 on Build GRCh38
rs546317437 Chr.7: 56057976-56057977 on Build GRCh38
rs187150768 Chr.7: 56058270-56058270 on Build GRCh38
rs148616984 Chr.7: 56058069-56058069 on Build GRCh38
rs750453724 Chr.7: 56057948-56057948 on Build GRCh38
rs758203138 Chr.7: 56058354-56058354 on Build GRCh38
rs756551563 Chr.7: 56058076-56058076 on Build GRCh38
rs756290750 Chr.7: 56058343-56058344 on Build GRCh38
rs778027927 Chr.7: 56058077-56058077 on Build GRCh38
rs141752926 Chr.7: 56058020-56058020 on Build GRCh38
rs768640457 Chr.7: 56058040-56058040 on Build GRCh38
rs1028356842 Chr.7: 56058291-56058291 on Build GRCh38
rs758115944 Chr.7: 56058359-56058359 on Build GRCh38
rs908218831 Chr.7: 56058024-56058024 on Build GRCh38
rs746885599 Chr.7: 56058034-56058034 on Build GRCh38
rs33922584 Chr.7: 56058064-56058064 on Build GRCh38
rs771667117 Chr.7: 56058014-56058014 on Build GRCh38
rs758075159 Chr.7: 56058154-56058154 on Build GRCh38
rs955168053 Chr.7: 56058211-56058212 on Build GRCh38
rs190455401 Chr.7: 56058317-56058317 on Build GRCh38
rs748550393 Chr.7: 56057990-56057990 on Build GRCh38
rs763196812 Chr.7: 56058057-56058057 on Build GRCh38
rs776282473 Chr.7: 56058045-56058045 on Build GRCh38
rs903196735 Chr.7: 56058073-56058073 on Build GRCh38
rs528101378 Chr.7: 56057977-56057977 on Build GRCh38
rs996674008 Chr.7: 56057966-56057966 on Build GRCh38
rs746581128 Chr.7: 56058117-56058117 on Build GRCh38
rs755602388 Chr.7: 56058376-56058376 on Build GRCh38
rs199694607 Chr.7: 56058060-56058060 on Build GRCh38
rs779638409 Chr.7: 56058116-56058116 on Build GRCh38
rs778686498 Chr.7: 56058008-56058008 on Build GRCh38
rs957449992 Chr.7: 56057951-56057951 on Build GRCh38
rs756855905 Chr.7: 56057999-56057999 on Build GRCh38
rs542702385 Chr.7: 56058322-56058322 on Build GRCh38
rs149533082 Chr.7: 56058062-56058062 on Build GRCh38
rs747944355 Chr.7: 56058344-56058347 on Build GRCh38
rs751088916 Chr.7: 56058378-56058378 on Build GRCh38
rs577890814 Chr.7: 56058216-56058216 on Build GRCh38
rs553204505 Chr.7: 56058177-56058177 on Build GRCh38
rs769595038 Chr.7: 56058146-56058146 on Build GRCh38
rs774410884 Chr.7: 56058153-56058153 on Build GRCh38
rs146242535 Chr.7: 56058051-56058051 on Build GRCh38
rs765352658 Chr.7: 56058052-56058052 on Build GRCh38
rs1015275838 Chr.7: 56058225-56058225 on Build GRCh38
rs750667827 Chr.7: 56058344-56058344 on Build GRCh38
rs903146169 Chr.7: 56057917-56057917 on Build GRCh38
rs1028152176 Chr.7: 56058209-56058209 on Build GRCh38
rs780294630 Chr.7: 56058344-56058345 on Build GRCh38
rs41298776 Chr.7: 56057943-56057943 on Build GRCh38
rs755505409 Chr.7: 56057979-56057979 on Build GRCh38
rs758518835 Chr.7: 56057949-56057949 on Build GRCh38
rs754258925 Chr.7: 56058354-56058354 on Build GRCh38
rs767890862 Chr.7: 56058311-56058311 on Build GRCh38
rs781357702 Chr.7: 56057982-56057982 on Build GRCh38
rs961002218 Chr.7: 56058036-56058036 on Build GRCh38
rs750045709 Chr.7: 56058091-56058091 on Build GRCh38
rs773014978 Chr.7: 56058148-56058148 on Build GRCh38
rs750131519 Chr.7: 56058142-56058142 on Build GRCh38
rs368984460 Chr.7: 56058005-56058005 on Build GRCh38
rs183269124 Chr.7: 56058339-56058339 on Build GRCh38
rs961054352 Chr.7: 56058263-56058263 on Build GRCh38

View Full Product Details