Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56059564-56060108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6A
Entrez Gene ID
908
Gene Name
chaperonin containing TCP1 subunit 6A
Gene Aliases
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
UniGene
Hs.82916
Gene Chromosome Location
Chr.7: 56051685-56063989 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
383492 c.1024G>T p.D342Y Chr.7: 56059599-56059599 on Build GRCh38
6623320 c.1012G>T p.D338Y Chr.7: 56059587-56059587 on Build GRCh38
3685191 c.1024G>C p.D342H Chr.7: 56059599-56059599 on Build GRCh38
6623321 c.1062A>G p.T354T Chr.7: 56059637-56059637 on Build GRCh38
3267095 c.1028G>T p.C343F Chr.7: 56059603-56059603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774734372 Chr.7: 56059692-56059699 on Build GRCh38
rs775590542 Chr.7: 56059991-56059991 on Build GRCh38
rs200832854 Chr.7: 56059688-56059690 on Build GRCh38
rs79776912 Chr.7: 56059858-56059858 on Build GRCh38
rs889972622 Chr.7: 56059916-56059916 on Build GRCh38
rs1006943233 Chr.7: 56059696-56059697 on Build GRCh38
rs952708890 Chr.7: 56059959-56059959 on Build GRCh38
rs973129712 Chr.7: 56059728-56059728 on Build GRCh38
rs1026065992 Chr.7: 56059944-56059944 on Build GRCh38
rs915661804 Chr.7: 56059931-56059931 on Build GRCh38
rs542012411 Chr.7: 56059834-56059834 on Build GRCh38
rs763285204 Chr.7: 56059673-56059673 on Build GRCh38
rs145667181 Chr.7: 56059929-56059929 on Build GRCh38
rs759456625 Chr.7: 56059678-56059678 on Build GRCh38
rs768275609 Chr.7: 56059657-56059657 on Build GRCh38
rs78567648 Chr.7: 56059856-56059856 on Build GRCh38
rs182670610 Chr.7: 56059732-56059732 on Build GRCh38
rs775183593 Chr.7: 56059659-56059660 on Build GRCh38
rs771484780 Chr.7: 56059625-56059625 on Build GRCh38
rs576730840 Chr.7: 56059713-56059713 on Build GRCh38
rs780127620 Chr.7: 56059833-56059833 on Build GRCh38
rs572890236 Chr.7: 56059624-56059624 on Build GRCh38
rs377037039 Chr.7: 56060014-56060014 on Build GRCh38
rs762734947 Chr.7: 56059672-56059672 on Build GRCh38
rs140044923 Chr.7: 56059683-56059683 on Build GRCh38
rs889918441 Chr.7: 56059694-56059697 on Build GRCh38
rs779118630 Chr.7: 56059731-56059731 on Build GRCh38
rs772790248 Chr.7: 56059671-56059671 on Build GRCh38
rs947637618 Chr.7: 56059858-56059858 on Build GRCh38
rs1053403915 Chr.7: 56059621-56059621 on Build GRCh38
rs747826456 Chr.7: 56059662-56059662 on Build GRCh38
rs560571092 Chr.7: 56059902-56059902 on Build GRCh38
rs981875242 Chr.7: 56059766-56059766 on Build GRCh38
rs779138315 Chr.7: 56059637-56059637 on Build GRCh38
rs575994115 Chr.7: 56060054-56060054 on Build GRCh38
rs893433893 Chr.7: 56059817-56059817 on Build GRCh38
rs1043287430 Chr.7: 56059915-56059915 on Build GRCh38
rs999166269 Chr.7: 56059620-56059620 on Build GRCh38
rs117697809 Chr.7: 56059715-56059715 on Build GRCh38
rs372800761 Chr.7: 56059586-56059586 on Build GRCh38
rs984457348 Chr.7: 56060043-56060043 on Build GRCh38
rs147515036 Chr.7: 56059599-56059599 on Build GRCh38
rs778155800 Chr.7: 56059668-56059668 on Build GRCh38
rs777940397 Chr.7: 56059610-56059610 on Build GRCh38
rs935019776 Chr.7: 56059785-56059785 on Build GRCh38
rs767817699 Chr.7: 56059681-56059681 on Build GRCh38
rs906209935 Chr.7: 56059618-56059618 on Build GRCh38
rs758590232 Chr.7: 56059717-56059717 on Build GRCh38
rs867911887 Chr.7: 56059764-56059764 on Build GRCh38
rs1006995812 Chr.7: 56059928-56059928 on Build GRCh38
rs35171408 Chr.7: 56059857-56059858 on Build GRCh38
rs781624320 Chr.7: 56059596-56059596 on Build GRCh38
rs994217996 Chr.7: 56059935-56059935 on Build GRCh38
rs1019634253 Chr.7: 56059698-56059698 on Build GRCh38
rs746288015 Chr.7: 56059638-56059638 on Build GRCh38
rs398004856 Chr.7: 56059868-56059869 on Build GRCh38
rs558629472 Chr.7: 56059684-56059684 on Build GRCh38
rs772074879 Chr.7: 56059930-56059930 on Build GRCh38
rs747300617 Chr.7: 56060057-56060057 on Build GRCh38
rs916425399 Chr.7: 56059761-56059761 on Build GRCh38
rs766409952 Chr.7: 56059673-56059673 on Build GRCh38
rs927678093 Chr.7: 56059774-56059774 on Build GRCh38
rs746021637 Chr.7: 56059768-56059768 on Build GRCh38
rs530363295 Chr.7: 56059800-56059800 on Build GRCh38
rs756188193 Chr.7: 56059603-56059603 on Build GRCh38

View Full Product Details