Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56106257-56106773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD2
Entrez Gene ID
51142
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Gene Aliases
C7orf17, MNRR1, NS2TP, PARK22
UniGene
Hs.389996
Gene Chromosome Location
Chr.7: 56101562-56106630 on Build GRCh38
Transcripts
NM_001320327.1 NM_016139.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4854358 c.43C>G p.P15A Chr.7: 56106371-56106371 on Build GRCh38
4537275 c.23G>A p.R8H Chr.7: 56106391-56106391 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776234231 Chr.7: 56106373-56106373 on Build GRCh38
rs570727110 Chr.7: 56106648-56106648 on Build GRCh38
rs753967131 Chr.7: 56106430-56106430 on Build GRCh38
rs373956381 Chr.7: 56106382-56106382 on Build GRCh38
rs1002307678 Chr.7: 56106558-56106558 on Build GRCh38
rs750193744 Chr.7: 56106319-56106319 on Build GRCh38
rs1010792606 Chr.7: 56106609-56106609 on Build GRCh38
rs146485961 Chr.7: 56106299-56106299 on Build GRCh38
rs921089829 Chr.7: 56106477-56106477 on Build GRCh38
rs781422604 Chr.7: 56106400-56106400 on Build GRCh38
rs975849397 Chr.7: 56106307-56106307 on Build GRCh38
rs773743531 Chr.7: 56106746-56106746 on Build GRCh38
rs1015602492 Chr.7: 56106606-56106606 on Build GRCh38
rs1007103678 Chr.7: 56106629-56106629 on Build GRCh38
rs921102429 Chr.7: 56106626-56106626 on Build GRCh38
rs768263808 Chr.7: 56106370-56106370 on Build GRCh38
rs750343228 Chr.7: 56106387-56106387 on Build GRCh38
rs946153795 Chr.7: 56106736-56106736 on Build GRCh38
rs904691925 Chr.7: 56106742-56106742 on Build GRCh38
rs575063134 Chr.7: 56106439-56106439 on Build GRCh38
rs761895736 Chr.7: 56106314-56106314 on Build GRCh38
rs142444896 Chr.7: 56106409-56106409 on Build GRCh38
rs929111466 Chr.7: 56106503-56106503 on Build GRCh38
rs753824117 Chr.7: 56106460-56106460 on Build GRCh38
rs751783132 Chr.7: 56106392-56106392 on Build GRCh38
rs770707415 Chr.7: 56106449-56106449 on Build GRCh38
rs113578920 Chr.7: 56106663-56106663 on Build GRCh38
rs1018513155 Chr.7: 56106645-56106645 on Build GRCh38
rs746271029 Chr.7: 56106417-56106417 on Build GRCh38
rs1004250620 Chr.7: 56106597-56106597 on Build GRCh38
rs201974351 Chr.7: 56106348-56106348 on Build GRCh38
rs1047028376 Chr.7: 56106538-56106538 on Build GRCh38
rs1043583544 Chr.7: 56106740-56106740 on Build GRCh38
rs77036114 Chr.7: 56106658-56106658 on Build GRCh38
rs965700477 Chr.7: 56106607-56106607 on Build GRCh38
rs943140091 Chr.7: 56106306-56106306 on Build GRCh38
rs1025137926 Chr.7: 56106375-56106375 on Build GRCh38
rs1007049652 Chr.7: 56106747-56106747 on Build GRCh38
rs77840769 Chr.7: 56106696-56106697 on Build GRCh38
rs764401963 Chr.7: 56106458-56106458 on Build GRCh38
rs755144061 Chr.7: 56106398-56106398 on Build GRCh38
rs369456404 Chr.7: 56106360-56106360 on Build GRCh38
rs567009389 Chr.7: 56106537-56106537 on Build GRCh38
rs749822788 Chr.7: 56106406-56106406 on Build GRCh38
rs773694807 Chr.7: 56106432-56106433 on Build GRCh38
rs1057458276 Chr.7: 56106571-56106571 on Build GRCh38
rs112876794 Chr.7: 56106425-56106425 on Build GRCh38
rs35422924 Chr.7: 56106697-56106698 on Build GRCh38
rs536091658 Chr.7: 56106659-56106659 on Build GRCh38
rs1048579929 Chr.7: 56106697-56106697 on Build GRCh38
rs780265780 Chr.7: 56106322-56106322 on Build GRCh38
rs765608255 Chr.7: 56106462-56106462 on Build GRCh38
rs769206892 Chr.7: 56106666-56106666 on Build GRCh38
rs775199912 Chr.7: 56106368-56106368 on Build GRCh38
rs137965562 Chr.7: 56106374-56106374 on Build GRCh38
rs540264762 Chr.7: 56106464-56106464 on Build GRCh38
rs146847249 Chr.7: 56106661-56106661 on Build GRCh38
rs762795852 Chr.7: 56106388-56106388 on Build GRCh38
rs767363392 Chr.7: 56106454-56106454 on Build GRCh38
rs557585915 Chr.7: 56106718-56106719 on Build GRCh38
rs816406 Chr.7: 56106478-56106478 on Build GRCh38
rs997161117 Chr.7: 56106650-56106650 on Build GRCh38
rs779510094 Chr.7: 56106441-56106441 on Build GRCh38
rs752972663 Chr.7: 56106433-56106433 on Build GRCh38
rs989460315 Chr.7: 56106276-56106276 on Build GRCh38
rs765200757 Chr.7: 56106385-56106385 on Build GRCh38
rs200226056 Chr.7: 56106424-56106424 on Build GRCh38
rs1046605969 Chr.7: 56106690-56106690 on Build GRCh38
rs375924428 Chr.7: 56106696-56106696 on Build GRCh38
rs1048101846 Chr.7: 56106377-56106377 on Build GRCh38
rs775655743 Chr.7: 56106457-56106457 on Build GRCh38
rs557143801 Chr.7: 56106434-56106434 on Build GRCh38
rs949677257 Chr.7: 56106683-56106683 on Build GRCh38
rs867193277 Chr.7: 56106490-56106490 on Build GRCh38
rs764116300 Chr.7: 56106435-56106435 on Build GRCh38
rs816408 Chr.7: 56106277-56106277 on Build GRCh38
rs755053648 Chr.7: 56106332-56106332 on Build GRCh38
rs766155505 Chr.7: 56106389-56106389 on Build GRCh38
rs568922394 Chr.7: 56106678-56106678 on Build GRCh38
rs909100683 Chr.7: 56106528-56106528 on Build GRCh38
rs771253445 Chr.7: 56106366-56106366 on Build GRCh38
rs767666028 Chr.7: 56106432-56106432 on Build GRCh38
rs200342583 Chr.7: 56106337-56106337 on Build GRCh38
rs750716521 Chr.7: 56106557-56106557 on Build GRCh38
rs760289183 Chr.7: 56106369-56106369 on Build GRCh38
rs953861639 Chr.7: 56106631-56106631 on Build GRCh38
rs554200294 Chr.7: 56106450-56106450 on Build GRCh38
rs538940651 Chr.7: 56106649-56106649 on Build GRCh38
rs764013716 Chr.7: 56106428-56106428 on Build GRCh38
rs185793529 Chr.7: 56106623-56106623 on Build GRCh38
rs976685580 Chr.7: 56106616-56106616 on Build GRCh38
rs746171975 Chr.7: 56106444-56106444 on Build GRCh38
rs748858900 Chr.7: 56106448-56106448 on Build GRCh38
rs11546416 Chr.7: 56106408-56106408 on Build GRCh38
rs979096924 Chr.7: 56106290-56106290 on Build GRCh38
rs527766236 Chr.7: 56106593-56106593 on Build GRCh38
rs751429500 Chr.7: 56106287-56106287 on Build GRCh38
rs778328496 Chr.7: 56106404-56106404 on Build GRCh38
rs765290689 Chr.7: 56106316-56106316 on Build GRCh38
rs539929834 Chr.7: 56106710-56106710 on Build GRCh38
rs747605451 Chr.7: 56106423-56106423 on Build GRCh38
rs367715869 Chr.7: 56106733-56106733 on Build GRCh38
rs909056860 Chr.7: 56106661-56106661 on Build GRCh38
rs572601459 Chr.7: 56106461-56106461 on Build GRCh38
rs10043 Chr.7: 56106422-56106422 on Build GRCh38
rs548797126 Chr.7: 56106535-56106535 on Build GRCh38
rs111801456 Chr.7: 56106532-56106532 on Build GRCh38
rs749606287 Chr.7: 56106363-56106363 on Build GRCh38
rs769196368 Chr.7: 56106564-56106564 on Build GRCh38
rs927218980 Chr.7: 56106689-56106689 on Build GRCh38
rs566875073 Chr.7: 56106403-56106403 on Build GRCh38
rs576618283 Chr.7: 56106514-56106514 on Build GRCh38
rs773975043 Chr.7: 56106429-56106429 on Build GRCh38
rs532870274 Chr.7: 56106479-56106479 on Build GRCh38
rs60847483 Chr.7: 56106506-56106506 on Build GRCh38
rs373823298 Chr.7: 56106416-56106416 on Build GRCh38
rs751787786 Chr.7: 56106323-56106323 on Build GRCh38
rs892748488 Chr.7: 56106515-56106515 on Build GRCh38
rs968101309 Chr.7: 56106278-56106278 on Build GRCh38
rs368503740 Chr.7: 56106436-56106436 on Build GRCh38
rs1001088436 Chr.7: 56106312-56106312 on Build GRCh38
rs780978381 Chr.7: 56106446-56106446 on Build GRCh38
rs1022282544 Chr.7: 56106445-56106445 on Build GRCh38
rs816407 Chr.7: 56106447-56106447 on Build GRCh38
rs372393265 Chr.7: 56106695-56106695 on Build GRCh38
rs7808463 Chr.7: 56106320-56106320 on Build GRCh38
rs925622078 Chr.7: 56106480-56106480 on Build GRCh38
rs752705344 Chr.7: 56106401-56106401 on Build GRCh38
rs905376881 Chr.7: 56106694-56106694 on Build GRCh38
rs371507821 Chr.7: 56106492-56106492 on Build GRCh38
rs572637014 Chr.7: 56106682-56106682 on Build GRCh38
rs952698648 Chr.7: 56106596-56106596 on Build GRCh38

View Full Product Details