Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66075697-66076197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASL
Entrez Gene ID
435
Gene Name
argininosuccinate lyase
Gene Aliases
ASAL
UniGene
Hs.632015
Gene Chromosome Location
Chr.7: 66075789-66093343 on Build GRCh38
Transcripts
NM_000048.3 NM_001024943.1 NM_001024944.1 NM_001024946.1

SNP Detail

SNP ID Chromosome Location
rs930815776 Chr.7: 66075755-66075755 on Build GRCh38
rs1035279429 Chr.7: 66075924-66075924 on Build GRCh38
rs1050266174 Chr.7: 66075825-66075825 on Build GRCh38
rs998541232 Chr.7: 66075909-66075909 on Build GRCh38
rs567697935 Chr.7: 66075779-66075779 on Build GRCh38
rs1002023330 Chr.7: 66075845-66075845 on Build GRCh38
rs888941126 Chr.7: 66075829-66075829 on Build GRCh38
rs768635601 Chr.7: 66076104-66076104 on Build GRCh38
rs773195623 Chr.7: 66076141-66076141 on Build GRCh38
rs895587887 Chr.7: 66075946-66075946 on Build GRCh38
rs149918397 Chr.7: 66076086-66076086 on Build GRCh38
rs995181145 Chr.7: 66076177-66076177 on Build GRCh38
rs909242711 Chr.7: 66075775-66075775 on Build GRCh38
rs762118548 Chr.7: 66076038-66076038 on Build GRCh38
rs577433994 Chr.7: 66075731-66075731 on Build GRCh38
rs140499284 Chr.7: 66075979-66075979 on Build GRCh38
rs776531650 Chr.7: 66076116-66076116 on Build GRCh38
rs376836768 Chr.7: 66076122-66076122 on Build GRCh38
rs1012606725 Chr.7: 66075959-66075959 on Build GRCh38
rs985040331 Chr.7: 66075750-66075750 on Build GRCh38
rs1040362831 Chr.7: 66075790-66075790 on Build GRCh38
rs535730908 Chr.7: 66075791-66075791 on Build GRCh38
rs913916151 Chr.7: 66076048-66076048 on Build GRCh38
rs944745788 Chr.7: 66075783-66075783 on Build GRCh38
rs966270687 Chr.7: 66075996-66075996 on Build GRCh38
rs756883050 Chr.7: 66076075-66076075 on Build GRCh38
rs910803249 Chr.7: 66075782-66075782 on Build GRCh38
rs985477836 Chr.7: 66075983-66075983 on Build GRCh38
rs372229504 Chr.7: 66076066-66076066 on Build GRCh38
rs763023035 Chr.7: 66076144-66076144 on Build GRCh38
rs796303307 Chr.7: 66076148-66076148 on Build GRCh38
rs556520035 Chr.7: 66076010-66076010 on Build GRCh38
rs573340046 Chr.7: 66076028-66076028 on Build GRCh38
rs112097520 Chr.7: 66075799-66075799 on Build GRCh38
rs952352136 Chr.7: 66075973-66075973 on Build GRCh38
rs995694201 Chr.7: 66075919-66075919 on Build GRCh38
rs973809638 Chr.7: 66075722-66075722 on Build GRCh38
rs766867890 Chr.7: 66076051-66076051 on Build GRCh38
rs543143954 Chr.7: 66075990-66075990 on Build GRCh38
rs1055488486 Chr.7: 66075860-66075860 on Build GRCh38
rs961008012 Chr.7: 66075918-66075918 on Build GRCh38
rs753529387 Chr.7: 66076073-66076073 on Build GRCh38
rs565969581 Chr.7: 66075800-66075800 on Build GRCh38
rs528791224 Chr.7: 66076165-66076165 on Build GRCh38
rs763390749 Chr.7: 66076044-66076044 on Build GRCh38
rs750491000 Chr.7: 66076042-66076042 on Build GRCh38
rs779988786 Chr.7: 66076089-66076089 on Build GRCh38
rs1023729210 Chr.7: 66075723-66075723 on Build GRCh38
rs776090425 Chr.7: 66075930-66075930 on Build GRCh38
rs920775517 Chr.7: 66075754-66075754 on Build GRCh38
rs557549042 Chr.7: 66075964-66075964 on Build GRCh38
rs1015270428 Chr.7: 66075887-66075887 on Build GRCh38
rs369860716 Chr.7: 66076123-66076123 on Build GRCh38
rs746934631 Chr.7: 66076103-66076103 on Build GRCh38
rs373177285 Chr.7: 66076145-66076145 on Build GRCh38
rs558890925 Chr.7: 66075865-66075865 on Build GRCh38
rs543301670 Chr.7: 66076039-66076039 on Build GRCh38
rs1036937767 Chr.7: 66076166-66076166 on Build GRCh38
rs781420770 Chr.7: 66076060-66076060 on Build GRCh38
rs902882952 Chr.7: 66075903-66075903 on Build GRCh38
rs1010672027 Chr.7: 66075842-66075842 on Build GRCh38
rs559747484 Chr.7: 66076052-66076052 on Build GRCh38
rs910689067 Chr.7: 66075988-66075988 on Build GRCh38
rs896849781 Chr.7: 66076175-66076175 on Build GRCh38
rs369399314 Chr.7: 66076041-66076041 on Build GRCh38
rs778309914 Chr.7: 66076078-66076078 on Build GRCh38
rs545638088 Chr.7: 66075879-66075879 on Build GRCh38

View Full Product Details