Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66082277-66082767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASL
Entrez Gene ID
435
Gene Name
argininosuccinate lyase
Gene Aliases
ASAL
UniGene
Hs.632015
Gene Chromosome Location
Chr.7: 66075789-66093343 on Build GRCh38
Transcripts
NM_000048.3 NM_001024943.1 NM_001024944.1 NM_001024946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
453297 c.219G>C p.E73D Chr.7: 66082379-66082379 on Build GRCh38
3720887 c.286C>G p.L96V Chr.7: 66082446-66082446 on Build GRCh38
7286747 c.261C>T p.D87D Chr.7: 66082421-66082421 on Build GRCh38
4819858 c.288G>A p.L96L Chr.7: 66082448-66082448 on Build GRCh38
5880526 c.232A>G p.T78A Chr.7: 66082392-66082392 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757507610 Chr.7: 66082341-66082341 on Build GRCh38
rs748674685 Chr.7: 66082466-66082466 on Build GRCh38
rs775520810 Chr.7: 66082379-66082379 on Build GRCh38
rs371162956 Chr.7: 66082458-66082458 on Build GRCh38
rs201523601 Chr.7: 66082452-66082452 on Build GRCh38
rs887932329 Chr.7: 66082681-66082681 on Build GRCh38
rs759257974 Chr.7: 66082411-66082411 on Build GRCh38
rs1018268240 Chr.7: 66082702-66082702 on Build GRCh38
rs767186104 Chr.7: 66082414-66082414 on Build GRCh38
rs569718973 Chr.7: 66082552-66082552 on Build GRCh38
rs536613106 Chr.7: 66082633-66082633 on Build GRCh38
rs778679108 Chr.7: 66082451-66082451 on Build GRCh38
rs1016374064 Chr.7: 66082338-66082338 on Build GRCh38
rs869312986 Chr.7: 66082417-66082417 on Build GRCh38
rs1042681659 Chr.7: 66082446-66082446 on Build GRCh38
rs1020917586 Chr.7: 66082737-66082737 on Build GRCh38
rs762014141 Chr.7: 66082390-66082390 on Build GRCh38
rs141046612 Chr.7: 66082402-66082402 on Build GRCh38
rs777647003 Chr.7: 66082322-66082322 on Build GRCh38
rs535769121 Chr.7: 66082518-66082518 on Build GRCh38
rs979264392 Chr.7: 66082491-66082491 on Build GRCh38
rs966103360 Chr.7: 66082720-66082720 on Build GRCh38
rs896646502 Chr.7: 66082510-66082510 on Build GRCh38
rs1027739421 Chr.7: 66082369-66082369 on Build GRCh38
rs534406964 Chr.7: 66082571-66082571 on Build GRCh38
rs780126368 Chr.7: 66082456-66082456 on Build GRCh38
rs933291133 Chr.7: 66082393-66082393 on Build GRCh38
rs148816474 Chr.7: 66082308-66082308 on Build GRCh38
rs143434632 Chr.7: 66082630-66082630 on Build GRCh38
rs1023669261 Chr.7: 66082302-66082302 on Build GRCh38
rs780895668 Chr.7: 66082436-66082436 on Build GRCh38
rs567573139 Chr.7: 66082319-66082319 on Build GRCh38
rs774204716 Chr.7: 66082403-66082403 on Build GRCh38
rs565578318 Chr.7: 66082556-66082556 on Build GRCh38
rs150418929 Chr.7: 66082542-66082542 on Build GRCh38
rs759284788 Chr.7: 66082475-66082475 on Build GRCh38
rs557383201 Chr.7: 66082619-66082619 on Build GRCh38
rs772209699 Chr.7: 66082362-66082362 on Build GRCh38
rs1015741360 Chr.7: 66082309-66082309 on Build GRCh38
rs373966403 Chr.7: 66082386-66082386 on Build GRCh38
rs969801136 Chr.7: 66082307-66082307 on Build GRCh38
rs937926250 Chr.7: 66082539-66082539 on Build GRCh38
rs1036665489 Chr.7: 66082553-66082553 on Build GRCh38
rs137915973 Chr.7: 66082389-66082389 on Build GRCh38
rs905418130 Chr.7: 66082624-66082624 on Build GRCh38
rs199547939 Chr.7: 66082345-66082345 on Build GRCh38
rs137955991 Chr.7: 66082747-66082747 on Build GRCh38
rs976229247 Chr.7: 66082467-66082467 on Build GRCh38
rs995193018 Chr.7: 66082618-66082618 on Build GRCh38
rs1014237581 Chr.7: 66082299-66082299 on Build GRCh38
rs752100894 Chr.7: 66082420-66082420 on Build GRCh38
rs757105613 Chr.7: 66082445-66082445 on Build GRCh38
rs200354837 Chr.7: 66082331-66082331 on Build GRCh38
rs781492633 Chr.7: 66082459-66082459 on Build GRCh38
rs561493592 Chr.7: 66082704-66082705 on Build GRCh38
rs771810598 Chr.7: 66082487-66082487 on Build GRCh38
rs966804115 Chr.7: 66082740-66082740 on Build GRCh38
rs777437569 Chr.7: 66082441-66082441 on Build GRCh38
rs374304304 Chr.7: 66082440-66082440 on Build GRCh38
rs1052642416 Chr.7: 66082468-66082468 on Build GRCh38
rs997935254 Chr.7: 66082724-66082724 on Build GRCh38
rs150244667 Chr.7: 66082444-66082444 on Build GRCh38
rs368188924 Chr.7: 66082455-66082455 on Build GRCh38
rs752199093 Chr.7: 66082314-66082314 on Build GRCh38
rs770348828 Chr.7: 66082473-66082473 on Build GRCh38
rs917615634 Chr.7: 66082547-66082547 on Build GRCh38
rs367590531 Chr.7: 66082488-66082488 on Build GRCh38
rs551946013 Chr.7: 66082344-66082344 on Build GRCh38
rs1007787504 Chr.7: 66082682-66082682 on Build GRCh38
rs949070234 Chr.7: 66082551-66082551 on Build GRCh38
rs556940515 Chr.7: 66082636-66082636 on Build GRCh38
rs760048145 Chr.7: 66082497-66082497 on Build GRCh38
rs28940585 Chr.7: 66082443-66082443 on Build GRCh38
rs773680222 Chr.7: 66082474-66082474 on Build GRCh38

View Full Product Details