Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66087208-66087751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASL
Entrez Gene ID
435
Gene Name
argininosuccinate lyase
Gene Aliases
ASAL
UniGene
Hs.632015
Gene Chromosome Location
Chr.7: 66075789-66093343 on Build GRCh38
Transcripts
NM_000048.3 NM_001024943.1 NM_001024944.1 NM_001024946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873232 c.617G>T p.G206V Chr.7: 66087348-66087348 on Build GRCh38
6036140 c.638G>A p.R213Q Chr.7: 66087369-66087369 on Build GRCh38
3381970 c.620A>G p.N207S Chr.7: 66087351-66087351 on Build GRCh38
6110758 c.660C>G p.L220L Chr.7: 66087733-66087733 on Build GRCh38
7171284 c.634G>C p.D212H Chr.7: 66087365-66087365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043858656 Chr.7: 66087423-66087423 on Build GRCh38
rs756638742 Chr.7: 66087715-66087715 on Build GRCh38
rs962018657 Chr.7: 66087561-66087561 on Build GRCh38
rs1007094019 Chr.7: 66087653-66087653 on Build GRCh38
rs771531271 Chr.7: 66087344-66087344 on Build GRCh38
rs901572464 Chr.7: 66087575-66087575 on Build GRCh38
rs770720936 Chr.7: 66087339-66087339 on Build GRCh38
rs774863220 Chr.7: 66087408-66087408 on Build GRCh38
rs968200125 Chr.7: 66087449-66087449 on Build GRCh38
rs761125427 Chr.7: 66087699-66087699 on Build GRCh38
rs761710556 Chr.7: 66087431-66087431 on Build GRCh38
rs369879957 Chr.7: 66087380-66087380 on Build GRCh38
rs762720984 Chr.7: 66087284-66087284 on Build GRCh38
rs773016193 Chr.7: 66087435-66087435 on Build GRCh38
rs138973946 Chr.7: 66087464-66087464 on Build GRCh38
rs745502369 Chr.7: 66087342-66087342 on Build GRCh38
rs962130781 Chr.7: 66087264-66087264 on Build GRCh38
rs60055215 Chr.7: 66087283-66087284 on Build GRCh38
rs746465424 Chr.7: 66087419-66087419 on Build GRCh38
rs571362354 Chr.7: 66087725-66087725 on Build GRCh38
rs776613796 Chr.7: 66087424-66087424 on Build GRCh38
rs892115459 Chr.7: 66087413-66087413 on Build GRCh38
rs991968140 Chr.7: 66087503-66087503 on Build GRCh38
rs313829 Chr.7: 66087510-66087510 on Build GRCh38
rs1015799966 Chr.7: 66087381-66087381 on Build GRCh38
rs985456847 Chr.7: 66087417-66087417 on Build GRCh38
rs920686074 Chr.7: 66087608-66087608 on Build GRCh38
rs372568540 Chr.7: 66087694-66087694 on Build GRCh38
rs766153120 Chr.7: 66087533-66087533 on Build GRCh38
rs765009028 Chr.7: 66087371-66087371 on Build GRCh38
rs774004637 Chr.7: 66087287-66087287 on Build GRCh38
rs756614378 Chr.7: 66087396-66087396 on Build GRCh38
rs879221934 Chr.7: 66087244-66087249 on Build GRCh38
rs865947057 Chr.7: 66087393-66087394 on Build GRCh38
rs866350721 Chr.7: 66087282-66087283 on Build GRCh38
rs369030197 Chr.7: 66087406-66087406 on Build GRCh38
rs778029825 Chr.7: 66087716-66087716 on Build GRCh38
rs778811159 Chr.7: 66087719-66087720 on Build GRCh38
rs887436276 Chr.7: 66087538-66087538 on Build GRCh38
rs751621800 Chr.7: 66087392-66087392 on Build GRCh38
rs769628469 Chr.7: 66087434-66087434 on Build GRCh38
rs541434058 Chr.7: 66087272-66087272 on Build GRCh38
rs200203985 Chr.7: 66087394-66087394 on Build GRCh38
rs753587033 Chr.7: 66087300-66087300 on Build GRCh38
rs998334666 Chr.7: 66087584-66087584 on Build GRCh38
rs755546894 Chr.7: 66087393-66087393 on Build GRCh38
rs1048752132 Chr.7: 66087514-66087514 on Build GRCh38
rs763897528 Chr.7: 66087298-66087298 on Build GRCh38
rs550365646 Chr.7: 66087502-66087502 on Build GRCh38
rs986380471 Chr.7: 66087621-66087621 on Build GRCh38
rs754763341 Chr.7: 66087302-66087302 on Build GRCh38
rs201814807 Chr.7: 66087724-66087724 on Build GRCh38
rs376008291 Chr.7: 66087314-66087314 on Build GRCh38
rs769683525 Chr.7: 66087726-66087726 on Build GRCh38
rs1042259170 Chr.7: 66087291-66087291 on Build GRCh38
rs367999318 Chr.7: 66087329-66087329 on Build GRCh38
rs763948849 Chr.7: 66087351-66087351 on Build GRCh38
rs751604140 Chr.7: 66087687-66087687 on Build GRCh38
rs867749155 Chr.7: 66087281-66087283 on Build GRCh38
rs778259514 Chr.7: 66087401-66087401 on Build GRCh38
rs753304575 Chr.7: 66087713-66087713 on Build GRCh38
rs760559303 Chr.7: 66087348-66087348 on Build GRCh38
rs541367510 Chr.7: 66087318-66087318 on Build GRCh38
rs117864317 Chr.7: 66087295-66087295 on Build GRCh38
rs895034522 Chr.7: 66087303-66087303 on Build GRCh38
rs796446156 Chr.7: 66087282-66087283 on Build GRCh38
rs796051925 Chr.7: 66087345-66087345 on Build GRCh38
rs1043358171 Chr.7: 66087708-66087708 on Build GRCh38
rs972210426 Chr.7: 66087581-66087581 on Build GRCh38
rs766396580 Chr.7: 66087385-66087385 on Build GRCh38
rs1034498654 Chr.7: 66087462-66087462 on Build GRCh38
rs768138859 Chr.7: 66087422-66087422 on Build GRCh38
rs375719771 Chr.7: 66087722-66087722 on Build GRCh38
rs765381051 Chr.7: 66087308-66087308 on Build GRCh38
rs115232785 Chr.7: 66087312-66087312 on Build GRCh38
rs978089617 Chr.7: 66087691-66087691 on Build GRCh38
rs767703228 Chr.7: 66087693-66087693 on Build GRCh38
rs946778999 Chr.7: 66087249-66087249 on Build GRCh38
rs758627014 Chr.7: 66087383-66087383 on Build GRCh38
rs903567779 Chr.7: 66087427-66087427 on Build GRCh38
rs759749791 Chr.7: 66087690-66087690 on Build GRCh38
rs944576125 Chr.7: 66087386-66087386 on Build GRCh38
rs944626973 Chr.7: 66087667-66087667 on Build GRCh38
rs868757400 Chr.7: 66087443-66087443 on Build GRCh38
rs369464777 Chr.7: 66087523-66087525 on Build GRCh38
rs748459510 Chr.7: 66087338-66087338 on Build GRCh38
rs758912888 Chr.7: 66087288-66087288 on Build GRCh38
rs60878697 Chr.7: 66087244-66087245 on Build GRCh38
rs952126896 Chr.7: 66087613-66087613 on Build GRCh38
rs781714725 Chr.7: 66087337-66087337 on Build GRCh38
rs770669613 Chr.7: 66087285-66087285 on Build GRCh38
rs776422049 Chr.7: 66087358-66087358 on Build GRCh38
rs747521107 Chr.7: 66087328-66087328 on Build GRCh38
rs1024284148 Chr.7: 66087522-66087522 on Build GRCh38
rs139105263 Chr.7: 66087340-66087340 on Build GRCh38
rs879164391 Chr.7: 66087243-66087244 on Build GRCh38
rs940327069 Chr.7: 66087544-66087544 on Build GRCh38
rs374879373 Chr.7: 66087292-66087292 on Build GRCh38
rs745526736 Chr.7: 66087402-66087402 on Build GRCh38
rs961844804 Chr.7: 66087388-66087388 on Build GRCh38
rs757473069 Chr.7: 66087302-66087302 on Build GRCh38
rs775329505 Chr.7: 66087294-66087294 on Build GRCh38
rs374259737 Chr.7: 66087723-66087723 on Build GRCh38
rs776596674 Chr.7: 66087244-66087247 on Build GRCh38
rs761651320 Chr.7: 66087368-66087368 on Build GRCh38
rs1040309625 Chr.7: 66087673-66087673 on Build GRCh38
rs915353679 Chr.7: 66087230-66087230 on Build GRCh38
rs313828 Chr.7: 66087627-66087627 on Build GRCh38
rs913603478 Chr.7: 66087677-66087677 on Build GRCh38
rs776185611 Chr.7: 66087246-66087247 on Build GRCh38
rs775013928 Chr.7: 66087346-66087346 on Build GRCh38
rs1036536909 Chr.7: 66087566-66087566 on Build GRCh38
rs766557243 Chr.7: 66087678-66087678 on Build GRCh38

View Full Product Details