Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 69599158-69599695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUTS2
Entrez Gene ID
26053
Gene Name
autism susceptibility candidate 2
Gene Aliases
FBRSL2, MRD26
UniGene
Hs.21631
Gene Chromosome Location
Chr.7: 69598475-70793068 on Build GRCh38
Transcripts
NM_001127231.2 NM_001127232.2 NM_015570.3 XM_011516010.2 XM_011516011.2 XM_011516012.2 XM_011516013.2 XM_011516014.2 XM_017011951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3750042 c.11C>T p.P4L Chr.7: 69599664-69599664 on Build GRCh38
6419453 c.14C>A p.T5K Chr.7: 69599667-69599667 on Build GRCh38
4635529 c.15G>A p.T5T Chr.7: 69599668-69599668 on Build GRCh38
4635528 c.15G>A p.T5T Chr.7: 69599668-69599668 on Build GRCh38
6419452 c.14C>A p.T5K Chr.7: 69599667-69599667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941569037 Chr.7: 69599360-69599360 on Build GRCh38
rs1050397327 Chr.7: 69599341-69599341 on Build GRCh38
rs927957148 Chr.7: 69599180-69599205 on Build GRCh38
rs993265275 Chr.7: 69599290-69599290 on Build GRCh38
rs566836925 Chr.7: 69599593-69599593 on Build GRCh38
rs992733166 Chr.7: 69599540-69599540 on Build GRCh38
rs559473184 Chr.7: 69599488-69599488 on Build GRCh38
rs1001380076 Chr.7: 69599523-69599523 on Build GRCh38
rs1020105958 Chr.7: 69599209-69599209 on Build GRCh38
rs1034658668 Chr.7: 69599525-69599525 on Build GRCh38
rs1041341061 Chr.7: 69599668-69599668 on Build GRCh38
rs985663753 Chr.7: 69599608-69599608 on Build GRCh38
rs1041495830 Chr.7: 69599486-69599486 on Build GRCh38
rs891065709 Chr.7: 69599633-69599633 on Build GRCh38
rs1014525052 Chr.7: 69599563-69599563 on Build GRCh38
rs920364812 Chr.7: 69599575-69599575 on Build GRCh38
rs959997244 Chr.7: 69599531-69599531 on Build GRCh38
rs968543349 Chr.7: 69599223-69599223 on Build GRCh38
rs978666886 Chr.7: 69599242-69599242 on Build GRCh38
rs1000053376 Chr.7: 69599520-69599520 on Build GRCh38
rs910017482 Chr.7: 69599330-69599330 on Build GRCh38
rs891828645 Chr.7: 69599440-69599440 on Build GRCh38
rs540754651 Chr.7: 69599453-69599453 on Build GRCh38
rs867140240 Chr.7: 69599445-69599445 on Build GRCh38
rs961726252 Chr.7: 69599569-69599569 on Build GRCh38
rs576514915 Chr.7: 69599431-69599431 on Build GRCh38
rs776323975 Chr.7: 69599395-69599395 on Build GRCh38
rs917268525 Chr.7: 69599294-69599294 on Build GRCh38
rs144481048 Chr.7: 69599484-69599485 on Build GRCh38
rs1041076989 Chr.7: 69599438-69599438 on Build GRCh38
rs943995195 Chr.7: 69599392-69599392 on Build GRCh38
rs890232996 Chr.7: 69599184-69599184 on Build GRCh38
rs200189077 Chr.7: 69599664-69599664 on Build GRCh38
rs3735260 Chr.7: 69599651-69599651 on Build GRCh38
rs958715483 Chr.7: 69599268-69599268 on Build GRCh38
rs973119300 Chr.7: 69599570-69599570 on Build GRCh38
rs1010004136 Chr.7: 69599191-69599191 on Build GRCh38
rs535698149 Chr.7: 69599301-69599301 on Build GRCh38
rs534062236 Chr.7: 69599612-69599612 on Build GRCh38
rs1014188084 Chr.7: 69599586-69599586 on Build GRCh38
rs890044778 Chr.7: 69599379-69599379 on Build GRCh38
rs1034719258 Chr.7: 69599246-69599246 on Build GRCh38
rs921538419 Chr.7: 69599400-69599400 on Build GRCh38
rs931531662 Chr.7: 69599202-69599202 on Build GRCh38
rs529864188 Chr.7: 69599496-69599496 on Build GRCh38
rs904323092 Chr.7: 69599517-69599517 on Build GRCh38
rs982830579 Chr.7: 69599315-69599315 on Build GRCh38
rs930026600 Chr.7: 69599307-69599310 on Build GRCh38

View Full Product Details