Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 70764668-70765181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUTS2
Entrez Gene ID
26053
Gene Name
autism susceptibility candidate 2
Gene Aliases
FBRSL2, MRD26
UniGene
Hs.21631
Gene Chromosome Location
Chr.7: 69598475-70793068 on Build GRCh38
Transcripts
NM_001127231.2 NM_015570.3 XM_005250257.2 XM_011516010.2 XM_011516011.2 XM_011516012.2 XM_011516013.2 XM_011516014.2 XM_011516017.2 XM_011516018.2 XM_017011951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776437 c.1468+1G>A p.? Chr.7: 70765006-70765006 on Build GRCh38
1091434 c.1316G>A p.S439N Chr.7: 70764853-70764853 on Build GRCh38
1091433 c.1252G>A p.A418T Chr.7: 70764789-70764789 on Build GRCh38
6436653 c.1271A>C p.H424P Chr.7: 70764808-70764808 on Build GRCh38
368980 c.1468+1G>T p.? Chr.7: 70765006-70765006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758059365 Chr.7: 70765039-70765039 on Build GRCh38
rs970531724 Chr.7: 70765119-70765119 on Build GRCh38
rs534340679 Chr.7: 70765091-70765091 on Build GRCh38
rs13228988 Chr.7: 70764751-70764751 on Build GRCh38
rs978681055 Chr.7: 70765154-70765154 on Build GRCh38
rs757228680 Chr.7: 70765033-70765033 on Build GRCh38
rs572933185 Chr.7: 70764808-70764808 on Build GRCh38
rs763397328 Chr.7: 70765020-70765020 on Build GRCh38
rs748069802 Chr.7: 70764766-70764766 on Build GRCh38
rs780713384 Chr.7: 70764886-70764886 on Build GRCh38
rs760653347 Chr.7: 70764926-70764926 on Build GRCh38
rs755576516 Chr.7: 70764983-70764983 on Build GRCh38
rs1004758747 Chr.7: 70765133-70765133 on Build GRCh38
rs772174582 Chr.7: 70764921-70764921 on Build GRCh38
rs67892322 Chr.7: 70764729-70764729 on Build GRCh38
rs746581320 Chr.7: 70764742-70764742 on Build GRCh38
rs1006308561 Chr.7: 70764704-70764704 on Build GRCh38
rs953027719 Chr.7: 70764795-70764796 on Build GRCh38
rs779837197 Chr.7: 70764739-70764739 on Build GRCh38
rs143679158 Chr.7: 70764784-70764784 on Build GRCh38
rs775766319 Chr.7: 70764977-70764977 on Build GRCh38
rs780131403 Chr.7: 70764991-70764991 on Build GRCh38
rs761208105 Chr.7: 70764835-70764835 on Build GRCh38
rs749176066 Chr.7: 70764891-70764891 on Build GRCh38
rs765645776 Chr.7: 70764947-70764947 on Build GRCh38
rs770649701 Chr.7: 70764896-70764896 on Build GRCh38
rs573827140 Chr.7: 70765126-70765126 on Build GRCh38
rs917640717 Chr.7: 70764709-70764709 on Build GRCh38
rs759619512 Chr.7: 70764831-70764831 on Build GRCh38
rs776652690 Chr.7: 70764884-70764885 on Build GRCh38
rs369919917 Chr.7: 70764985-70764985 on Build GRCh38
rs768557523 Chr.7: 70765008-70765008 on Build GRCh38
rs770968653 Chr.7: 70764726-70764726 on Build GRCh38
rs773760057 Chr.7: 70765018-70765018 on Build GRCh38
rs750280573 Chr.7: 70765037-70765037 on Build GRCh38
rs767056271 Chr.7: 70764978-70764978 on Build GRCh38
rs549088098 Chr.7: 70765129-70765129 on Build GRCh38
rs145730337 Chr.7: 70765088-70765088 on Build GRCh38
rs370414676 Chr.7: 70764747-70764747 on Build GRCh38
rs567637069 Chr.7: 70765136-70765136 on Build GRCh38
rs772392460 Chr.7: 70764990-70764990 on Build GRCh38
rs1016368213 Chr.7: 70765083-70765083 on Build GRCh38
rs528878518 Chr.7: 70764989-70764989 on Build GRCh38
rs758169328 Chr.7: 70764731-70764731 on Build GRCh38
rs1035401310 Chr.7: 70765109-70765109 on Build GRCh38
rs1056205169 Chr.7: 70765086-70765086 on Build GRCh38
rs556918487 Chr.7: 70764716-70764716 on Build GRCh38
rs746046031 Chr.7: 70764919-70764919 on Build GRCh38
rs374122014 Chr.7: 70765066-70765066 on Build GRCh38
rs57488007 Chr.7: 70764858-70764859 on Build GRCh38
rs369831199 Chr.7: 70765009-70765009 on Build GRCh38
rs900790027 Chr.7: 70764794-70764794 on Build GRCh38
rs148916760 Chr.7: 70765142-70765142 on Build GRCh38
rs773100260 Chr.7: 70764788-70764788 on Build GRCh38
rs1010288143 Chr.7: 70764902-70764902 on Build GRCh38
rs756126420 Chr.7: 70764890-70764890 on Build GRCh38
rs116591851 Chr.7: 70765028-70765028 on Build GRCh38
rs377507477 Chr.7: 70764986-70764986 on Build GRCh38
rs376584055 Chr.7: 70764801-70764801 on Build GRCh38
rs774553373 Chr.7: 70764826-70764826 on Build GRCh38
rs747142063 Chr.7: 70764710-70764710 on Build GRCh38
rs751287276 Chr.7: 70764956-70764956 on Build GRCh38
rs771202006 Chr.7: 70764805-70764805 on Build GRCh38
rs201372643 Chr.7: 70764881-70764881 on Build GRCh38
rs754117579 Chr.7: 70764730-70764730 on Build GRCh38
rs571670071 Chr.7: 70765047-70765047 on Build GRCh38
rs904502698 Chr.7: 70764936-70764947 on Build GRCh38
rs765141969 Chr.7: 70765036-70765036 on Build GRCh38
rs748216311 Chr.7: 70765053-70765053 on Build GRCh38
rs776933516 Chr.7: 70764940-70764940 on Build GRCh38
rs764482237 Chr.7: 70764847-70764847 on Build GRCh38
rs752117240 Chr.7: 70764887-70764887 on Build GRCh38
rs774940823 Chr.7: 70765026-70765026 on Build GRCh38
rs1012186178 Chr.7: 70765093-70765093 on Build GRCh38
rs560622993 Chr.7: 70764834-70764834 on Build GRCh38
rs770393819 Chr.7: 70765013-70765013 on Build GRCh38
rs971771415 Chr.7: 70764740-70764740 on Build GRCh38
rs1035517466 Chr.7: 70764781-70764781 on Build GRCh38
rs1000641183 Chr.7: 70764965-70764965 on Build GRCh38
rs370647270 Chr.7: 70764836-70764836 on Build GRCh38
rs767529359 Chr.7: 70764832-70764832 on Build GRCh38
rs752310034 Chr.7: 70764979-70764979 on Build GRCh38
rs1005779686 Chr.7: 70765081-70765081 on Build GRCh38
rs754469709 Chr.7: 70764746-70764746 on Build GRCh38
rs751197272 Chr.7: 70764880-70764880 on Build GRCh38
rs1015902790 Chr.7: 70764710-70764710 on Build GRCh38
rs1013171425 Chr.7: 70765132-70765132 on Build GRCh38
rs769167826 Chr.7: 70765046-70765046 on Build GRCh38
rs374398806 Chr.7: 70765012-70765012 on Build GRCh38
rs534028659 Chr.7: 70765071-70765071 on Build GRCh38
rs374318676 Chr.7: 70764856-70764856 on Build GRCh38
rs370278297 Chr.7: 70765029-70765029 on Build GRCh38
rs762289043 Chr.7: 70765054-70765054 on Build GRCh38
rs762261704 Chr.7: 70764876-70764876 on Build GRCh38
rs1983488 Chr.7: 70764699-70764699 on Build GRCh38
rs1057518198 Chr.7: 70764835-70764835 on Build GRCh38
rs754282610 Chr.7: 70764853-70764853 on Build GRCh38
rs781462397 Chr.7: 70765049-70765049 on Build GRCh38
rs760875170 Chr.7: 70765084-70765084 on Build GRCh38
rs762280300 Chr.7: 70764941-70764941 on Build GRCh38
rs375248939 Chr.7: 70764924-70764924 on Build GRCh38
rs371714496 Chr.7: 70764879-70764879 on Build GRCh38
rs922167941 Chr.7: 70765032-70765032 on Build GRCh38
rs143055091 Chr.7: 70765001-70765001 on Build GRCh38
rs200026249 Chr.7: 70764910-70764910 on Build GRCh38
rs1041778868 Chr.7: 70765056-70765056 on Build GRCh38
rs769270137 Chr.7: 70764931-70764931 on Build GRCh38
rs780333398 Chr.7: 70765040-70765040 on Build GRCh38
rs767268365 Chr.7: 70765025-70765025 on Build GRCh38
rs796406304 Chr.7: 70765087-70765087 on Build GRCh38

View Full Product Details