Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 71781009-71781519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALN1
Entrez Gene ID
83698
Gene Name
calneuron 1
Gene Aliases
CABP8
UniGene
Hs.592750
Gene Chromosome Location
Chr.7: 71779491-72504271 on Build GRCh38
Transcripts
NM_001017440.2 NM_031468.3 XM_011516594.2 XM_011516595.1 XM_011516596.2 XM_011516597.2 XM_011516599.1 XM_017012675.1 XM_017012676.1 XM_017012677.1 XM_017012678.1 XM_017012679.1 XM_017012680.1 XM_017012681.1 XM_017012682.1 XM_017012683.1

SNP Detail

SNP ID Chromosome Location
rs749459897 Chr.7: 71781492-71781492 on Build GRCh38
rs764756192 Chr.7: 71781301-71781301 on Build GRCh38
rs534481874 Chr.7: 71781379-71781379 on Build GRCh38
rs150960157 Chr.7: 71781246-71781246 on Build GRCh38
rs561933893 Chr.7: 71781471-71781471 on Build GRCh38
rs1024710586 Chr.7: 71781043-71781043 on Build GRCh38
rs189876541 Chr.7: 71781184-71781184 on Build GRCh38
rs752258250 Chr.7: 71781314-71781314 on Build GRCh38
rs961429919 Chr.7: 71781272-71781272 on Build GRCh38
rs965905091 Chr.7: 71781330-71781330 on Build GRCh38
rs764349047 Chr.7: 71781353-71781353 on Build GRCh38
rs987630534 Chr.7: 71781197-71781197 on Build GRCh38
rs1048815849 Chr.7: 71781304-71781304 on Build GRCh38
rs533341222 Chr.7: 71781412-71781412 on Build GRCh38
rs776562673 Chr.7: 71781224-71781224 on Build GRCh38
rs975211367 Chr.7: 71781457-71781457 on Build GRCh38
rs987641789 Chr.7: 71781444-71781444 on Build GRCh38
rs756234199 Chr.7: 71781483-71781483 on Build GRCh38
rs914139417 Chr.7: 71781171-71781171 on Build GRCh38
rs544759045 Chr.7: 71781450-71781450 on Build GRCh38
rs771567211 Chr.7: 71781340-71781340 on Build GRCh38
rs766236884 Chr.7: 71781078-71781078 on Build GRCh38
rs112171100 Chr.7: 71781437-71781437 on Build GRCh38
rs781747960 Chr.7: 71781451-71781451 on Build GRCh38
rs1033726790 Chr.7: 71781093-71781093 on Build GRCh38
rs1002330299 Chr.7: 71781351-71781351 on Build GRCh38
rs990115021 Chr.7: 71781164-71781164 on Build GRCh38
rs1016066159 Chr.7: 71781269-71781269 on Build GRCh38
rs575397876 Chr.7: 71781473-71781473 on Build GRCh38
rs966895825 Chr.7: 71781173-71781173 on Build GRCh38
rs916218067 Chr.7: 71781447-71781447 on Build GRCh38
rs78074718 Chr.7: 71781453-71781453 on Build GRCh38
rs920565062 Chr.7: 71781293-71781293 on Build GRCh38
rs983464846 Chr.7: 71781484-71781484 on Build GRCh38
rs923647461 Chr.7: 71781175-71781175 on Build GRCh38
rs140759012 Chr.7: 71781321-71781321 on Build GRCh38
rs906297537 Chr.7: 71781345-71781345 on Build GRCh38
rs1002979363 Chr.7: 71781247-71781250 on Build GRCh38
rs758995934 Chr.7: 71781260-71781260 on Build GRCh38
rs116744034 Chr.7: 71781300-71781300 on Build GRCh38
rs1001826008 Chr.7: 71781067-71781067 on Build GRCh38
rs370382206 Chr.7: 71781138-71781138 on Build GRCh38
rs954236207 Chr.7: 71781055-71781055 on Build GRCh38
rs912021300 Chr.7: 71781206-71781206 on Build GRCh38
rs567700930 Chr.7: 71781107-71781107 on Build GRCh38
rs145790107 Chr.7: 71781358-71781358 on Build GRCh38
rs528980100 Chr.7: 71781150-71781150 on Build GRCh38
rs970447259 Chr.7: 71781054-71781054 on Build GRCh38
rs1012122617 Chr.7: 71781223-71781223 on Build GRCh38
rs138634296 Chr.7: 71781413-71781413 on Build GRCh38
rs957965629 Chr.7: 71781380-71781380 on Build GRCh38
rs943395076 Chr.7: 71781232-71781232 on Build GRCh38
rs958326190 Chr.7: 71781163-71781163 on Build GRCh38
rs920674339 Chr.7: 71781479-71781479 on Build GRCh38
rs544305449 Chr.7: 71781287-71781287 on Build GRCh38
rs943810079 Chr.7: 71781151-71781151 on Build GRCh38
rs79577030 Chr.7: 71781488-71781488 on Build GRCh38
rs1027233563 Chr.7: 71781303-71781303 on Build GRCh38

View Full Product Details