Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 71782019-71782561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALN1
Entrez Gene ID
83698
Gene Name
calneuron 1
Gene Aliases
CABP8
UniGene
Hs.592750
Gene Chromosome Location
Chr.7: 71779491-72504271 on Build GRCh38
Transcripts
NM_001017440.2 NM_031468.3 XM_011516594.2 XM_011516595.1 XM_011516596.2 XM_011516597.2 XM_011516599.1 XM_017012675.1 XM_017012676.1 XM_017012677.1 XM_017012678.1 XM_017012679.1 XM_017012680.1 XM_017012681.1 XM_017012682.1 XM_017012683.1

SNP Detail

SNP ID Chromosome Location
rs756745006 Chr.7: 71782238-71782238 on Build GRCh38
rs915050935 Chr.7: 71782210-71782210 on Build GRCh38
rs942794182 Chr.7: 71782131-71782131 on Build GRCh38
rs77762702 Chr.7: 71782437-71782437 on Build GRCh38
rs750944822 Chr.7: 71782236-71782236 on Build GRCh38
rs554227560 Chr.7: 71782465-71782465 on Build GRCh38
rs10275990 Chr.7: 71782150-71782150 on Build GRCh38
rs1014996198 Chr.7: 71782335-71782335 on Build GRCh38
rs1005045999 Chr.7: 71782539-71782539 on Build GRCh38
rs1053140605 Chr.7: 71782192-71782192 on Build GRCh38
rs1042186766 Chr.7: 71782234-71782234 on Build GRCh38
rs927909613 Chr.7: 71782125-71782125 on Build GRCh38
rs920510388 Chr.7: 71782087-71782087 on Build GRCh38
rs2270209 Chr.7: 71782266-71782266 on Build GRCh38
rs1056637940 Chr.7: 71782530-71782530 on Build GRCh38
rs762231086 Chr.7: 71782097-71782097 on Build GRCh38
rs184451757 Chr.7: 71782436-71782436 on Build GRCh38
rs911339181 Chr.7: 71782113-71782113 on Build GRCh38
rs931620391 Chr.7: 71782500-71782500 on Build GRCh38
rs781010555 Chr.7: 71782474-71782474 on Build GRCh38
rs905321510 Chr.7: 71782237-71782237 on Build GRCh38
rs79350211 Chr.7: 71782430-71782430 on Build GRCh38
rs143533016 Chr.7: 71782045-71782045 on Build GRCh38
rs76347112 Chr.7: 71782426-71782426 on Build GRCh38
rs977912693 Chr.7: 71782464-71782464 on Build GRCh38
rs572707372 Chr.7: 71782489-71782489 on Build GRCh38
rs150799301 Chr.7: 71782049-71782049 on Build GRCh38
rs118054711 Chr.7: 71782183-71782183 on Build GRCh38
rs946477844 Chr.7: 71782213-71782213 on Build GRCh38
rs972403348 Chr.7: 71782055-71782055 on Build GRCh38
rs111947289 Chr.7: 71782318-71782318 on Build GRCh38
rs1034321410 Chr.7: 71782421-71782421 on Build GRCh38
rs549701204 Chr.7: 71782358-71782358 on Build GRCh38
rs1004993314 Chr.7: 71782316-71782316 on Build GRCh38
rs866499665 Chr.7: 71782475-71782475 on Build GRCh38
rs538800980 Chr.7: 71782461-71782461 on Build GRCh38
rs951902262 Chr.7: 71782096-71782096 on Build GRCh38
rs910405727 Chr.7: 71782294-71782296 on Build GRCh38
rs60710563 Chr.7: 71782292-71782292 on Build GRCh38
rs1002830860 Chr.7: 71782405-71782405 on Build GRCh38
rs921588333 Chr.7: 71782499-71782499 on Build GRCh38
rs1041114254 Chr.7: 71782137-71782137 on Build GRCh38
rs745789649 Chr.7: 71782531-71782531 on Build GRCh38
rs779147237 Chr.7: 71782291-71782291 on Build GRCh38
rs60030903 Chr.7: 71782282-71782282 on Build GRCh38
rs779993505 Chr.7: 71782265-71782265 on Build GRCh38
rs888018293 Chr.7: 71782538-71782538 on Build GRCh38
rs935372249 Chr.7: 71782379-71782379 on Build GRCh38
rs900757144 Chr.7: 71782270-71782270 on Build GRCh38
rs772383401 Chr.7: 71782327-71782328 on Build GRCh38
rs967904006 Chr.7: 71782462-71782462 on Build GRCh38
rs944401211 Chr.7: 71782480-71782480 on Build GRCh38

View Full Product Details