Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 71783533-71784034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALN1
Entrez Gene ID
83698
Gene Name
calneuron 1
Gene Aliases
CABP8
UniGene
Hs.592750
Gene Chromosome Location
Chr.7: 71779491-72504271 on Build GRCh38
Transcripts
NM_001017440.2 NM_031468.3 XM_011516594.2 XM_011516595.1 XM_011516596.2 XM_011516597.2 XM_011516599.1 XM_017012675.1 XM_017012676.1 XM_017012677.1 XM_017012678.1 XM_017012679.1 XM_017012680.1 XM_017012681.1 XM_017012682.1 XM_017012683.1

SNP Detail

SNP ID Chromosome Location
rs895611056 Chr.7: 71783656-71783656 on Build GRCh38
rs900740868 Chr.7: 71783713-71783713 on Build GRCh38
rs749239371 Chr.7: 71783752-71783752 on Build GRCh38
rs868068437 Chr.7: 71783640-71783640 on Build GRCh38
rs760009438 Chr.7: 71783582-71783582 on Build GRCh38
rs1037671208 Chr.7: 71783833-71783833 on Build GRCh38
rs7809896 Chr.7: 71783715-71783715 on Build GRCh38
rs1016508663 Chr.7: 71783711-71783711 on Build GRCh38
rs537998987 Chr.7: 71783644-71783644 on Build GRCh38
rs147781968 Chr.7: 71783778-71783778 on Build GRCh38
rs927192872 Chr.7: 71783626-71783626 on Build GRCh38
rs949520261 Chr.7: 71783664-71783664 on Build GRCh38
rs758475722 Chr.7: 71783669-71783669 on Build GRCh38
rs10225199 Chr.7: 71783981-71783981 on Build GRCh38
rs922571484 Chr.7: 71783593-71783593 on Build GRCh38
rs878991930 Chr.7: 71783719-71783719 on Build GRCh38
rs114008942 Chr.7: 71783815-71783815 on Build GRCh38
rs796590542 Chr.7: 71783622-71783640 on Build GRCh38
rs112694990 Chr.7: 71783800-71783800 on Build GRCh38
rs1031813570 Chr.7: 71783839-71783839 on Build GRCh38
rs754999826 Chr.7: 71783575-71783575 on Build GRCh38
rs370266239 Chr.7: 71783651-71783651 on Build GRCh38
rs1051993367 Chr.7: 71783854-71783854 on Build GRCh38
rs548267859 Chr.7: 71783561-71783561 on Build GRCh38
rs968465406 Chr.7: 71783937-71783937 on Build GRCh38
rs538540513 Chr.7: 71783710-71783710 on Build GRCh38
rs189986140 Chr.7: 71783861-71783861 on Build GRCh38
rs1010414321 Chr.7: 71783742-71783742 on Build GRCh38
rs754938163 Chr.7: 71783793-71783793 on Build GRCh38
rs746415494 Chr.7: 71783884-71783884 on Build GRCh38
rs978702683 Chr.7: 71783620-71783620 on Build GRCh38
rs567360045 Chr.7: 71783612-71783612 on Build GRCh38
rs541325173 Chr.7: 71783829-71783829 on Build GRCh38
rs554661851 Chr.7: 71783766-71783766 on Build GRCh38
rs889031745 Chr.7: 71783652-71783652 on Build GRCh38
rs186051918 Chr.7: 71783716-71783716 on Build GRCh38
rs3801156 Chr.7: 71783691-71783691 on Build GRCh38
rs954876896 Chr.7: 71783726-71783727 on Build GRCh38
rs932650841 Chr.7: 71783598-71783598 on Build GRCh38
rs890646246 Chr.7: 71783863-71783863 on Build GRCh38
rs544618000 Chr.7: 71783678-71783678 on Build GRCh38
rs548279558 Chr.7: 71784003-71784003 on Build GRCh38
rs934697202 Chr.7: 71783564-71783564 on Build GRCh38
rs926912550 Chr.7: 71783768-71783768 on Build GRCh38
rs1047827751 Chr.7: 71783827-71783827 on Build GRCh38
rs372415877 Chr.7: 71783622-71783639 on Build GRCh38
rs376182911 Chr.7: 71783592-71783592 on Build GRCh38
rs1037459751 Chr.7: 71783675-71783675 on Build GRCh38
rs922697193 Chr.7: 71783916-71783916 on Build GRCh38
rs1010390670 Chr.7: 71783915-71783915 on Build GRCh38
rs1034035367 Chr.7: 71783992-71783992 on Build GRCh38
rs565324800 Chr.7: 71783806-71783806 on Build GRCh38
rs947005382 Chr.7: 71783619-71783619 on Build GRCh38
rs900458949 Chr.7: 71783837-71783837 on Build GRCh38
rs1006158304 Chr.7: 71783709-71783709 on Build GRCh38
rs988538769 Chr.7: 71783591-71783591 on Build GRCh38
rs1020081116 Chr.7: 71783922-71783922 on Build GRCh38
rs562840131 Chr.7: 71783832-71783832 on Build GRCh38
rs752837777 Chr.7: 71783628-71783628 on Build GRCh38
rs747605439 Chr.7: 71783878-71783878 on Build GRCh38
rs958371879 Chr.7: 71783993-71783993 on Build GRCh38
rs978417877 Chr.7: 71783767-71783767 on Build GRCh38
rs958876258 Chr.7: 71783792-71783792 on Build GRCh38

View Full Product Details