Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73434463-73434983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD9
Entrez Gene ID
8326
Gene Name
frizzled class receptor 9
Gene Aliases
CD349, FZD3
UniGene
Hs.647029
Gene Chromosome Location
Chr.7: 73433779-73436120 on Build GRCh38
Transcripts
NM_003508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1451961 c.861C>T p.A287A Chr.7: 73434868-73434868 on Build GRCh38
4685999 c.873C>T p.S291S Chr.7: 73434880-73434880 on Build GRCh38
6668229 c.874G>A p.V292M Chr.7: 73434881-73434881 on Build GRCh38
5672948 c.831C>G p.Y277* Chr.7: 73434838-73434838 on Build GRCh38
747181 c.815T>A p.M272K Chr.7: 73434822-73434822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782026570 Chr.7: 73434868-73434868 on Build GRCh38
rs781938252 Chr.7: 73434592-73434592 on Build GRCh38
rs782604885 Chr.7: 73434904-73434904 on Build GRCh38
rs977938373 Chr.7: 73434928-73434928 on Build GRCh38
rs782076832 Chr.7: 73434940-73434940 on Build GRCh38
rs971589447 Chr.7: 73434681-73434681 on Build GRCh38
rs781853577 Chr.7: 73434605-73434605 on Build GRCh38
rs536929629 Chr.7: 73434524-73434524 on Build GRCh38
rs575621716 Chr.7: 73434582-73434582 on Build GRCh38
rs782580729 Chr.7: 73434633-73434633 on Build GRCh38
rs376132966 Chr.7: 73434800-73434800 on Build GRCh38
rs782330628 Chr.7: 73434506-73434506 on Build GRCh38
rs1034602722 Chr.7: 73434781-73434781 on Build GRCh38
rs531718356 Chr.7: 73434770-73434770 on Build GRCh38
rs782281964 Chr.7: 73434927-73434927 on Build GRCh38
rs782170377 Chr.7: 73434714-73434714 on Build GRCh38
rs782459639 Chr.7: 73434901-73434901 on Build GRCh38
rs782767239 Chr.7: 73434739-73434739 on Build GRCh38
rs73134914 Chr.7: 73434891-73434891 on Build GRCh38
rs782136427 Chr.7: 73434606-73434606 on Build GRCh38
rs141168260 Chr.7: 73434697-73434697 on Build GRCh38
rs781980067 Chr.7: 73434947-73434947 on Build GRCh38
rs782300865 Chr.7: 73434581-73434581 on Build GRCh38
rs781960801 Chr.7: 73434906-73434906 on Build GRCh38
rs782133022 Chr.7: 73434959-73434959 on Build GRCh38
rs138414411 Chr.7: 73434563-73434563 on Build GRCh38
rs782599690 Chr.7: 73434788-73434788 on Build GRCh38
rs376108024 Chr.7: 73434797-73434797 on Build GRCh38
rs782340783 Chr.7: 73434686-73434686 on Build GRCh38
rs946134081 Chr.7: 73434911-73434911 on Build GRCh38
rs782384125 Chr.7: 73434547-73434547 on Build GRCh38
rs782180751 Chr.7: 73434505-73434505 on Build GRCh38
rs147849239 Chr.7: 73434661-73434661 on Build GRCh38
rs782659170 Chr.7: 73434799-73434799 on Build GRCh38
rs375403747 Chr.7: 73434632-73434632 on Build GRCh38
rs782562809 Chr.7: 73434682-73434682 on Build GRCh38
rs769369970 Chr.7: 73434573-73434573 on Build GRCh38
rs781785546 Chr.7: 73434761-73434761 on Build GRCh38
rs373157288 Chr.7: 73434779-73434779 on Build GRCh38
rs782119421 Chr.7: 73434851-73434851 on Build GRCh38
rs782712314 Chr.7: 73434880-73434880 on Build GRCh38
rs781869354 Chr.7: 73434886-73434886 on Build GRCh38
rs782039651 Chr.7: 73434873-73434873 on Build GRCh38
rs781942977 Chr.7: 73434724-73434724 on Build GRCh38
rs782228364 Chr.7: 73434793-73434793 on Build GRCh38
rs782761805 Chr.7: 73434884-73434884 on Build GRCh38
rs782085952 Chr.7: 73434735-73434735 on Build GRCh38
rs782413002 Chr.7: 73434670-73434670 on Build GRCh38
rs782385561 Chr.7: 73434700-73434700 on Build GRCh38
rs782023448 Chr.7: 73434704-73434704 on Build GRCh38
rs782546523 Chr.7: 73434890-73434890 on Build GRCh38
rs115007009 Chr.7: 73434842-73434842 on Build GRCh38
rs782203045 Chr.7: 73434832-73434832 on Build GRCh38
rs782795346 Chr.7: 73434610-73434610 on Build GRCh38
rs781793071 Chr.7: 73434900-73434900 on Build GRCh38
rs782651060 Chr.7: 73434660-73434660 on Build GRCh38
rs1015078235 Chr.7: 73434675-73434675 on Build GRCh38
rs782778281 Chr.7: 73434858-73434858 on Build GRCh38
rs781923345 Chr.7: 73434808-73434808 on Build GRCh38
rs781828087 Chr.7: 73434644-73434644 on Build GRCh38
rs782077630 Chr.7: 73434602-73434602 on Build GRCh38
rs17852398 Chr.7: 73434603-73434603 on Build GRCh38
rs782193718 Chr.7: 73434685-73434685 on Build GRCh38
rs782382695 Chr.7: 73434907-73434907 on Build GRCh38
rs782805481 Chr.7: 73434757-73434757 on Build GRCh38
rs782357130 Chr.7: 73434944-73434944 on Build GRCh38
rs782760906 Chr.7: 73434604-73434604 on Build GRCh38
rs782237874 Chr.7: 73434905-73434905 on Build GRCh38
rs781861224 Chr.7: 73434745-73434745 on Build GRCh38
rs782486490 Chr.7: 73434652-73434652 on Build GRCh38
rs782243429 Chr.7: 73434698-73434698 on Build GRCh38
rs558152721 Chr.7: 73434758-73434758 on Build GRCh38
rs782703347 Chr.7: 73434716-73434716 on Build GRCh38
rs782538318 Chr.7: 73434746-73434746 on Build GRCh38
rs781907051 Chr.7: 73434613-73434613 on Build GRCh38
rs782657739 Chr.7: 73434914-73434914 on Build GRCh38
rs781928891 Chr.7: 73434939-73434939 on Build GRCh38
rs570248551 Chr.7: 73434529-73434529 on Build GRCh38
rs567917703 Chr.7: 73434881-73434881 on Build GRCh38
rs782692525 Chr.7: 73434497-73434497 on Build GRCh38

View Full Product Details