Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73435998-73436496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD9
Entrez Gene ID
8326
Gene Name
frizzled class receptor 9
Gene Aliases
CD349, FZD3
UniGene
Hs.647029
Gene Chromosome Location
Chr.7: 73433779-73436120 on Build GRCh38
Transcripts
NM_003508.2

SNP Detail

SNP ID Chromosome Location
rs982024180 Chr.7: 73436314-73436314 on Build GRCh38
rs1005658496 Chr.7: 73436286-73436286 on Build GRCh38
rs532360992 Chr.7: 73436244-73436244 on Build GRCh38
rs782054830 Chr.7: 73436243-73436243 on Build GRCh38
rs575534859 Chr.7: 73436147-73436147 on Build GRCh38
rs563308060 Chr.7: 73436028-73436028 on Build GRCh38
rs372383503 Chr.7: 73436041-73436041 on Build GRCh38
rs961572082 Chr.7: 73436045-73436045 on Build GRCh38
rs377119475 Chr.7: 73436225-73436225 on Build GRCh38
rs151034742 Chr.7: 73436047-73436047 on Build GRCh38
rs888546769 Chr.7: 73436261-73436261 on Build GRCh38
rs750504116 Chr.7: 73436185-73436185 on Build GRCh38
rs575772278 Chr.7: 73436079-73436079 on Build GRCh38
rs1034938916 Chr.7: 73436335-73436335 on Build GRCh38
rs927607632 Chr.7: 73436083-73436083 on Build GRCh38
rs914983937 Chr.7: 73436112-73436112 on Build GRCh38
rs192149363 Chr.7: 73436463-73436463 on Build GRCh38
rs541019597 Chr.7: 73436363-73436363 on Build GRCh38
rs782107236 Chr.7: 73436358-73436358 on Build GRCh38
rs782266329 Chr.7: 73436107-73436107 on Build GRCh38
rs914976360 Chr.7: 73436379-73436379 on Build GRCh38
rs781961546 Chr.7: 73436340-73436340 on Build GRCh38
rs937700444 Chr.7: 73436095-73436095 on Build GRCh38
rs1015831143 Chr.7: 73436289-73436289 on Build GRCh38
rs968186400 Chr.7: 73436421-73436421 on Build GRCh38
rs35122531 Chr.7: 73436360-73436361 on Build GRCh38
rs149791898 Chr.7: 73436238-73436238 on Build GRCh38
rs990500208 Chr.7: 73436371-73436371 on Build GRCh38
rs961268794 Chr.7: 73436293-73436293 on Build GRCh38
rs782038311 Chr.7: 73436176-73436176 on Build GRCh38
rs573101757 Chr.7: 73436305-73436305 on Build GRCh38
rs559718462 Chr.7: 73436440-73436440 on Build GRCh38
rs903378338 Chr.7: 73436208-73436208 on Build GRCh38
rs1041318963 Chr.7: 73436228-73436228 on Build GRCh38
rs1043637300 Chr.7: 73436184-73436184 on Build GRCh38
rs139709248 Chr.7: 73436143-73436143 on Build GRCh38

View Full Product Details