Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73537015-73537511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL7B
Entrez Gene ID
9275
Gene Name
BCL tumor suppressor 7B
Gene Aliases
-
UniGene
Hs.647051
Gene Chromosome Location
Chr.7: 73536353-73557735 on Build GRCh38
Transcripts
NM_001197244.1 NM_001301061.1 NM_001707.3 NR_036682.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5450398 c.581C>T p.T194I Chr.7: 73537326-73537326 on Build GRCh38
353540 c.609G>A p.*203* Chr.7: 73537298-73537298 on Build GRCh38
4567062 c.551_552CC>TT p.P184L Chr.7: 73537355-73537356 on Build GRCh38
6610769 c.585G>A p.V195V Chr.7: 73537322-73537322 on Build GRCh38
6034447 c.587C>T p.P196L Chr.7: 73537320-73537320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978834449 Chr.7: 73537485-73537485 on Build GRCh38
rs782738627 Chr.7: 73537261-73537262 on Build GRCh38
rs782342027 Chr.7: 73537278-73537278 on Build GRCh38
rs781978459 Chr.7: 73537270-73537270 on Build GRCh38
rs782452325 Chr.7: 73537359-73537359 on Build GRCh38
rs34252694 Chr.7: 73537438-73537439 on Build GRCh38
rs782026116 Chr.7: 73537314-73537314 on Build GRCh38
rs1042700476 Chr.7: 73537243-73537243 on Build GRCh38
rs373582004 Chr.7: 73537289-73537289 on Build GRCh38
rs915826794 Chr.7: 73537470-73537470 on Build GRCh38
rs782039889 Chr.7: 73537405-73537405 on Build GRCh38
rs755853719 Chr.7: 73537095-73537095 on Build GRCh38
rs778427893 Chr.7: 73537104-73537104 on Build GRCh38
rs782140570 Chr.7: 73537288-73537288 on Build GRCh38
rs938511475 Chr.7: 73537184-73537184 on Build GRCh38
rs782079710 Chr.7: 73537423-73537423 on Build GRCh38
rs782292155 Chr.7: 73537267-73537267 on Build GRCh38
rs777561329 Chr.7: 73537097-73537097 on Build GRCh38
rs782530597 Chr.7: 73537376-73537376 on Build GRCh38
rs781825282 Chr.7: 73537358-73537358 on Build GRCh38
rs782643696 Chr.7: 73537375-73537375 on Build GRCh38
rs748613410 Chr.7: 73537215-73537215 on Build GRCh38
rs745352851 Chr.7: 73537106-73537107 on Build GRCh38
rs782715921 Chr.7: 73537293-73537293 on Build GRCh38
rs80056196 Chr.7: 73537111-73537111 on Build GRCh38
rs748955585 Chr.7: 73537100-73537102 on Build GRCh38
rs1029239264 Chr.7: 73537140-73537140 on Build GRCh38
rs997804790 Chr.7: 73537130-73537130 on Build GRCh38
rs138977022 Chr.7: 73537176-73537176 on Build GRCh38
rs782366199 Chr.7: 73537411-73537411 on Build GRCh38
rs373913398 Chr.7: 73537281-73537281 on Build GRCh38
rs782209775 Chr.7: 73537253-73537253 on Build GRCh38
rs782736225 Chr.7: 73537325-73537325 on Build GRCh38
rs781802427 Chr.7: 73537292-73537293 on Build GRCh38
rs782603452 Chr.7: 73537311-73537311 on Build GRCh38
rs947058929 Chr.7: 73537237-73537237 on Build GRCh38
rs782185455 Chr.7: 73537209-73537209 on Build GRCh38
rs769255998 Chr.7: 73537108-73537108 on Build GRCh38
rs776990992 Chr.7: 73537110-73537110 on Build GRCh38
rs200214637 Chr.7: 73537436-73537436 on Build GRCh38
rs991392816 Chr.7: 73537186-73537186 on Build GRCh38
rs375576005 Chr.7: 73537276-73537276 on Build GRCh38
rs781968905 Chr.7: 73537414-73537414 on Build GRCh38
rs773661748 Chr.7: 73537335-73537335 on Build GRCh38
rs953528752 Chr.7: 73537150-73537150 on Build GRCh38
rs781938776 Chr.7: 73537336-73537336 on Build GRCh38
rs782153220 Chr.7: 73537407-73537407 on Build GRCh38
rs782246562 Chr.7: 73537387-73537387 on Build GRCh38
rs782819772 Chr.7: 73537348-73537348 on Build GRCh38
rs902908461 Chr.7: 73537087-73537087 on Build GRCh38
rs752592157 Chr.7: 73537048-73537048 on Build GRCh38
rs782314388 Chr.7: 73537402-73537402 on Build GRCh38
rs1016776522 Chr.7: 73537160-73537160 on Build GRCh38
rs782465879 Chr.7: 73537265-73537265 on Build GRCh38
rs782388247 Chr.7: 73537268-73537268 on Build GRCh38
rs201805044 Chr.7: 73537338-73537338 on Build GRCh38
rs756856634 Chr.7: 73537103-73537103 on Build GRCh38
rs782792047 Chr.7: 73537442-73537442 on Build GRCh38
rs781901018 Chr.7: 73537374-73537374 on Build GRCh38
rs191652486 Chr.7: 73537175-73537175 on Build GRCh38
rs552864297 Chr.7: 73537233-73537233 on Build GRCh38
rs370470463 Chr.7: 73537279-73537279 on Build GRCh38
rs781889639 Chr.7: 73537349-73537349 on Build GRCh38
rs536648176 Chr.7: 73537159-73537159 on Build GRCh38
rs1000115740 Chr.7: 73537091-73537091 on Build GRCh38
rs991258499 Chr.7: 73537453-73537453 on Build GRCh38
rs782728263 Chr.7: 73537425-73537425 on Build GRCh38
rs959888534 Chr.7: 73537435-73537435 on Build GRCh38
rs915811428 Chr.7: 73537232-73537232 on Build GRCh38
rs576551566 Chr.7: 73537320-73537320 on Build GRCh38
rs782201415 Chr.7: 73537395-73537395 on Build GRCh38
rs782560828 Chr.7: 73537370-73537370 on Build GRCh38
rs1035725737 Chr.7: 73537409-73537409 on Build GRCh38
rs782269872 Chr.7: 73537316-73537316 on Build GRCh38
rs146117610 Chr.7: 73537049-73537049 on Build GRCh38
rs968630940 Chr.7: 73537475-73537475 on Build GRCh38
rs534849362 Chr.7: 73537230-73537230 on Build GRCh38
rs781793182 Chr.7: 73537299-73537299 on Build GRCh38
rs113146366 Chr.7: 73537109-73537109 on Build GRCh38
rs61734974 Chr.7: 73537355-73537355 on Build GRCh38
rs782390609 Chr.7: 73537319-73537319 on Build GRCh38
rs781783012 Chr.7: 73537429-73537429 on Build GRCh38
rs782735560 Chr.7: 73537356-73537356 on Build GRCh38
rs782548045 Chr.7: 73537303-73537303 on Build GRCh38
rs962121678 Chr.7: 73537170-73537170 on Build GRCh38
rs574800772 Chr.7: 73537240-73537240 on Build GRCh38
rs782474413 Chr.7: 73537291-73537291 on Build GRCh38
rs370881412 Chr.7: 73537410-73537410 on Build GRCh38

View Full Product Details