Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74372613-74373118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIP2
Entrez Gene ID
7461
Gene Name
CAP-Gly domain containing linker protein 2
Gene Aliases
CLIP, CLIP-115, CYLN2, WBSCR3, WBSCR4, WSCR3, WSCR4
UniGene
Hs.647018
Gene Chromosome Location
Chr.7: 74289437-74405943 on Build GRCh38
Transcripts
NM_003388.4 NM_032421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1452029 c.1444G>T p.A482S Chr.7: 74372995-74372995 on Build GRCh38
6020509 c.1381-1G>C p.? Chr.7: 74372931-74372931 on Build GRCh38
5457348 c.1485+7G>A p.? Chr.7: 74373043-74373043 on Build GRCh38
5979364 c.1457G>A p.R486Q Chr.7: 74373008-74373008 on Build GRCh38
4421352 c.1457G>C p.R486P Chr.7: 74373008-74373008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941985357 Chr.7: 74372803-74372803 on Build GRCh38
rs536279606 Chr.7: 74372849-74372849 on Build GRCh38
rs958920470 Chr.7: 74372645-74372645 on Build GRCh38
rs782030543 Chr.7: 74373043-74373043 on Build GRCh38
rs782331938 Chr.7: 74373066-74373066 on Build GRCh38
rs199579765 Chr.7: 74372901-74372901 on Build GRCh38
rs1036370813 Chr.7: 74372808-74372808 on Build GRCh38
rs938734696 Chr.7: 74373061-74373061 on Build GRCh38
rs962122242 Chr.7: 74372634-74372634 on Build GRCh38
rs117357496 Chr.7: 74372783-74372783 on Build GRCh38
rs1002974159 Chr.7: 74372838-74372838 on Build GRCh38
rs782810749 Chr.7: 74373027-74373027 on Build GRCh38
rs782121130 Chr.7: 74372892-74372892 on Build GRCh38
rs781884928 Chr.7: 74373053-74373053 on Build GRCh38
rs923653747 Chr.7: 74372762-74372762 on Build GRCh38
rs897851938 Chr.7: 74373060-74373060 on Build GRCh38
rs887779768 Chr.7: 74372802-74372802 on Build GRCh38
rs549917264 Chr.7: 74372633-74372633 on Build GRCh38
rs782529593 Chr.7: 74372907-74372907 on Build GRCh38
rs376115830 Chr.7: 74372996-74372996 on Build GRCh38
rs201741678 Chr.7: 74373056-74373056 on Build GRCh38
rs897882877 Chr.7: 74372821-74372821 on Build GRCh38
rs782785004 Chr.7: 74372898-74372898 on Build GRCh38
rs554559654 Chr.7: 74372882-74372882 on Build GRCh38
rs1003005077 Chr.7: 74372643-74372643 on Build GRCh38
rs782790707 Chr.7: 74372899-74372900 on Build GRCh38
rs553856119 Chr.7: 74372698-74372698 on Build GRCh38
rs558749004 Chr.7: 74372952-74372952 on Build GRCh38
rs782223604 Chr.7: 74373003-74373003 on Build GRCh38
rs986065447 Chr.7: 74373008-74373008 on Build GRCh38
rs782180052 Chr.7: 74372954-74372954 on Build GRCh38
rs781828771 Chr.7: 74372899-74372899 on Build GRCh38
rs979202926 Chr.7: 74372755-74372755 on Build GRCh38
rs377649215 Chr.7: 74373065-74373065 on Build GRCh38
rs373141145 Chr.7: 74372957-74372957 on Build GRCh38
rs776118408 Chr.7: 74372680-74372680 on Build GRCh38
rs782614469 Chr.7: 74373059-74373059 on Build GRCh38
rs782635400 Chr.7: 74373054-74373054 on Build GRCh38
rs1013160843 Chr.7: 74372917-74372917 on Build GRCh38
rs923128174 Chr.7: 74372786-74372786 on Build GRCh38
rs370792041 Chr.7: 74373004-74373004 on Build GRCh38
rs534319062 Chr.7: 74373058-74373058 on Build GRCh38
rs543052949 Chr.7: 74373078-74373078 on Build GRCh38
rs956433572 Chr.7: 74372977-74372977 on Build GRCh38
rs781806818 Chr.7: 74372905-74372905 on Build GRCh38
rs931844388 Chr.7: 74373007-74373007 on Build GRCh38
rs782615936 Chr.7: 74372953-74372953 on Build GRCh38
rs781925728 Chr.7: 74373006-74373006 on Build GRCh38
rs781838222 Chr.7: 74373049-74373049 on Build GRCh38
rs369824735 Chr.7: 74372928-74372928 on Build GRCh38
rs782115030 Chr.7: 74373044-74373044 on Build GRCh38
rs1035841948 Chr.7: 74372842-74372842 on Build GRCh38
rs782469723 Chr.7: 74372934-74372934 on Build GRCh38
rs1050803745 Chr.7: 74372798-74372798 on Build GRCh38
rs968152035 Chr.7: 74372751-74372751 on Build GRCh38
rs782023870 Chr.7: 74373068-74373068 on Build GRCh38
rs782321095 Chr.7: 74372955-74372955 on Build GRCh38
rs969000132 Chr.7: 74372923-74372923 on Build GRCh38
rs782690565 Chr.7: 74372910-74372910 on Build GRCh38
rs531846694 Chr.7: 74373090-74373090 on Build GRCh38
rs935091004 Chr.7: 74372763-74372763 on Build GRCh38
rs137993993 Chr.7: 74372997-74372997 on Build GRCh38
rs1030638385 Chr.7: 74372968-74372968 on Build GRCh38
rs373887488 Chr.7: 74373088-74373088 on Build GRCh38
rs537350900 Chr.7: 74372902-74372902 on Build GRCh38
rs1022333281 Chr.7: 74372922-74372922 on Build GRCh38
rs782489591 Chr.7: 74373057-74373057 on Build GRCh38
rs782282342 Chr.7: 74372982-74372982 on Build GRCh38
rs560667523 Chr.7: 74373050-74373050 on Build GRCh38
rs931883885 Chr.7: 74372793-74372793 on Build GRCh38
rs545494836 Chr.7: 74373045-74373045 on Build GRCh38
rs782070393 Chr.7: 74373017-74373017 on Build GRCh38
rs6975173 Chr.7: 74372644-74372644 on Build GRCh38

View Full Product Details