Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99456741-99457244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Gene Symbol
CPSF4
Entrez Gene ID
10898
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
CPSF30, NAR, NEB-1, NEB1
UniGene
Hs.489287
Gene Chromosome Location
Chr.7: 99438920-99457377 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

SNP Detail

SNP ID Chromosome Location
rs1046671088 Chr.7: 99457054-99457054 on Build GRCh38
rs373413008 Chr.7: 99457071-99457071 on Build GRCh38
rs577957963 Chr.7: 99456805-99456805 on Build GRCh38
rs189479313 Chr.7: 99456908-99456908 on Build GRCh38
rs905440989 Chr.7: 99457047-99457047 on Build GRCh38
rs953248644 Chr.7: 99456981-99456981 on Build GRCh38
rs115205187 Chr.7: 99456782-99456782 on Build GRCh38
rs984587720 Chr.7: 99456986-99456986 on Build GRCh38
rs78391420 Chr.7: 99457065-99457065 on Build GRCh38
rs1045733739 Chr.7: 99457043-99457043 on Build GRCh38
rs147044374 Chr.7: 99456869-99456869 on Build GRCh38
rs115904935 Chr.7: 99456848-99456848 on Build GRCh38
rs779938093 Chr.7: 99456993-99456993 on Build GRCh38
rs1030186649 Chr.7: 99457185-99457185 on Build GRCh38
rs770389307 Chr.7: 99456858-99456858 on Build GRCh38
rs756699053 Chr.7: 99456957-99456957 on Build GRCh38
rs1020382063 Chr.7: 99456948-99456948 on Build GRCh38
rs925366399 Chr.7: 99457009-99457009 on Build GRCh38
rs895398610 Chr.7: 99456817-99456817 on Build GRCh38
rs1006344675 Chr.7: 99457123-99457123 on Build GRCh38
rs192853553 Chr.7: 99456988-99456988 on Build GRCh38
rs548881756 Chr.7: 99457107-99457107 on Build GRCh38
rs114036924 Chr.7: 99456760-99456760 on Build GRCh38
rs1045298882 Chr.7: 99456820-99456820 on Build GRCh38
rs561857517 Chr.7: 99456877-99456877 on Build GRCh38
rs374958556 Chr.7: 99456999-99456999 on Build GRCh38
rs976406207 Chr.7: 99456958-99456958 on Build GRCh38
rs1032099591 Chr.7: 99457212-99457212 on Build GRCh38
rs553998110 Chr.7: 99456831-99456831 on Build GRCh38
rs556134098 Chr.7: 99456791-99456791 on Build GRCh38
rs887543046 Chr.7: 99457117-99457117 on Build GRCh38
rs371951331 Chr.7: 99457216-99457216 on Build GRCh38
rs45459197 Chr.7: 99457040-99457040 on Build GRCh38
rs544274031 Chr.7: 99456924-99456924 on Build GRCh38
rs892607621 Chr.7: 99457138-99457138 on Build GRCh38
rs917929152 Chr.7: 99457034-99457034 on Build GRCh38
rs371684801 Chr.7: 99456849-99456849 on Build GRCh38
rs138281757 Chr.7: 99457165-99457165 on Build GRCh38
rs368117982 Chr.7: 99456822-99456822 on Build GRCh38
rs755152036 Chr.7: 99457055-99457055 on Build GRCh38
rs8794 Chr.7: 99457006-99457006 on Build GRCh38
rs550946933 Chr.7: 99457137-99457137 on Build GRCh38
rs930127193 Chr.7: 99457068-99457068 on Build GRCh38
rs972146257 Chr.7: 99457033-99457033 on Build GRCh38
rs958149474 Chr.7: 99457217-99457217 on Build GRCh38
rs901512387 Chr.7: 99457178-99457178 on Build GRCh38
rs949682146 Chr.7: 99456810-99456810 on Build GRCh38
rs1009934077 Chr.7: 99456917-99456917 on Build GRCh38
rs1028874925 Chr.7: 99456973-99456973 on Build GRCh38
rs1017655662 Chr.7: 99456870-99456870 on Build GRCh38
rs1053944765 Chr.7: 99457112-99457112 on Build GRCh38
rs940767943 Chr.7: 99457019-99457019 on Build GRCh38
rs567617087 Chr.7: 99457160-99457160 on Build GRCh38
rs1019991656 Chr.7: 99457177-99457177 on Build GRCh38
rs551718593 Chr.7: 99457001-99457001 on Build GRCh38
rs977885870 Chr.7: 99457193-99457193 on Build GRCh38
rs966553322 Chr.7: 99457192-99457192 on Build GRCh38

View Full Product Details