Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99457137-99457644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Gene Symbol
CPSF4
Entrez Gene ID
10898
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
CPSF30, NAR, NEB-1, NEB1
UniGene
Hs.489287
Gene Chromosome Location
Chr.7: 99438920-99457377 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

SNP Detail

SNP ID Chromosome Location
rs997185152 Chr.7: 99457243-99457243 on Build GRCh38
rs184226989 Chr.7: 99457234-99457234 on Build GRCh38
rs373201795 Chr.7: 99457383-99457383 on Build GRCh38
rs961829821 Chr.7: 99457318-99457318 on Build GRCh38
rs779128884 Chr.7: 99457270-99457270 on Build GRCh38
rs1016064803 Chr.7: 99457289-99457289 on Build GRCh38
rs982401501 Chr.7: 99457331-99457331 on Build GRCh38
rs971119830 Chr.7: 99457308-99457308 on Build GRCh38
rs1024138279 Chr.7: 99457619-99457619 on Build GRCh38
rs1039179590 Chr.7: 99457471-99457471 on Build GRCh38
rs1030186649 Chr.7: 99457185-99457185 on Build GRCh38
rs1050040268 Chr.7: 99457506-99457506 on Build GRCh38
rs910017284 Chr.7: 99457450-99457450 on Build GRCh38
rs768880498 Chr.7: 99457570-99457570 on Build GRCh38
rs879224739 Chr.7: 99457264-99457264 on Build GRCh38
rs1804948 Chr.7: 99457298-99457298 on Build GRCh38
rs113662347 Chr.7: 99457363-99457363 on Build GRCh38
rs941803043 Chr.7: 99457466-99457466 on Build GRCh38
rs958099799 Chr.7: 99457604-99457604 on Build GRCh38
rs900188841 Chr.7: 99457480-99457480 on Build GRCh38
rs770708973 Chr.7: 99457507-99457507 on Build GRCh38
rs997133618 Chr.7: 99457481-99457481 on Build GRCh38
rs538620153 Chr.7: 99457329-99457329 on Build GRCh38
rs901592275 Chr.7: 99457463-99457463 on Build GRCh38
rs945530249 Chr.7: 99457424-99457424 on Build GRCh38
rs536880609 Chr.7: 99457611-99457611 on Build GRCh38
rs763414598 Chr.7: 99457488-99457488 on Build GRCh38
rs1032099591 Chr.7: 99457212-99457212 on Build GRCh38
rs371951331 Chr.7: 99457216-99457216 on Build GRCh38
rs748417834 Chr.7: 99457453-99457453 on Build GRCh38
rs138281757 Chr.7: 99457165-99457165 on Build GRCh38
rs368558648 Chr.7: 99457413-99457414 on Build GRCh38
rs980777114 Chr.7: 99457361-99457361 on Build GRCh38
rs972561663 Chr.7: 99457327-99457327 on Build GRCh38
rs369462309 Chr.7: 99457236-99457236 on Build GRCh38
rs535885046 Chr.7: 99457434-99457434 on Build GRCh38
rs571311381 Chr.7: 99457279-99457279 on Build GRCh38
rs1054014919 Chr.7: 99457411-99457411 on Build GRCh38
rs142325695 Chr.7: 99457514-99457514 on Build GRCh38
rs958149474 Chr.7: 99457217-99457217 on Build GRCh38
rs936920300 Chr.7: 99457406-99457406 on Build GRCh38
rs997616635 Chr.7: 99457491-99457491 on Build GRCh38
rs901512387 Chr.7: 99457178-99457178 on Build GRCh38
rs552537806 Chr.7: 99457614-99457614 on Build GRCh38
rs971173960 Chr.7: 99457354-99457354 on Build GRCh38
rs902444227 Chr.7: 99457498-99457498 on Build GRCh38
rs917975519 Chr.7: 99457338-99457338 on Build GRCh38
rs567617087 Chr.7: 99457160-99457160 on Build GRCh38
rs1019991656 Chr.7: 99457177-99457177 on Build GRCh38
rs545774820 Chr.7: 99457430-99457430 on Build GRCh38
rs977885870 Chr.7: 99457193-99457193 on Build GRCh38
rs966553322 Chr.7: 99457192-99457192 on Build GRCh38
rs888756439 Chr.7: 99457512-99457512 on Build GRCh38
rs914072912 Chr.7: 99457423-99457423 on Build GRCh38

View Full Product Details