Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99827899-99828324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A43
Entrez Gene ID
64816
Gene Name
cytochrome P450 family 3 subfamily A member 43
Gene Aliases
-
UniGene
Hs.306220
Gene Chromosome Location
Chr.7: 99828013-99867138 on Build GRCh38
Transcripts
NM_001278921.1 NM_022820.4 NM_057095.2 NM_057096.3 NR_103868.1 NR_103869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4467635 c.14C>T p.P5L Chr.7: 99828129-99828129 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371606320 Chr.7: 99828117-99828117 on Build GRCh38
rs1000729523 Chr.7: 99828063-99828063 on Build GRCh38
rs772466109 Chr.7: 99828124-99828124 on Build GRCh38
rs891805211 Chr.7: 99828012-99828012 on Build GRCh38
rs959209416 Chr.7: 99828097-99828097 on Build GRCh38
rs763610341 Chr.7: 99828148-99828148 on Build GRCh38
rs559197324 Chr.7: 99828064-99828064 on Build GRCh38
rs944176090 Chr.7: 99828268-99828268 on Build GRCh38
rs914787766 Chr.7: 99828249-99828249 on Build GRCh38
rs202031279 Chr.7: 99828082-99828082 on Build GRCh38
rs751352157 Chr.7: 99828182-99828182 on Build GRCh38
rs749331044 Chr.7: 99828110-99828110 on Build GRCh38
rs577285838 Chr.7: 99828034-99828034 on Build GRCh38
rs777879810 Chr.7: 99828101-99828101 on Build GRCh38
rs553497612 Chr.7: 99828163-99828163 on Build GRCh38
rs374620083 Chr.7: 99828128-99828128 on Build GRCh38
rs779189527 Chr.7: 99828211-99828211 on Build GRCh38
rs1038659728 Chr.7: 99827991-99827991 on Build GRCh38
rs562959556 Chr.7: 99828157-99828157 on Build GRCh38
rs145433087 Chr.7: 99828149-99828149 on Build GRCh38
rs377124793 Chr.7: 99828183-99828183 on Build GRCh38
rs540844577 Chr.7: 99828039-99828039 on Build GRCh38
rs772815209 Chr.7: 99828078-99828078 on Build GRCh38
rs146431983 Chr.7: 99828158-99828158 on Build GRCh38
rs774930852 Chr.7: 99828113-99828113 on Build GRCh38
rs748390766 Chr.7: 99828088-99828088 on Build GRCh38
rs201024192 Chr.7: 99828145-99828145 on Build GRCh38
rs958829233 Chr.7: 99828239-99828239 on Build GRCh38
rs758125742 Chr.7: 99828076-99828076 on Build GRCh38
rs775764854 Chr.7: 99828127-99828127 on Build GRCh38
rs758326938 Chr.7: 99828165-99828165 on Build GRCh38
rs193296612 Chr.7: 99828248-99828248 on Build GRCh38
rs766174900 Chr.7: 99828168-99828168 on Build GRCh38
rs931035474 Chr.7: 99828235-99828235 on Build GRCh38
rs200290120 Chr.7: 99828229-99828229 on Build GRCh38
rs371298927 Chr.7: 99828216-99828216 on Build GRCh38
rs45600736 Chr.7: 99827992-99827992 on Build GRCh38
rs1050877477 Chr.7: 99827954-99827954 on Build GRCh38
rs564652728 Chr.7: 99828295-99828295 on Build GRCh38
rs558962186 Chr.7: 99828018-99828018 on Build GRCh38
rs779781051 Chr.7: 99828081-99828081 on Build GRCh38
rs1006680112 Chr.7: 99827972-99827972 on Build GRCh38
rs971121236 Chr.7: 99828125-99828125 on Build GRCh38
rs889654458 Chr.7: 99827955-99827955 on Build GRCh38
rs375286435 Chr.7: 99827983-99827983 on Build GRCh38
rs771148123 Chr.7: 99828111-99828111 on Build GRCh38
rs919714560 Chr.7: 99828197-99828197 on Build GRCh38
rs188624641 Chr.7: 99828152-99828152 on Build GRCh38
rs45577236 Chr.7: 99828035-99828035 on Build GRCh38
rs749525707 Chr.7: 99828203-99828203 on Build GRCh38
rs777797970 Chr.7: 99828192-99828192 on Build GRCh38
rs764735572 Chr.7: 99828156-99828156 on Build GRCh38

View Full Product Details