Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99848085-99848591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A43
Entrez Gene ID
64816
Gene Name
cytochrome P450 family 3 subfamily A member 43
Gene Aliases
-
UniGene
Hs.306220
Gene Chromosome Location
Chr.7: 99828013-99867138 on Build GRCh38
Transcripts
NM_001278921.1 NM_022820.4 NM_057095.2 NM_057096.3 NR_103868.1 NR_103869.1 XM_017012542.1 XM_017012543.1 XM_017012544.1 XM_017012545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2867549 c.447T>G p.I149M Chr.7: 99848180-99848180 on Build GRCh38
4518057 c.93_94CC>TT p.H32Y Chr.7: 99848239-99848240 on Build GRCh38
6111232 c.56G>T p.G19V Chr.7: 99848202-99848202 on Build GRCh38
6111231 c.469G>T p.V157L Chr.7: 99848202-99848202 on Build GRCh38
7425646 c.83A>T p.E28V Chr.7: 99848229-99848229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054785720 Chr.7: 99848520-99848520 on Build GRCh38
rs923223498 Chr.7: 99848476-99848476 on Build GRCh38
rs371251791 Chr.7: 99848157-99848157 on Build GRCh38
rs763656555 Chr.7: 99848131-99848131 on Build GRCh38
rs897408250 Chr.7: 99848570-99848570 on Build GRCh38
rs183826531 Chr.7: 99848316-99848316 on Build GRCh38
rs776160563 Chr.7: 99848253-99848253 on Build GRCh38
rs1003373717 Chr.7: 99848511-99848511 on Build GRCh38
rs907326307 Chr.7: 99848500-99848500 on Build GRCh38
rs1026947253 Chr.7: 99848335-99848335 on Build GRCh38
rs542436399 Chr.7: 99848544-99848544 on Build GRCh38
rs771523961 Chr.7: 99848186-99848186 on Build GRCh38
rs568218999 Chr.7: 99848126-99848126 on Build GRCh38
rs750387816 Chr.7: 99848263-99848263 on Build GRCh38
rs757422577 Chr.7: 99848290-99848290 on Build GRCh38
rs772485834 Chr.7: 99848201-99848201 on Build GRCh38
rs574623821 Chr.7: 99848454-99848454 on Build GRCh38
rs930008369 Chr.7: 99848447-99848447 on Build GRCh38
rs898484554 Chr.7: 99848436-99848436 on Build GRCh38
rs569931868 Chr.7: 99848275-99848275 on Build GRCh38
rs778059301 Chr.7: 99848172-99848172 on Build GRCh38
rs764988602 Chr.7: 99848149-99848149 on Build GRCh38
rs187120994 Chr.7: 99848323-99848323 on Build GRCh38
rs1047128830 Chr.7: 99848487-99848487 on Build GRCh38
rs911387649 Chr.7: 99848366-99848366 on Build GRCh38
rs754356184 Chr.7: 99848264-99848264 on Build GRCh38
rs767027924 Chr.7: 99848122-99848122 on Build GRCh38
rs534604656 Chr.7: 99848332-99848332 on Build GRCh38
rs371581147 Chr.7: 99848197-99848197 on Build GRCh38
rs942936249 Chr.7: 99848379-99848379 on Build GRCh38
rs45515501 Chr.7: 99848555-99848555 on Build GRCh38
rs778169761 Chr.7: 99848286-99848286 on Build GRCh38
rs572896798 Chr.7: 99848552-99848552 on Build GRCh38
rs28669087 Chr.7: 99848351-99848351 on Build GRCh38
rs750148915 Chr.7: 99848150-99848150 on Build GRCh38
rs375014878 Chr.7: 99848266-99848266 on Build GRCh38
rs924944832 Chr.7: 99848495-99848495 on Build GRCh38
rs943314406 Chr.7: 99848127-99848127 on Build GRCh38
rs759975587 Chr.7: 99848120-99848120 on Build GRCh38
rs376469708 Chr.7: 99848385-99848385 on Build GRCh38
rs1012145039 Chr.7: 99848567-99848567 on Build GRCh38
rs753916770 Chr.7: 99848289-99848289 on Build GRCh38
rs753759775 Chr.7: 99848134-99848134 on Build GRCh38
rs751683182 Chr.7: 99848276-99848276 on Build GRCh38
rs749780001 Chr.7: 99848183-99848183 on Build GRCh38
rs936465296 Chr.7: 99848496-99848496 on Build GRCh38
rs779217617 Chr.7: 99848295-99848295 on Build GRCh38
rs977737222 Chr.7: 99848492-99848492 on Build GRCh38
rs45450092 Chr.7: 99848168-99848168 on Build GRCh38
rs1038608227 Chr.7: 99848421-99848421 on Build GRCh38
rs985534064 Chr.7: 99848369-99848369 on Build GRCh38
rs895333139 Chr.7: 99848524-99848524 on Build GRCh38
rs949593979 Chr.7: 99848546-99848546 on Build GRCh38
rs554636533 Chr.7: 99848138-99848138 on Build GRCh38
rs140500492 Chr.7: 99848301-99848301 on Build GRCh38
rs764042128 Chr.7: 99848153-99848155 on Build GRCh38
rs765036212 Chr.7: 99848262-99848262 on Build GRCh38
rs376128279 Chr.7: 99848160-99848160 on Build GRCh38
rs200952381 Chr.7: 99848155-99848155 on Build GRCh38
rs749959233 Chr.7: 99848114-99848114 on Build GRCh38
rs760377549 Chr.7: 99848245-99848245 on Build GRCh38
rs774997883 Chr.7: 99848210-99848210 on Build GRCh38
rs760243610 Chr.7: 99848135-99848135 on Build GRCh38

View Full Product Details