Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100158851-100159381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST4
Entrez Gene ID
79690
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
GAL3ST-4
UniGene
Hs.44856
Gene Chromosome Location
Chr.7: 100159242-100168750 on Build GRCh38
Transcripts
NM_024637.4

SNP Detail

SNP ID Chromosome Location
rs948179851 Chr.7: 100159075-100159075 on Build GRCh38
rs963184247 Chr.7: 100159255-100159258 on Build GRCh38
rs139461829 Chr.7: 100159154-100159155 on Build GRCh38
rs574479409 Chr.7: 100159229-100159229 on Build GRCh38
rs1045176878 Chr.7: 100159099-100159099 on Build GRCh38
rs960123278 Chr.7: 100159303-100159303 on Build GRCh38
rs923627973 Chr.7: 100159011-100159011 on Build GRCh38
rs943671161 Chr.7: 100159171-100159171 on Build GRCh38
rs543046815 Chr.7: 100159169-100159170 on Build GRCh38
rs541525855 Chr.7: 100159241-100159241 on Build GRCh38
rs1004359507 Chr.7: 100159250-100159250 on Build GRCh38
rs866248515 Chr.7: 100159168-100159168 on Build GRCh38
rs868603319 Chr.7: 100158930-100158930 on Build GRCh38
rs190922165 Chr.7: 100159192-100159192 on Build GRCh38
rs113955884 Chr.7: 100159157-100159157 on Build GRCh38
rs1003625851 Chr.7: 100159157-100159160 on Build GRCh38
rs1052207042 Chr.7: 100159107-100159107 on Build GRCh38
rs916450311 Chr.7: 100158970-100158970 on Build GRCh38
rs900042299 Chr.7: 100159208-100159208 on Build GRCh38
rs746640302 Chr.7: 100158989-100158989 on Build GRCh38
rs1028773251 Chr.7: 100159281-100159284 on Build GRCh38
rs923630329 Chr.7: 100159355-100159355 on Build GRCh38
rs534653814 Chr.7: 100159185-100159185 on Build GRCh38
rs1035639398 Chr.7: 100158881-100158881 on Build GRCh38
rs35262758 Chr.7: 100159157-100159158 on Build GRCh38
rs951460189 Chr.7: 100158878-100158878 on Build GRCh38
rs995809404 Chr.7: 100159213-100159213 on Build GRCh38
rs141967364 Chr.7: 100158987-100158987 on Build GRCh38
rs987219366 Chr.7: 100159316-100159316 on Build GRCh38
rs868676602 Chr.7: 100159181-100159181 on Build GRCh38
rs969701431 Chr.7: 100159334-100159334 on Build GRCh38
rs1035707649 Chr.7: 100159296-100159296 on Build GRCh38
rs893992729 Chr.7: 100159044-100159044 on Build GRCh38
rs890895442 Chr.7: 100159166-100159166 on Build GRCh38
rs906576820 Chr.7: 100159120-100159120 on Build GRCh38
rs563892584 Chr.7: 100159248-100159248 on Build GRCh38
rs925088658 Chr.7: 100159352-100159352 on Build GRCh38
rs1027099488 Chr.7: 100159226-100159226 on Build GRCh38
rs1054026337 Chr.7: 100159025-100159025 on Build GRCh38
rs771446330 Chr.7: 100159325-100159325 on Build GRCh38
rs7807641 Chr.7: 100158880-100158880 on Build GRCh38
rs979401927 Chr.7: 100159345-100159345 on Build GRCh38
rs991860857 Chr.7: 100159307-100159307 on Build GRCh38
rs559059843 Chr.7: 100159169-100159169 on Build GRCh38
rs554492042 Chr.7: 100158942-100158942 on Build GRCh38
rs935054265 Chr.7: 100159065-100159065 on Build GRCh38
rs201953361 Chr.7: 100158880-100158880 on Build GRCh38
rs933006539 Chr.7: 100158983-100158983 on Build GRCh38
rs960009709 Chr.7: 100158923-100158923 on Build GRCh38
rs370875894 Chr.7: 100159357-100159357 on Build GRCh38
rs569480930 Chr.7: 100158931-100158931 on Build GRCh38
rs948000013 Chr.7: 100158975-100158975 on Build GRCh38
rs1023006697 Chr.7: 100159328-100159328 on Build GRCh38
rs397814676 Chr.7: 100159171-100159172 on Build GRCh38
rs974512482 Chr.7: 100159262-100159262 on Build GRCh38
rs185955336 Chr.7: 100159022-100159022 on Build GRCh38
rs866770505 Chr.7: 100159172-100159172 on Build GRCh38
rs925140790 Chr.7: 100159057-100159057 on Build GRCh38

View Full Product Details