Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100159392-100159892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST4
Entrez Gene ID
79690
Gene Name
galactose-3-O-sulfotransferase 4
Gene Aliases
GAL3ST-4
UniGene
Hs.44856
Gene Chromosome Location
Chr.7: 100159242-100168750 on Build GRCh38
Transcripts
NM_024637.4

SNP Detail

SNP ID Chromosome Location
rs987639699 Chr.7: 100159804-100159804 on Build GRCh38
rs370049334 Chr.7: 100159579-100159579 on Build GRCh38
rs1048605956 Chr.7: 100159461-100159461 on Build GRCh38
rs533734994 Chr.7: 100159470-100159470 on Build GRCh38
rs931541516 Chr.7: 100159460-100159460 on Build GRCh38
rs948210894 Chr.7: 100159489-100159489 on Build GRCh38
rs780228789 Chr.7: 100159650-100159650 on Build GRCh38
rs770141391 Chr.7: 100159563-100159563 on Build GRCh38
rs912437528 Chr.7: 100159684-100159684 on Build GRCh38
rs1032230373 Chr.7: 100159646-100159646 on Build GRCh38
rs996044574 Chr.7: 100159752-100159752 on Build GRCh38
rs540623522 Chr.7: 100159738-100159738 on Build GRCh38
rs546180234 Chr.7: 100159416-100159416 on Build GRCh38
rs899119854 Chr.7: 100159722-100159722 on Build GRCh38
rs915457534 Chr.7: 100159447-100159447 on Build GRCh38
rs529407027 Chr.7: 100159772-100159772 on Build GRCh38
rs78325297 Chr.7: 100159719-100159719 on Build GRCh38
rs550366866 Chr.7: 100159720-100159721 on Build GRCh38
rs550771073 Chr.7: 100159849-100159849 on Build GRCh38
rs59528184 Chr.7: 100159577-100159577 on Build GRCh38
rs887375515 Chr.7: 100159505-100159505 on Build GRCh38
rs965490293 Chr.7: 100159754-100159754 on Build GRCh38
rs1030727967 Chr.7: 100159825-100159825 on Build GRCh38
rs530654612 Chr.7: 100159863-100159863 on Build GRCh38
rs74883328 Chr.7: 100159524-100159524 on Build GRCh38
rs968771223 Chr.7: 100159593-100159593 on Build GRCh38
rs528774292 Chr.7: 100159709-100159709 on Build GRCh38
rs866169211 Chr.7: 100159675-100159675 on Build GRCh38
rs564807878 Chr.7: 100159602-100159602 on Build GRCh38
rs954554258 Chr.7: 100159785-100159785 on Build GRCh38
rs568990471 Chr.7: 100159859-100159859 on Build GRCh38
rs1048057699 Chr.7: 100159862-100159862 on Build GRCh38
rs777091098 Chr.7: 100159813-100159813 on Build GRCh38
rs1013037161 Chr.7: 100159576-100159576 on Build GRCh38
rs1028178117 Chr.7: 100159764-100159764 on Build GRCh38
rs1047131623 Chr.7: 100159565-100159565 on Build GRCh38
rs751699797 Chr.7: 100159852-100159853 on Build GRCh38
rs374600359 Chr.7: 100159719-100159720 on Build GRCh38
rs975215473 Chr.7: 100159763-100159763 on Build GRCh38
rs763007922 Chr.7: 100159618-100159618 on Build GRCh38
rs762664890 Chr.7: 100159712-100159712 on Build GRCh38
rs956614388 Chr.7: 100159651-100159651 on Build GRCh38
rs1004304078 Chr.7: 100159509-100159509 on Build GRCh38
rs561890684 Chr.7: 100159768-100159768 on Build GRCh38
rs1000622990 Chr.7: 100159645-100159645 on Build GRCh38

View Full Product Details