Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100199249-100199745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATS
Entrez Gene ID
352954
Gene Name
GATS, stromal antigen 3 opposite strand
Gene Aliases
STAG3OS
UniGene
Hs.556063
Gene Chromosome Location
Chr.7: 100187988-100272274 on Build GRCh38
Transcripts
XM_017012156.1 NM_001282716.1 NM_001282717.1 NM_001282718.1 NM_012447.3 XM_011515742.1 XM_017011683.1 XM_017011684.1 XM_017011685.1 XM_017011686.1 XM_017011687.1

Gene Symbol
STAG3
Entrez Gene ID
10734
Gene Name
stromal antigen 3
Gene Aliases
-
UniGene
Hs.592283
Gene Chromosome Location
Chr.7: 100177563-100219334 on Build GRCh38
Transcripts
XM_017012156.1 NM_001282716.1 NM_001282717.1 NM_001282718.1 NM_012447.3 XM_011515742.1 XM_017011683.1 XM_017011684.1 XM_017011685.1 XM_017011686.1 XM_017011687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5617439 c.1573+9C>G p.? Chr.7: 100199376-100199376 on Build GRCh38
4804222 c.1659C>A p.G553G Chr.7: 100199626-100199626 on Build GRCh38
5710775 c.1562A>G p.E521G Chr.7: 100199356-100199356 on Build GRCh38
603118 c.1523G>A p.R508Q Chr.7: 100199317-100199317 on Build GRCh38
1673367 c.1556T>G p.L519R Chr.7: 100199350-100199350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370347717 Chr.7: 100199389-100199389 on Build GRCh38
rs752618983 Chr.7: 100199340-100199340 on Build GRCh38
rs762664360 Chr.7: 100199316-100199316 on Build GRCh38
rs762089807 Chr.7: 100199278-100199278 on Build GRCh38
rs765988932 Chr.7: 100199317-100199317 on Build GRCh38
rs2272345 Chr.7: 100199408-100199408 on Build GRCh38
rs774562681 Chr.7: 100199566-100199566 on Build GRCh38
rs932766305 Chr.7: 100199696-100199696 on Build GRCh38
rs866637489 Chr.7: 100199452-100199452 on Build GRCh38
rs748198889 Chr.7: 100199537-100199537 on Build GRCh38
rs758468666 Chr.7: 100199391-100199392 on Build GRCh38
rs545360000 Chr.7: 100199309-100199309 on Build GRCh38
rs750181986 Chr.7: 100199656-100199656 on Build GRCh38
rs758978551 Chr.7: 100199403-100199403 on Build GRCh38
rs781256757 Chr.7: 100199608-100199608 on Build GRCh38
rs1010757663 Chr.7: 100199478-100199478 on Build GRCh38
rs751084132 Chr.7: 100199320-100199320 on Build GRCh38
rs748714185 Chr.7: 100199381-100199381 on Build GRCh38
rs373807249 Chr.7: 100199491-100199491 on Build GRCh38
rs767357096 Chr.7: 100199337-100199337 on Build GRCh38
rs756105314 Chr.7: 100199341-100199341 on Build GRCh38
rs760213165 Chr.7: 100199617-100199617 on Build GRCh38
rs879341671 Chr.7: 100199459-100199459 on Build GRCh38
rs545346538 Chr.7: 100199699-100199699 on Build GRCh38
rs143920664 Chr.7: 100199707-100199707 on Build GRCh38
rs1051623243 Chr.7: 100199700-100199700 on Build GRCh38
rs921471150 Chr.7: 100199684-100199684 on Build GRCh38
rs769672494 Chr.7: 100199697-100199697 on Build GRCh38
rs555000414 Chr.7: 100199474-100199474 on Build GRCh38
rs771590735 Chr.7: 100199613-100199613 on Build GRCh38
rs766909978 Chr.7: 100199371-100199371 on Build GRCh38
rs573945610 Chr.7: 100199504-100199504 on Build GRCh38
rs747089940 Chr.7: 100199361-100199361 on Build GRCh38
rs563074605 Chr.7: 100199623-100199623 on Build GRCh38
rs760732550 Chr.7: 100199407-100199407 on Build GRCh38
rs114537080 Chr.7: 100199405-100199405 on Build GRCh38
rs763285803 Chr.7: 100199391-100199391 on Build GRCh38
rs773983532 Chr.7: 100199399-100199399 on Build GRCh38
rs200869015 Chr.7: 100199291-100199291 on Build GRCh38
rs980933139 Chr.7: 100199564-100199564 on Build GRCh38
rs773610101 Chr.7: 100199390-100199390 on Build GRCh38
rs775113819 Chr.7: 100199616-100199616 on Build GRCh38
rs999278100 Chr.7: 100199720-100199720 on Build GRCh38
rs143187428 Chr.7: 100199273-100199273 on Build GRCh38
rs765138899 Chr.7: 100199652-100199652 on Build GRCh38
rs770338935 Chr.7: 100199551-100199551 on Build GRCh38
rs770596809 Chr.7: 100199393-100199393 on Build GRCh38
rs781401928 Chr.7: 100199534-100199534 on Build GRCh38
rs759043323 Chr.7: 100199331-100199331 on Build GRCh38
rs778208860 Chr.7: 100199552-100199552 on Build GRCh38
rs775751338 Chr.7: 100199271-100199271 on Build GRCh38
rs961408883 Chr.7: 100199628-100199628 on Build GRCh38
rs757077887 Chr.7: 100199411-100199411 on Build GRCh38
rs141330490 Chr.7: 100199406-100199406 on Build GRCh38
rs1024887840 Chr.7: 100199679-100199679 on Build GRCh38
rs539090342 Chr.7: 100199305-100199305 on Build GRCh38
rs376380968 Chr.7: 100199654-100199654 on Build GRCh38
rs544394583 Chr.7: 100199594-100199594 on Build GRCh38
rs781564543 Chr.7: 100199380-100199380 on Build GRCh38
rs956597291 Chr.7: 100199473-100199473 on Build GRCh38
rs760850659 Chr.7: 100199468-100199468 on Build GRCh38
rs375088944 Chr.7: 100199360-100199360 on Build GRCh38
rs578178474 Chr.7: 100199627-100199627 on Build GRCh38
rs368796564 Chr.7: 100199556-100199556 on Build GRCh38
rs376787666 Chr.7: 100199372-100199372 on Build GRCh38
rs753652685 Chr.7: 100199351-100199351 on Build GRCh38
rs751748479 Chr.7: 100199502-100199502 on Build GRCh38
rs758742892 Chr.7: 100199352-100199352 on Build GRCh38
rs891122661 Chr.7: 100199433-100199433 on Build GRCh38
rs745844628 Chr.7: 100199683-100199684 on Build GRCh38
rs773555060 Chr.7: 100199294-100199294 on Build GRCh38
rs941623435 Chr.7: 100199650-100199650 on Build GRCh38
rs761714259 Chr.7: 100199641-100199641 on Build GRCh38
rs540870980 Chr.7: 100199430-100199430 on Build GRCh38
rs372391791 Chr.7: 100199560-100199560 on Build GRCh38
rs755270421 Chr.7: 100199657-100199657 on Build GRCh38
rs767614675 Chr.7: 100199676-100199676 on Build GRCh38

View Full Product Details