Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100550331-100550827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
UniGene
Hs.521083
Gene Chromosome Location
Chr.7: 100539211-100568220 on Build GRCh38
Transcripts
NM_006076.4 XM_005250306.2 XM_017012112.1 XM_017012113.1 XM_017012114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
172591 c.327G>T p.K109N Chr.7: 100550407-100550407 on Build GRCh38
1082864 c.414A>T p.K138N Chr.7: 100550494-100550494 on Build GRCh38
5946454 c.347A>T p.D116V Chr.7: 100550427-100550427 on Build GRCh38
4517573 c.374_375CC>TT p.S125F Chr.7: 100550454-100550455 on Build GRCh38
6596231 c.377G>A p.R126K Chr.7: 100550457-100550457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370191289 Chr.7: 100550517-100550517 on Build GRCh38
rs766494809 Chr.7: 100550758-100550758 on Build GRCh38
rs753230251 Chr.7: 100550562-100550562 on Build GRCh38
rs985967230 Chr.7: 100550649-100550649 on Build GRCh38
rs900272221 Chr.7: 100550741-100550741 on Build GRCh38
rs766563298 Chr.7: 100550548-100550548 on Build GRCh38
rs748973775 Chr.7: 100550494-100550494 on Build GRCh38
rs758596353 Chr.7: 100550436-100550436 on Build GRCh38
rs375257050 Chr.7: 100550483-100550483 on Build GRCh38
rs948775820 Chr.7: 100550447-100550447 on Build GRCh38
rs1048007207 Chr.7: 100550613-100550617 on Build GRCh38
rs893877093 Chr.7: 100550369-100550369 on Build GRCh38
rs929568267 Chr.7: 100550559-100550559 on Build GRCh38
rs963176398 Chr.7: 100550743-100550743 on Build GRCh38
rs372318437 Chr.7: 100550537-100550537 on Build GRCh38
rs777713041 Chr.7: 100550383-100550384 on Build GRCh38
rs769490472 Chr.7: 100550368-100550368 on Build GRCh38
rs773875205 Chr.7: 100550393-100550393 on Build GRCh38
rs760595528 Chr.7: 100550533-100550533 on Build GRCh38
rs748450379 Chr.7: 100550367-100550367 on Build GRCh38
rs910315268 Chr.7: 100550677-100550677 on Build GRCh38
rs751644727 Chr.7: 100550552-100550552 on Build GRCh38
rs35579128 Chr.7: 100550685-100550685 on Build GRCh38
rs755078076 Chr.7: 100550554-100550554 on Build GRCh38
rs765595747 Chr.7: 100550432-100550432 on Build GRCh38
rs773749861 Chr.7: 100550511-100550511 on Build GRCh38
rs546873771 Chr.7: 100550756-100550756 on Build GRCh38
rs888021696 Chr.7: 100550623-100550623 on Build GRCh38
rs998773866 Chr.7: 100550549-100550549 on Build GRCh38
rs144744845 Chr.7: 100550400-100550400 on Build GRCh38
rs770694129 Chr.7: 100550543-100550546 on Build GRCh38
rs748556561 Chr.7: 100550475-100550475 on Build GRCh38
rs770552533 Chr.7: 100550507-100550507 on Build GRCh38
rs771970852 Chr.7: 100550519-100550519 on Build GRCh38
rs775536622 Chr.7: 100550529-100550529 on Build GRCh38
rs1030180554 Chr.7: 100550752-100550752 on Build GRCh38
rs762503806 Chr.7: 100550385-100550385 on Build GRCh38
rs781703349 Chr.7: 100550557-100550557 on Build GRCh38
rs750650620 Chr.7: 100550433-100550433 on Build GRCh38
rs187147239 Chr.7: 100550749-100550749 on Build GRCh38
rs781652659 Chr.7: 100550462-100550462 on Build GRCh38
rs528811835 Chr.7: 100550751-100550751 on Build GRCh38
rs767431926 Chr.7: 100550403-100550403 on Build GRCh38
rs753644967 Chr.7: 100550543-100550543 on Build GRCh38
rs755449202 Chr.7: 100550446-100550446 on Build GRCh38
rs367548306 Chr.7: 100550389-100550389 on Build GRCh38
rs954736035 Chr.7: 100550648-100550648 on Build GRCh38
rs181645141 Chr.7: 100550729-100550729 on Build GRCh38
rs556715227 Chr.7: 100550414-100550414 on Build GRCh38
rs866739243 Chr.7: 100550585-100550585 on Build GRCh38
rs1046670838 Chr.7: 100550794-100550794 on Build GRCh38
rs772823759 Chr.7: 100550370-100550370 on Build GRCh38
rs1039095586 Chr.7: 100550681-100550681 on Build GRCh38
rs966666193 Chr.7: 100550489-100550489 on Build GRCh38
rs575336072 Chr.7: 100550608-100550608 on Build GRCh38
rs747041982 Chr.7: 100550439-100550439 on Build GRCh38
rs375827427 Chr.7: 100550437-100550437 on Build GRCh38
rs755967504 Chr.7: 100550430-100550430 on Build GRCh38
rs146728315 Chr.7: 100550402-100550402 on Build GRCh38
rs746865620 Chr.7: 100550510-100550511 on Build GRCh38
rs776862088 Chr.7: 100550361-100550361 on Build GRCh38

View Full Product Details