Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100647117-100647609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL6B
Entrez Gene ID
51412
Gene Name
actin like 6B
Gene Aliases
ACTL6, BAF53B, arpNalpha
UniGene
Hs.259831
Gene Chromosome Location
Chr.7: 100643097-100656461 on Build GRCh38
Transcripts
NM_016188.4 NR_134539.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5886507 c.748T>G p.Y250D Chr.7: 100647455-100647455 on Build GRCh38
6044752 c.694_695insC p.N233fs*17 Chr.7: 100647508-100647509 on Build GRCh38
5576372 c.809C>T p.P270L Chr.7: 100647235-100647235 on Build GRCh38
3080620 c.760-2A>C p.? Chr.7: 100647286-100647286 on Build GRCh38
5589528 c.814G>A p.D272N Chr.7: 100647230-100647230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778374696 Chr.7: 100647316-100647316 on Build GRCh38
rs916689221 Chr.7: 100647345-100647345 on Build GRCh38
rs781115123 Chr.7: 100647542-100647542 on Build GRCh38
rs779550102 Chr.7: 100647507-100647508 on Build GRCh38
rs577709097 Chr.7: 100647297-100647297 on Build GRCh38
rs778056678 Chr.7: 100647486-100647486 on Build GRCh38
rs755552228 Chr.7: 100647491-100647492 on Build GRCh38
rs773511386 Chr.7: 100647396-100647396 on Build GRCh38
rs756801418 Chr.7: 100647311-100647311 on Build GRCh38
rs886981555 Chr.7: 100647521-100647521 on Build GRCh38
rs979636076 Chr.7: 100647355-100647355 on Build GRCh38
rs773244976 Chr.7: 100647257-100647257 on Build GRCh38
rs1039453059 Chr.7: 100647437-100647437 on Build GRCh38
rs771181055 Chr.7: 100647408-100647408 on Build GRCh38
rs369078607 Chr.7: 100647305-100647305 on Build GRCh38
rs752059926 Chr.7: 100647174-100647174 on Build GRCh38
rs567168998 Chr.7: 100647179-100647179 on Build GRCh38
rs376232920 Chr.7: 100647189-100647189 on Build GRCh38
rs182364275 Chr.7: 100647178-100647178 on Build GRCh38
rs774557186 Chr.7: 100647409-100647409 on Build GRCh38
rs780066798 Chr.7: 100647513-100647513 on Build GRCh38
rs560092975 Chr.7: 100647387-100647387 on Build GRCh38
rs556388048 Chr.7: 100647263-100647263 on Build GRCh38
rs778854802 Chr.7: 100647220-100647220 on Build GRCh38
rs2231175 Chr.7: 100647564-100647564 on Build GRCh38
rs756887243 Chr.7: 100647507-100647507 on Build GRCh38
rs200805270 Chr.7: 100647509-100647509 on Build GRCh38
rs767828491 Chr.7: 100647296-100647296 on Build GRCh38
rs574288531 Chr.7: 100647143-100647143 on Build GRCh38
rs779793080 Chr.7: 100647324-100647324 on Build GRCh38
rs760096533 Chr.7: 100647285-100647285 on Build GRCh38
rs745865581 Chr.7: 100647317-100647317 on Build GRCh38
rs901940947 Chr.7: 100647171-100647171 on Build GRCh38
rs777213917 Chr.7: 100647202-100647202 on Build GRCh38
rs1044348178 Chr.7: 100647138-100647138 on Build GRCh38
rs755777522 Chr.7: 100647195-100647195 on Build GRCh38
rs140422470 Chr.7: 100647504-100647504 on Build GRCh38
rs925490306 Chr.7: 100647357-100647357 on Build GRCh38
rs114099705 Chr.7: 100647415-100647415 on Build GRCh38
rs745775554 Chr.7: 100647231-100647231 on Build GRCh38
rs771364875 Chr.7: 100647568-100647568 on Build GRCh38
rs757123439 Chr.7: 100647274-100647275 on Build GRCh38
rs760188690 Chr.7: 100647412-100647412 on Build GRCh38
rs899598302 Chr.7: 100647438-100647438 on Build GRCh38
rs934960865 Chr.7: 100647149-100647149 on Build GRCh38
rs375678559 Chr.7: 100647298-100647298 on Build GRCh38
rs748834517 Chr.7: 100647205-100647205 on Build GRCh38
rs991826127 Chr.7: 100647334-100647334 on Build GRCh38
rs952513805 Chr.7: 100647358-100647358 on Build GRCh38
rs1053155437 Chr.7: 100647375-100647375 on Build GRCh38
rs1025032812 Chr.7: 100647341-100647341 on Build GRCh38
rs754846041 Chr.7: 100647538-100647538 on Build GRCh38
rs753088200 Chr.7: 100647416-100647416 on Build GRCh38
rs143726613 Chr.7: 100647571-100647571 on Build GRCh38
rs750120089 Chr.7: 100647409-100647410 on Build GRCh38
rs762011843 Chr.7: 100647248-100647248 on Build GRCh38
rs370335831 Chr.7: 100647282-100647282 on Build GRCh38
rs771906747 Chr.7: 100647235-100647235 on Build GRCh38
rs767026063 Chr.7: 100647394-100647394 on Build GRCh38
rs914744923 Chr.7: 100647345-100647357 on Build GRCh38
rs890839346 Chr.7: 100647176-100647176 on Build GRCh38
rs770379963 Chr.7: 100647206-100647206 on Build GRCh38
rs763233278 Chr.7: 100647403-100647403 on Build GRCh38
rs757785400 Chr.7: 100647510-100647510 on Build GRCh38
rs992192261 Chr.7: 100647344-100647344 on Build GRCh38
rs766523164 Chr.7: 100647261-100647261 on Build GRCh38
rs753449657 Chr.7: 100647309-100647309 on Build GRCh38
rs779400172 Chr.7: 100647351-100647351 on Build GRCh38
rs761132428 Chr.7: 100647427-100647427 on Build GRCh38
rs1042445712 Chr.7: 100647489-100647489 on Build GRCh38
rs545134426 Chr.7: 100647323-100647323 on Build GRCh38
rs764791421 Chr.7: 100647508-100647508 on Build GRCh38
rs375468195 Chr.7: 100647430-100647430 on Build GRCh38
rs748851049 Chr.7: 100647582-100647583 on Build GRCh38
rs749531229 Chr.7: 100647524-100647524 on Build GRCh38
rs935597426 Chr.7: 100647373-100647373 on Build GRCh38
rs371565196 Chr.7: 100647581-100647581 on Build GRCh38
rs771275027 Chr.7: 100647569-100647569 on Build GRCh38
rs751280377 Chr.7: 100647271-100647272 on Build GRCh38
rs147582494 Chr.7: 100647258-100647258 on Build GRCh38
rs768283279 Chr.7: 100647326-100647326 on Build GRCh38
rs1018069417 Chr.7: 100647200-100647200 on Build GRCh38
rs1052113184 Chr.7: 100647152-100647152 on Build GRCh38

View Full Product Details